Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 69957424-69957675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSN2
Entrez Gene ID
1447
Gene Name
casein beta
Gene Aliases
CASB
UniGene
Hs.2242
Gene Chromosome Location
Chr.4: 69955256-69965762 on Build GRCh38
Transcripts
NM_001302770.1 NM_001891.3 XM_017007760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6512297 c.463C>T p.Q155* Chr.4: 69957486-69957486 on Build GRCh38
2829990 c.366G>A p.T122T Chr.4: 69957583-69957583 on Build GRCh38
6545495 c.430C>A p.P144T Chr.4: 69957519-69957519 on Build GRCh38
2829991 c.347C>T p.P116L Chr.4: 69957602-69957602 on Build GRCh38
6563826 c.378_379insT p.D127fs*1 Chr.4: 69957570-69957571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773401690 Chr.4: 69957589-69957589 on Build GRCh38
rs140189537 Chr.4: 69957584-69957584 on Build GRCh38
rs555260643 Chr.4: 69957549-69957549 on Build GRCh38
rs777791012 Chr.4: 69957648-69957648 on Build GRCh38
rs766870827 Chr.4: 69957476-69957476 on Build GRCh38
rs1003669682 Chr.4: 69957567-69957567 on Build GRCh38
rs775132407 Chr.4: 69957609-69957609 on Build GRCh38
rs200065857 Chr.4: 69957585-69957585 on Build GRCh38
rs752426575 Chr.4: 69957492-69957492 on Build GRCh38
rs767933151 Chr.4: 69957469-69957469 on Build GRCh38
rs77091095 Chr.4: 69957550-69957550 on Build GRCh38
rs755271919 Chr.4: 69957531-69957531 on Build GRCh38
rs760870667 Chr.4: 69957543-69957543 on Build GRCh38
rs556908560 Chr.4: 69957639-69957639 on Build GRCh38
rs747186095 Chr.4: 69957592-69957592 on Build GRCh38
rs996665461 Chr.4: 69957564-69957564 on Build GRCh38
rs750732984 Chr.4: 69957470-69957470 on Build GRCh38
rs943018219 Chr.4: 69957457-69957457 on Build GRCh38
rs766922127 Chr.4: 69957548-69957548 on Build GRCh38
rs770935273 Chr.4: 69957447-69957447 on Build GRCh38
rs751438548 Chr.4: 69957575-69957575 on Build GRCh38
rs774606832 Chr.4: 69957456-69957456 on Build GRCh38
rs761889094 Chr.4: 69957460-69957460 on Build GRCh38
rs1037982412 Chr.4: 69957471-69957471 on Build GRCh38
rs891740577 Chr.4: 69957468-69957468 on Build GRCh38
rs1009057209 Chr.4: 69957597-69957597 on Build GRCh38
rs185772786 Chr.4: 69957583-69957583 on Build GRCh38
rs201368395 Chr.4: 69957650-69957650 on Build GRCh38
rs201164541 Chr.4: 69957602-69957602 on Build GRCh38
rs192835335 Chr.4: 69957449-69957449 on Build GRCh38
rs761231909 Chr.4: 69957473-69957473 on Build GRCh38
rs758431693 Chr.4: 69957644-69957644 on Build GRCh38
rs767421388 Chr.4: 69957621-69957621 on Build GRCh38
rs267600229 Chr.4: 69957614-69957614 on Build GRCh38
rs757093924 Chr.4: 69957576-69957576 on Build GRCh38
rs267600228 Chr.4: 69957578-69957578 on Build GRCh38
rs779449498 Chr.4: 69957454-69957455 on Build GRCh38
rs776474472 Chr.4: 69957611-69957611 on Build GRCh38
rs773636475 Chr.4: 69957534-69957534 on Build GRCh38
rs368977729 Chr.4: 69957503-69957503 on Build GRCh38
rs774252717 Chr.4: 69957525-69957525 on Build GRCh38
rs753388171 Chr.4: 69957477-69957477 on Build GRCh38
rs1015030365 Chr.4: 69957601-69957601 on Build GRCh38
rs764809165 Chr.4: 69957558-69957558 on Build GRCh38
rs754463985 Chr.4: 69957482-69957482 on Build GRCh38
rs190197314 Chr.4: 69957615-69957615 on Build GRCh38
rs765646805 Chr.4: 69957607-69957607 on Build GRCh38
rs768253575 Chr.4: 69957596-69957602 on Build GRCh38
rs890527571 Chr.4: 69957596-69957596 on Build GRCh38
rs200412722 Chr.4: 69957603-69957603 on Build GRCh38
rs141152689 Chr.4: 69957450-69957450 on Build GRCh38
rs745828002 Chr.4: 69957581-69957581 on Build GRCh38
rs952374561 Chr.4: 69957633-69957633 on Build GRCh38
rs746485707 Chr.4: 69957471-69957472 on Build GRCh38
rs374407737 Chr.4: 69957533-69957533 on Build GRCh38
rs757328792 Chr.4: 69957504-69957504 on Build GRCh38
rs376566334 Chr.4: 69957502-69957502 on Build GRCh38
rs779234858 Chr.4: 69957571-69957571 on Build GRCh38
rs749425664 Chr.4: 69957511-69957511 on Build GRCh38
rs778238023 Chr.4: 69957483-69957483 on Build GRCh38
rs752089071 Chr.4: 69957570-69957570 on Build GRCh38
rs771318281 Chr.4: 69957599-69957599 on Build GRCh38
rs781240580 Chr.4: 69957507-69957507 on Build GRCh38
rs752874310 Chr.4: 69957641-69957641 on Build GRCh38
rs375072668 Chr.4: 69957579-69957579 on Build GRCh38
rs760999263 Chr.4: 69957547-69957547 on Build GRCh38
rs192725460 Chr.4: 69957619-69957619 on Build GRCh38
rs755950545 Chr.4: 69957582-69957582 on Build GRCh38
rs756927150 Chr.4: 69957620-69957620 on Build GRCh38
rs899433396 Chr.4: 69957491-69957491 on Build GRCh38
rs768798471 Chr.4: 69957454-69957454 on Build GRCh38
rs750175387 Chr.4: 69957634-69957634 on Build GRCh38
rs757920475 Chr.4: 69957493-69957493 on Build GRCh38
rs768799924 Chr.4: 69957524-69957524 on Build GRCh38
rs1027087323 Chr.4: 69957628-69957628 on Build GRCh38

View Full Product Details