Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 195601-195828 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM20C
Entrez Gene ID
56975
Gene Name
family with sequence similarity 20 member C
Gene Aliases
DMP-4, DMP4, GEF-CK, RNS
UniGene
Hs.134742
Gene Chromosome Location
Chr.7: 192939-260774 on Build GRCh38
Transcripts
NM_020223.3 XM_017012450.1 XM_017012451.1 XM_017012452.1 XM_017012453.1 XM_017012454.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6659418 c.732G>A p.P244P Chr.7: 195680-195680 on Build GRCh38
3698327 c.715T>C p.Y239H Chr.7: 195663-195663 on Build GRCh38
4682900 c.661G>C p.D221H Chr.7: 195609-195609 on Build GRCh38
3637102 c.739G>A p.E247K Chr.7: 195687-195687 on Build GRCh38
4391172 c.759C>T p.L253L Chr.7: 195707-195707 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371079871 Chr.7: 195758-195758 on Build GRCh38
rs777009380 Chr.7: 195701-195701 on Build GRCh38
rs116181849 Chr.7: 195679-195679 on Build GRCh38
rs377584782 Chr.7: 195772-195772 on Build GRCh38
rs745637661 Chr.7: 195704-195704 on Build GRCh38
rs574668517 Chr.7: 195686-195686 on Build GRCh38
rs760967233 Chr.7: 195729-195729 on Build GRCh38
rs764407296 Chr.7: 195741-195741 on Build GRCh38
rs545268162 Chr.7: 195656-195656 on Build GRCh38
rs752896209 Chr.7: 195643-195643 on Build GRCh38
rs960130028 Chr.7: 195629-195629 on Build GRCh38
rs771123460 Chr.7: 195761-195761 on Build GRCh38
rs771316261 Chr.7: 195776-195776 on Build GRCh38
rs796051874 Chr.7: 195685-195685 on Build GRCh38
rs963699043 Chr.7: 195651-195651 on Build GRCh38
rs374098657 Chr.7: 195687-195687 on Build GRCh38
rs760049058 Chr.7: 195702-195702 on Build GRCh38
rs749188339 Chr.7: 195670-195670 on Build GRCh38
rs752800765 Chr.7: 195725-195725 on Build GRCh38
rs201720047 Chr.7: 195760-195760 on Build GRCh38
rs765294314 Chr.7: 195638-195638 on Build GRCh38
rs758547451 Chr.7: 195712-195712 on Build GRCh38
rs777006445 Chr.7: 195780-195780 on Build GRCh38
rs61730252 Chr.7: 195681-195681 on Build GRCh38
rs760011462 Chr.7: 195782-195782 on Build GRCh38
rs756187054 Chr.7: 195661-195661 on Build GRCh38
rs998015927 Chr.7: 195737-195737 on Build GRCh38
rs748161528 Chr.7: 195728-195728 on Build GRCh38
rs768052338 Chr.7: 195673-195673 on Build GRCh38
rs773497163 Chr.7: 195690-195690 on Build GRCh38
rs949577912 Chr.7: 195676-195676 on Build GRCh38
rs531086852 Chr.7: 195771-195771 on Build GRCh38
rs778330143 Chr.7: 195766-195766 on Build GRCh38
rs983478775 Chr.7: 195632-195632 on Build GRCh38
rs780125740 Chr.7: 195662-195662 on Build GRCh38
rs758646891 Chr.7: 195652-195652 on Build GRCh38
rs376874896 Chr.7: 195680-195680 on Build GRCh38
rs200225309 Chr.7: 195657-195657 on Build GRCh38
rs765256799 Chr.7: 195713-195713 on Build GRCh38
rs375588971 Chr.7: 195769-195769 on Build GRCh38
rs1016709355 Chr.7: 195637-195637 on Build GRCh38
rs1024044113 Chr.7: 195793-195793 on Build GRCh38
rs192542992 Chr.7: 195751-195751 on Build GRCh38
rs1014953797 Chr.7: 195645-195645 on Build GRCh38
rs61732569 Chr.7: 195622-195622 on Build GRCh38
rs904847120 Chr.7: 195720-195720 on Build GRCh38
rs553263336 Chr.7: 195765-195765 on Build GRCh38
rs971335232 Chr.7: 195805-195805 on Build GRCh38

View Full Product Details