Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186595592-186595865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT1
Entrez Gene ID
2195
Gene Name
FAT atypical cadherin 1
Gene Aliases
CDHF7, CDHR8, FAT, ME5, hFat1
UniGene
Hs.481371
Gene Chromosome Location
Chr.4: 186587783-186726915 on Build GRCh38
Transcripts
NM_005245.3 XM_005262834.2 XM_005262835.2 XM_006714139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3428401 c.13120G>A p.E4374K Chr.4: 186595707-186595707 on Build GRCh38
6407126 c.13098G>T p.Q4366H Chr.4: 186595729-186595729 on Build GRCh38
5892964 c.13051A>T p.K4351* Chr.4: 186595776-186595776 on Build GRCh38
6407125 c.13098G>T p.Q4366H Chr.4: 186595729-186595729 on Build GRCh38
6915334 c.13060C>T p.Q4354* Chr.4: 186595767-186595767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528665067 Chr.4: 186595648-186595648 on Build GRCh38
rs75306482 Chr.4: 186595616-186595616 on Build GRCh38
rs574164474 Chr.4: 186595621-186595621 on Build GRCh38
rs773679488 Chr.4: 186595704-186595704 on Build GRCh38
rs1027383845 Chr.4: 186595638-186595638 on Build GRCh38
rs759930842 Chr.4: 186595824-186595824 on Build GRCh38
rs775248043 Chr.4: 186595833-186595833 on Build GRCh38
rs752186670 Chr.4: 186595667-186595667 on Build GRCh38
rs761872072 Chr.4: 186595702-186595702 on Build GRCh38
rs2306988 Chr.4: 186595635-186595635 on Build GRCh38
rs781392214 Chr.4: 186595779-186595779 on Build GRCh38
rs776855582 Chr.4: 186595698-186595698 on Build GRCh38
rs771401093 Chr.4: 186595802-186595802 on Build GRCh38
rs765367010 Chr.4: 186595831-186595834 on Build GRCh38
rs371207320 Chr.4: 186595686-186595686 on Build GRCh38
rs370346177 Chr.4: 186595753-186595753 on Build GRCh38
rs769928509 Chr.4: 186595703-186595703 on Build GRCh38
rs4862721 Chr.4: 186595660-186595660 on Build GRCh38
rs894692817 Chr.4: 186595615-186595615 on Build GRCh38
rs753693029 Chr.4: 186595764-186595764 on Build GRCh38
rs766808676 Chr.4: 186595647-186595647 on Build GRCh38
rs762309111 Chr.4: 186595655-186595655 on Build GRCh38
rs749357346 Chr.4: 186595793-186595793 on Build GRCh38
rs80120846 Chr.4: 186595785-186595785 on Build GRCh38
rs755699033 Chr.4: 186595681-186595681 on Build GRCh38
rs1298865 Chr.4: 186595726-186595726 on Build GRCh38
rs778815135 Chr.4: 186595693-186595693 on Build GRCh38
rs143315416 Chr.4: 186595622-186595622 on Build GRCh38
rs777461585 Chr.4: 186595760-186595760 on Build GRCh38
rs777835734 Chr.4: 186595685-186595685 on Build GRCh38
rs910507309 Chr.4: 186595672-186595672 on Build GRCh38
rs1023998325 Chr.4: 186595769-186595769 on Build GRCh38
rs377350329 Chr.4: 186595751-186595751 on Build GRCh38
rs568172303 Chr.4: 186595661-186595661 on Build GRCh38
rs768057436 Chr.4: 186595825-186595825 on Build GRCh38
rs375073515 Chr.4: 186595773-186595773 on Build GRCh38
rs757166365 Chr.4: 186595770-186595770 on Build GRCh38
rs771915345 Chr.4: 186595774-186595774 on Build GRCh38
rs759822733 Chr.4: 186595719-186595719 on Build GRCh38
rs774732432 Chr.4: 186595805-186595805 on Build GRCh38
rs745683952 Chr.4: 186595696-186595696 on Build GRCh38
rs951826857 Chr.4: 186595819-186595819 on Build GRCh38
rs372275533 Chr.4: 186595838-186595838 on Build GRCh38
rs770597395 Chr.4: 186595691-186595691 on Build GRCh38
rs936159059 Chr.4: 186595709-186595709 on Build GRCh38
rs867833343 Chr.4: 186595699-186595699 on Build GRCh38
rs763565837 Chr.4: 186595707-186595707 on Build GRCh38
rs922222190 Chr.4: 186595834-186595834 on Build GRCh38
rs766819444 Chr.4: 186595712-186595712 on Build GRCh38
rs535865840 Chr.4: 186595787-186595787 on Build GRCh38
rs944616503 Chr.4: 186595673-186595673 on Build GRCh38
rs961851696 Chr.4: 186595623-186595623 on Build GRCh38
rs546255152 Chr.4: 186595789-186595789 on Build GRCh38

View Full Product Details