Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 14326401-14326628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLRT3
Entrez Gene ID
23767
Gene Name
fibronectin leucine rich transmembrane protein 3
Gene Aliases
HH21
UniGene
Hs.41296
Gene Chromosome Location
Chr.20: 14322985-14337847 on Build GRCh38
Transcripts
NM_013281.3 NM_198391.2 XM_005260682.4 XM_011529204.2 XM_011529205.2 NM_080676.5 XM_017027677.1

Gene Symbol
MACROD2
Entrez Gene ID
140733
Gene Name
MACRO domain containing 2
Gene Aliases
C20orf133
UniGene
Hs.661576
Gene Chromosome Location
Chr.20: 13995476-16053197 on Build GRCh38
Transcripts
NM_013281.3 NM_198391.2 XM_005260682.4 XM_011529204.2 XM_011529205.2 NM_080676.5 XM_017027677.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6665201 c.944G>A p.W315* Chr.20: 14326563-14326563 on Build GRCh38
281192 c.907C>T p.R303C Chr.20: 14326600-14326600 on Build GRCh38
6665200 c.933C>A p.C311* Chr.20: 14326574-14326574 on Build GRCh38
3910877 c.917C>T p.P306L Chr.20: 14326590-14326590 on Build GRCh38
1410456 c.1100C>A p.T367N Chr.20: 14326407-14326407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748518747 Chr.20: 14326544-14326544 on Build GRCh38
rs759473524 Chr.20: 14326428-14326428 on Build GRCh38
rs398124654 Chr.20: 14326491-14326491 on Build GRCh38
rs911759953 Chr.20: 14326477-14326477 on Build GRCh38
rs980632823 Chr.20: 14326474-14326474 on Build GRCh38
rs376427607 Chr.20: 14326436-14326436 on Build GRCh38
rs775343139 Chr.20: 14326579-14326579 on Build GRCh38
rs375651610 Chr.20: 14326555-14326555 on Build GRCh38
rs983233462 Chr.20: 14326476-14326476 on Build GRCh38
rs944637009 Chr.20: 14326485-14326485 on Build GRCh38
rs762893918 Chr.20: 14326580-14326580 on Build GRCh38
rs769769921 Chr.20: 14326561-14326561 on Build GRCh38
rs763048882 Chr.20: 14326558-14326558 on Build GRCh38
rs369748001 Chr.20: 14326599-14326599 on Build GRCh38
rs763883408 Chr.20: 14326456-14326456 on Build GRCh38
rs756352350 Chr.20: 14326489-14326489 on Build GRCh38
rs768394306 Chr.20: 14326585-14326585 on Build GRCh38
rs6033944 Chr.20: 14326469-14326469 on Build GRCh38
rs936019282 Chr.20: 14326577-14326577 on Build GRCh38
rs372667293 Chr.20: 14326454-14326454 on Build GRCh38
rs779346527 Chr.20: 14326524-14326524 on Build GRCh38
rs776024502 Chr.20: 14326426-14326426 on Build GRCh38
rs774163669 Chr.20: 14326589-14326589 on Build GRCh38
rs775345656 Chr.20: 14326437-14326437 on Build GRCh38
rs753905921 Chr.20: 14326515-14326515 on Build GRCh38
rs562745017 Chr.20: 14326459-14326459 on Build GRCh38
rs141238925 Chr.20: 14326559-14326559 on Build GRCh38
rs541810428 Chr.20: 14326523-14326523 on Build GRCh38
rs755488912 Chr.20: 14326518-14326518 on Build GRCh38
rs148251977 Chr.20: 14326557-14326557 on Build GRCh38
rs750588557 Chr.20: 14326457-14326457 on Build GRCh38
rs758767646 Chr.20: 14326553-14326553 on Build GRCh38
rs766443058 Chr.20: 14326511-14326511 on Build GRCh38
rs144067037 Chr.20: 14326461-14326461 on Build GRCh38

View Full Product Details