Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70480891-70481150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG4
Entrez Gene ID
25839
Gene Name
component of oligomeric golgi complex 4
Gene Aliases
CDG2J, COD1
UniGene
Hs.208680
Gene Chromosome Location
Chr.16: 70480569-70523554 on Build GRCh38
Transcripts
NM_001195139.1 NM_015386.2 XM_011522981.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
973020 c.2308C>T p.R770C Chr.16: 70481072-70481072 on Build GRCh38
4062367 c.2363G>A p.R788H Chr.16: 70481017-70481017 on Build GRCh38
239387 c.2242G>A p.E748K Chr.16: 70481138-70481138 on Build GRCh38
6632226 c.2271C>T p.S757S Chr.16: 70481109-70481109 on Build GRCh38
4579225 c.2246T>G p.I749S Chr.16: 70481134-70481134 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752445461 Chr.16: 70481046-70481046 on Build GRCh38
rs1006772304 Chr.16: 70480962-70480962 on Build GRCh38
rs747527122 Chr.16: 70481097-70481097 on Build GRCh38
rs1052373781 Chr.16: 70480926-70480926 on Build GRCh38
rs188102141 Chr.16: 70480938-70480938 on Build GRCh38
rs776332724 Chr.16: 70481036-70481036 on Build GRCh38
rs113010015 Chr.16: 70481102-70481102 on Build GRCh38
rs1035186212 Chr.16: 70480951-70480951 on Build GRCh38
rs765871294 Chr.16: 70481032-70481032 on Build GRCh38
rs749494856 Chr.16: 70481081-70481081 on Build GRCh38
rs887314289 Chr.16: 70480955-70480955 on Build GRCh38
rs387907203 Chr.16: 70481062-70481062 on Build GRCh38
rs144846241 Chr.16: 70481084-70481084 on Build GRCh38
rs751066284 Chr.16: 70481018-70481018 on Build GRCh38
rs905277603 Chr.16: 70480943-70480943 on Build GRCh38
rs780050636 Chr.16: 70481078-70481078 on Build GRCh38
rs935189450 Chr.16: 70481054-70481054 on Build GRCh38
rs372160853 Chr.16: 70481114-70481114 on Build GRCh38
rs200293003 Chr.16: 70481053-70481053 on Build GRCh38
rs759073858 Chr.16: 70481103-70481103 on Build GRCh38
rs376925549 Chr.16: 70481098-70481098 on Build GRCh38
rs368014374 Chr.16: 70481110-70481110 on Build GRCh38
rs200485721 Chr.16: 70481060-70481060 on Build GRCh38
rs987662091 Chr.16: 70481037-70481037 on Build GRCh38
rs771106863 Chr.16: 70481105-70481105 on Build GRCh38
rs751496426 Chr.16: 70481115-70481115 on Build GRCh38
rs533161794 Chr.16: 70481070-70481070 on Build GRCh38
rs201867211 Chr.16: 70480989-70480989 on Build GRCh38
rs192714339 Chr.16: 70481003-70481003 on Build GRCh38
rs758368956 Chr.16: 70480977-70480977 on Build GRCh38
rs201107005 Chr.16: 70480949-70480949 on Build GRCh38
rs760163512 Chr.16: 70481022-70481022 on Build GRCh38
rs769770565 Chr.16: 70480992-70480992 on Build GRCh38
rs1025304275 Chr.16: 70481007-70481007 on Build GRCh38
rs780321299 Chr.16: 70480993-70480993 on Build GRCh38
rs772659272 Chr.16: 70481013-70481013 on Build GRCh38
rs142971847 Chr.16: 70481017-70481017 on Build GRCh38
rs377725094 Chr.16: 70481002-70481002 on Build GRCh38
rs893798618 Chr.16: 70480928-70480928 on Build GRCh38
rs543968780 Chr.16: 70481092-70481092 on Build GRCh38
rs183503243 Chr.16: 70480934-70480934 on Build GRCh38
rs759235205 Chr.16: 70481041-70481041 on Build GRCh38
rs773762934 Chr.16: 70481093-70481093 on Build GRCh38
rs911765572 Chr.16: 70480917-70480917 on Build GRCh38
rs749535137 Chr.16: 70480996-70480996 on Build GRCh38
rs745909388 Chr.16: 70480990-70480990 on Build GRCh38
rs7919 Chr.16: 70480925-70480925 on Build GRCh38
rs938661433 Chr.16: 70481130-70481130 on Build GRCh38
rs368766795 Chr.16: 70480963-70480963 on Build GRCh38
rs535935157 Chr.16: 70480972-70480972 on Build GRCh38
rs184431716 Chr.16: 70481087-70481087 on Build GRCh38
rs370178627 Chr.16: 70481108-70481108 on Build GRCh38
rs199688575 Chr.16: 70481050-70481050 on Build GRCh38
rs200083914 Chr.16: 70481129-70481129 on Build GRCh38
rs202087905 Chr.16: 70481073-70481073 on Build GRCh38
rs540169455 Chr.16: 70481010-70481010 on Build GRCh38
rs764982187 Chr.16: 70481042-70481042 on Build GRCh38
rs140873482 Chr.16: 70481066-70481066 on Build GRCh38
rs150695410 Chr.16: 70481057-70481057 on Build GRCh38
rs200772778 Chr.16: 70481109-70481109 on Build GRCh38
rs1056975658 Chr.16: 70481112-70481112 on Build GRCh38

View Full Product Details