Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 104304805-104305034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EID3
Entrez Gene ID
493861
Gene Name
EP300 interacting inhibitor of differentiation 3
Gene Aliases
NS4EB, NSE4B, NSMCE4B
UniGene
Hs.659857
Gene Chromosome Location
Chr.12: 104303732-104305205 on Build GRCh38
Transcripts
NM_001008394.2 NM_001093771.2 NM_001261445.1 NM_001261446.1 NM_003330.3 NM_182729.2 NM_182742.2 NM_182743.2

Gene Symbol
TXNRD1
Entrez Gene ID
7296
Gene Name
thioredoxin reductase 1
Gene Aliases
GRIM-12, TR, TR1, TRXR1, TXNR
UniGene
Hs.654922
Gene Chromosome Location
Chr.12: 104215779-104350307 on Build GRCh38
Transcripts
NM_001008394.2 NM_001093771.2 NM_001261445.1 NM_001261446.1 NM_003330.3 NM_182729.2 NM_182742.2 NM_182743.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5699725 c.875G>T p.G292V Chr.12: 104304809-104304809 on Build GRCh38
6232389 c.995C>T p.S332L Chr.12: 104304929-104304929 on Build GRCh38
6231985 c.986C>T p.T329I Chr.12: 104304920-104304920 on Build GRCh38
3416413 c.881G>A p.G294E Chr.12: 104304815-104304815 on Build GRCh38
6234506 c.886G>T p.D296Y Chr.12: 104304820-104304820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs894932620 Chr.12: 104304890-104304890 on Build GRCh38
rs779817123 Chr.12: 104304849-104304850 on Build GRCh38
rs1032608365 Chr.12: 104304950-104304951 on Build GRCh38
rs377091329 Chr.12: 104304876-104304876 on Build GRCh38
rs746261186 Chr.12: 104304906-104304906 on Build GRCh38
rs775806781 Chr.12: 104304921-104304921 on Build GRCh38
rs761125074 Chr.12: 104304927-104304927 on Build GRCh38
rs746894135 Chr.12: 104304865-104304865 on Build GRCh38
rs749343384 Chr.12: 104304984-104304984 on Build GRCh38
rs1018857764 Chr.12: 104304967-104304967 on Build GRCh38
rs965469081 Chr.12: 104304988-104304988 on Build GRCh38
rs761328553 Chr.12: 104304956-104304956 on Build GRCh38
rs751372684 Chr.12: 104304971-104304971 on Build GRCh38
rs999167792 Chr.12: 104304944-104304944 on Build GRCh38
rs35757377 Chr.12: 104304897-104304898 on Build GRCh38
rs754921010 Chr.12: 104304870-104304870 on Build GRCh38
rs749385426 Chr.12: 104304874-104304874 on Build GRCh38
rs200258450 Chr.12: 104304951-104304952 on Build GRCh38
rs772369124 Chr.12: 104304913-104304913 on Build GRCh38
rs751171911 Chr.12: 104304924-104304926 on Build GRCh38
rs997225109 Chr.12: 104304886-104304886 on Build GRCh38
rs777759118 Chr.12: 104304871-104304871 on Build GRCh38
rs976792979 Chr.12: 104304991-104304991 on Build GRCh38
rs774486177 Chr.12: 104304899-104304899 on Build GRCh38
rs371069815 Chr.12: 104304900-104304900 on Build GRCh38
rs372620381 Chr.12: 104304856-104304856 on Build GRCh38
rs909100860 Chr.12: 104304860-104304860 on Build GRCh38
rs780827201 Chr.12: 104304965-104304966 on Build GRCh38
rs764802723 Chr.12: 104304957-104304957 on Build GRCh38
rs757317328 Chr.12: 104304986-104304986 on Build GRCh38
rs766326037 Chr.12: 104304965-104304965 on Build GRCh38
rs529973442 Chr.12: 104304929-104304929 on Build GRCh38
rs776061265 Chr.12: 104304930-104304930 on Build GRCh38
rs754560070 Chr.12: 104304947-104304948 on Build GRCh38
rs747613307 Chr.12: 104304966-104304966 on Build GRCh38
rs781178230 Chr.12: 104304973-104304973 on Build GRCh38
rs779022548 Chr.12: 104304880-104304880 on Build GRCh38
rs758290002 Chr.12: 104304961-104304961 on Build GRCh38

View Full Product Details