Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 86579644-86579891 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXL1
Entrez Gene ID
2300
Gene Name
forkhead box L1
Gene Aliases
FKH6, FKHL11, FREAC7
UniGene
Hs.533830
Gene Chromosome Location
Chr.16: 86578509-86581698 on Build GRCh38
Transcripts
NM_005250.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7075979 c.924C>G p.F308L Chr.16: 86579647-86579647 on Build GRCh38
4685411 c.959G>A p.G320D Chr.16: 86579682-86579682 on Build GRCh38
4456524 c.1012C>T p.P338S Chr.16: 86579735-86579735 on Build GRCh38
7202898 c.934C>A p.L312M Chr.16: 86579657-86579657 on Build GRCh38
4646529 c.929C>T p.A310V Chr.16: 86579652-86579652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199660959 Chr.16: 86579765-86579765 on Build GRCh38
rs1019884415 Chr.16: 86579767-86579767 on Build GRCh38
rs180865818 Chr.16: 86579801-86579801 on Build GRCh38
rs781034400 Chr.16: 86579743-86579743 on Build GRCh38
rs374156925 Chr.16: 86579807-86579807 on Build GRCh38
rs928798404 Chr.16: 86579834-86579834 on Build GRCh38
rs759793722 Chr.16: 86579711-86579711 on Build GRCh38
rs754156142 Chr.16: 86579700-86579700 on Build GRCh38
rs747707854 Chr.16: 86579813-86579813 on Build GRCh38
rs539599013 Chr.16: 86579741-86579741 on Build GRCh38
rs963425937 Chr.16: 86579728-86579728 on Build GRCh38
rs757203142 Chr.16: 86579796-86579796 on Build GRCh38
rs376492717 Chr.16: 86579785-86579785 on Build GRCh38
rs778388787 Chr.16: 86579811-86579811 on Build GRCh38
rs376251742 Chr.16: 86579809-86579809 on Build GRCh38
rs1053364634 Chr.16: 86579867-86579867 on Build GRCh38
rs541981355 Chr.16: 86579769-86579769 on Build GRCh38
rs745706511 Chr.16: 86579745-86579745 on Build GRCh38
rs987378998 Chr.16: 86579770-86579770 on Build GRCh38
rs934851128 Chr.16: 86579861-86579861 on Build GRCh38
rs752797610 Chr.16: 86579715-86579715 on Build GRCh38
rs981597554 Chr.16: 86579833-86579833 on Build GRCh38
rs564421915 Chr.16: 86579814-86579814 on Build GRCh38
rs371495333 Chr.16: 86579667-86579667 on Build GRCh38
rs751757798 Chr.16: 86579794-86579794 on Build GRCh38
rs573254555 Chr.16: 86579768-86579768 on Build GRCh38
rs1052740188 Chr.16: 86579725-86579725 on Build GRCh38
rs777034884 Chr.16: 86579777-86579777 on Build GRCh38
rs772537317 Chr.16: 86579695-86579695 on Build GRCh38
rs201605395 Chr.16: 86579778-86579778 on Build GRCh38
rs376837550 Chr.16: 86579699-86579699 on Build GRCh38
rs778077859 Chr.16: 86579750-86579750 on Build GRCh38
rs773665323 Chr.16: 86579698-86579698 on Build GRCh38
rs767402138 Chr.16: 86579797-86579797 on Build GRCh38
rs769464450 Chr.16: 86579755-86579755 on Build GRCh38
rs914848765 Chr.16: 86579871-86579871 on Build GRCh38
rs758392330 Chr.16: 86579665-86579665 on Build GRCh38
rs765429672 Chr.16: 86579713-86579713 on Build GRCh38
rs548926439 Chr.16: 86579673-86579673 on Build GRCh38
rs1011287442 Chr.16: 86579742-86579742 on Build GRCh38
rs764026385 Chr.16: 86579696-86579710 on Build GRCh38
rs895957157 Chr.16: 86579853-86579853 on Build GRCh38
rs1049471503 Chr.16: 86579747-86579747 on Build GRCh38
rs370452719 Chr.16: 86579786-86579786 on Build GRCh38
rs185528186 Chr.16: 86579677-86579677 on Build GRCh38
rs138392858 Chr.16: 86579722-86579722 on Build GRCh38
rs143767765 Chr.16: 86579686-86579686 on Build GRCh38
rs773861513 Chr.16: 86579775-86579775 on Build GRCh38
rs780864241 Chr.16: 86579670-86579670 on Build GRCh38
rs919070291 Chr.16: 86579756-86579756 on Build GRCh38
rs116490033 Chr.16: 86579830-86579830 on Build GRCh38
rs907430078 Chr.16: 86579719-86579719 on Build GRCh38
rs892801101 Chr.16: 86579736-86579736 on Build GRCh38
rs904612236 Chr.16: 86579788-86579788 on Build GRCh38
rs376500241 Chr.16: 86579668-86579668 on Build GRCh38
rs200228439 Chr.16: 86579776-86579776 on Build GRCh38
rs778510104 Chr.16: 86579808-86579808 on Build GRCh38
rs56324750 Chr.16: 86579818-86579818 on Build GRCh38
rs200555965 Chr.16: 86579718-86579718 on Build GRCh38

View Full Product Details