Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 40042452-40042705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCAM
Entrez Gene ID
1826
Gene Name
DS cell adhesion molecule
Gene Aliases
CHD2, CHD2-42, CHD2-52
UniGene
Hs.397800
Gene Chromosome Location
Chr.21: 40010999-40847113 on Build GRCh38
Transcripts
NM_001271534.1 NM_001389.3 NR_073202.1 XM_017028281.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6350318 c.5476C>A p.Q1826K Chr.21: 40042581-40042581 on Build GRCh38
725136 c.5400C>A p.S1800R Chr.21: 40042657-40042657 on Build GRCh38
6494714 c.5506G>T p.E1836* Chr.21: 40042551-40042551 on Build GRCh38
3550786 c.5470G>A p.E1824K Chr.21: 40042587-40042587 on Build GRCh38
4101646 c.5469G>C p.M1823I Chr.21: 40042588-40042588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200410460 Chr.21: 40042493-40042493 on Build GRCh38
rs771726952 Chr.21: 40042480-40042480 on Build GRCh38
rs771399250 Chr.21: 40042504-40042504 on Build GRCh38
rs958167486 Chr.21: 40042552-40042552 on Build GRCh38
rs371190781 Chr.21: 40042531-40042531 on Build GRCh38
rs750278142 Chr.21: 40042557-40042557 on Build GRCh38
rs758281162 Chr.21: 40042671-40042671 on Build GRCh38
rs189404834 Chr.21: 40042492-40042492 on Build GRCh38
rs199659065 Chr.21: 40042525-40042525 on Build GRCh38
rs375850828 Chr.21: 40042491-40042491 on Build GRCh38
rs199512904 Chr.21: 40042603-40042603 on Build GRCh38
rs776587730 Chr.21: 40042648-40042648 on Build GRCh38
rs775951471 Chr.21: 40042615-40042615 on Build GRCh38
rs114741050 Chr.21: 40042597-40042597 on Build GRCh38
rs772544835 Chr.21: 40042614-40042614 on Build GRCh38
rs145903838 Chr.21: 40042666-40042666 on Build GRCh38
rs775828668 Chr.21: 40042526-40042526 on Build GRCh38
rs759961978 Chr.21: 40042513-40042513 on Build GRCh38
rs757148200 Chr.21: 40042553-40042553 on Build GRCh38
rs763398671 Chr.21: 40042495-40042495 on Build GRCh38
rs754617380 Chr.21: 40042570-40042570 on Build GRCh38
rs991212273 Chr.21: 40042667-40042667 on Build GRCh38
rs1020734882 Chr.21: 40042591-40042591 on Build GRCh38
rs747632409 Chr.21: 40042577-40042577 on Build GRCh38
rs746684524 Chr.21: 40042569-40042569 on Build GRCh38
rs200764944 Chr.21: 40042663-40042663 on Build GRCh38
rs375445425 Chr.21: 40042558-40042558 on Build GRCh38
rs765163525 Chr.21: 40042554-40042554 on Build GRCh38
rs779675403 Chr.21: 40042489-40042489 on Build GRCh38
rs181977833 Chr.21: 40042669-40042669 on Build GRCh38
rs771396349 Chr.21: 40042582-40042582 on Build GRCh38
rs375289717 Chr.21: 40042624-40042624 on Build GRCh38
rs766191893 Chr.21: 40042679-40042679 on Build GRCh38
rs753915774 Chr.21: 40042541-40042541 on Build GRCh38
rs774865490 Chr.21: 40042507-40042507 on Build GRCh38
rs779730383 Chr.21: 40042568-40042568 on Build GRCh38
rs764312075 Chr.21: 40042637-40042637 on Build GRCh38

View Full Product Details