Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218399880-218400129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSP1
Entrez Gene ID
58190
Gene Name
CTD small phosphatase 1
Gene Aliases
NIF3, NLI-IF, NLIIF, SCP1
UniGene
Hs.444468
Gene Chromosome Location
Chr.2: 218398338-218405941 on Build GRCh38
Transcripts
NM_001206878.1 NM_021198.2 NM_182642.2 XM_017004617.1 XM_017004618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382065 c.6C>A p.D2E Chr.2: 218400096-218400096 on Build GRCh38
1614359 c.17_18insCA p.I7fs*104 Chr.2: 218400107-218400108 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs942020089 Chr.2: 218400038-218400038 on Build GRCh38
rs1037617948 Chr.2: 218399992-218399992 on Build GRCh38
rs976581068 Chr.2: 218399960-218399960 on Build GRCh38
rs1012874192 Chr.2: 218399930-218399930 on Build GRCh38
rs932483188 Chr.2: 218399989-218399992 on Build GRCh38
rs895399532 Chr.2: 218399925-218399925 on Build GRCh38
rs1022553157 Chr.2: 218399937-218399937 on Build GRCh38
rs957662181 Chr.2: 218399913-218399913 on Build GRCh38
rs922403988 Chr.2: 218399955-218399955 on Build GRCh38
rs909819301 Chr.2: 218399994-218399994 on Build GRCh38
rs757010032 Chr.2: 218400096-218400096 on Build GRCh38
rs533705122 Chr.2: 218400004-218400004 on Build GRCh38
rs1056476748 Chr.2: 218400069-218400069 on Build GRCh38
rs955801892 Chr.2: 218399959-218399959 on Build GRCh38
rs954785777 Chr.2: 218400033-218400033 on Build GRCh38
rs1010863052 Chr.2: 218399917-218399917 on Build GRCh38
rs1021281848 Chr.2: 218399932-218399932 on Build GRCh38
rs777751173 Chr.2: 218400075-218400075 on Build GRCh38
rs976972132 Chr.2: 218399940-218399940 on Build GRCh38
rs549174201 Chr.2: 218400034-218400034 on Build GRCh38
rs780862243 Chr.2: 218400105-218400105 on Build GRCh38
rs939475911 Chr.2: 218400063-218400063 on Build GRCh38
rs910598314 Chr.2: 218400035-218400035 on Build GRCh38
rs966596518 Chr.2: 218399938-218399938 on Build GRCh38
rs922392329 Chr.2: 218399981-218399981 on Build GRCh38
rs566992143 Chr.2: 218400090-218400090 on Build GRCh38
rs941341241 Chr.2: 218400031-218400031 on Build GRCh38
rs566808221 Chr.2: 218399993-218399993 on Build GRCh38
rs551378353 Chr.2: 218399950-218399950 on Build GRCh38
rs1037987118 Chr.2: 218400098-218400098 on Build GRCh38
rs918664389 Chr.2: 218400051-218400051 on Build GRCh38

View Full Product Details