Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 60767698-60767969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL7
Entrez Gene ID
79993
Gene Name
ELOVL fatty acid elongase 7
Gene Aliases
-
UniGene
Hs.274256
Gene Chromosome Location
Chr.5: 60751789-60844292 on Build GRCh38
Transcripts
NM_001104558.1 NM_001297617.1 NM_001297618.1 NM_024930.2 XM_005248606.4 XM_005248607.3 XM_006714695.3 XM_011543651.2 XM_017009885.1 XM_017009886.1 XM_017009887.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855790 c.279A>G p.I93M Chr.5: 60767880-60767880 on Build GRCh38
5038430 c.276T>G p.G92G Chr.5: 60767883-60767883 on Build GRCh38
391226 c.277delA p.I93fs*1 Chr.5: 60767882-60767882 on Build GRCh38
3617318 c.335G>A p.R112K Chr.5: 60767824-60767824 on Build GRCh38
5061137 c.316C>T p.R106W Chr.5: 60767843-60767843 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs553735764 Chr.5: 60767940-60767940 on Build GRCh38
rs201806045 Chr.5: 60767919-60767919 on Build GRCh38
rs754915095 Chr.5: 60767947-60767947 on Build GRCh38
rs777724115 Chr.5: 60767924-60767924 on Build GRCh38
rs754148974 Chr.5: 60767782-60767782 on Build GRCh38
rs749088345 Chr.5: 60767843-60767843 on Build GRCh38
rs748301033 Chr.5: 60767863-60767863 on Build GRCh38
rs999026197 Chr.5: 60767755-60767755 on Build GRCh38
rs777088532 Chr.5: 60767934-60767934 on Build GRCh38
rs756057238 Chr.5: 60767841-60767841 on Build GRCh38
rs185682294 Chr.5: 60767921-60767921 on Build GRCh38
rs761104177 Chr.5: 60767879-60767879 on Build GRCh38
rs373667081 Chr.5: 60767785-60767785 on Build GRCh38
rs765402621 Chr.5: 60767919-60767925 on Build GRCh38
rs1033206278 Chr.5: 60767906-60767906 on Build GRCh38
rs772283847 Chr.5: 60767922-60767922 on Build GRCh38
rs759883343 Chr.5: 60767888-60767888 on Build GRCh38
rs761924036 Chr.5: 60767809-60767809 on Build GRCh38
rs763772946 Chr.5: 60767894-60767894 on Build GRCh38
rs866433834 Chr.5: 60767866-60767866 on Build GRCh38
rs753859584 Chr.5: 60767946-60767946 on Build GRCh38
rs371208705 Chr.5: 60767812-60767812 on Build GRCh38
rs747090572 Chr.5: 60767926-60767926 on Build GRCh38
rs771365279 Chr.5: 60767880-60767880 on Build GRCh38
rs773295195 Chr.5: 60767870-60767870 on Build GRCh38
rs867391130 Chr.5: 60767910-60767910 on Build GRCh38
rs766756450 Chr.5: 60767777-60767777 on Build GRCh38
rs779257014 Chr.5: 60767858-60767858 on Build GRCh38
rs772782089 Chr.5: 60767800-60767800 on Build GRCh38
rs771489747 Chr.5: 60767933-60767933 on Build GRCh38
rs756688463 Chr.5: 60767802-60767802 on Build GRCh38
rs758054064 Chr.5: 60767784-60767784 on Build GRCh38
rs750435449 Chr.5: 60767944-60767945 on Build GRCh38
rs777049585 Chr.5: 60767884-60767884 on Build GRCh38
rs375506846 Chr.5: 60767819-60767819 on Build GRCh38
rs112505252 Chr.5: 60767913-60767913 on Build GRCh38
rs374576689 Chr.5: 60767912-60767912 on Build GRCh38
rs200699082 Chr.5: 60767867-60767867 on Build GRCh38
rs573554345 Chr.5: 60767944-60767944 on Build GRCh38
rs1460958 Chr.5: 60767749-60767749 on Build GRCh38
rs770161587 Chr.5: 60767938-60767938 on Build GRCh38
rs766460854 Chr.5: 60767945-60767945 on Build GRCh38
rs761376176 Chr.5: 60767939-60767939 on Build GRCh38
rs751200482 Chr.5: 60767895-60767895 on Build GRCh38
rs753736432 Chr.5: 60767915-60767915 on Build GRCh38
rs779998858 Chr.5: 60767842-60767842 on Build GRCh38
rs902911649 Chr.5: 60767754-60767754 on Build GRCh38
rs79940333 Chr.5: 60767936-60767936 on Build GRCh38
rs199575400 Chr.5: 60767900-60767900 on Build GRCh38
rs371579989 Chr.5: 60767898-60767898 on Build GRCh38
rs201538282 Chr.5: 60767801-60767801 on Build GRCh38
rs761475311 Chr.5: 60767897-60767897 on Build GRCh38
rs142412675 Chr.5: 60767856-60767856 on Build GRCh38

View Full Product Details