Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 746074-746335 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN4
Entrez Gene ID
50628
Gene Name
gem nuclear organelle associated protein 4
Gene Aliases
HC56, HCAP1, HHRF-1, p97
UniGene
Hs.499620
Gene Chromosome Location
Chr.17: 744421-752868 on Build GRCh38
Transcripts
NM_015721.2 XM_005256667.4 XM_005256670.4 XM_011523910.2 XM_011523911.2 XM_011523912.2 XM_011523913.2 XM_017024709.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4473856 c.1879C>T p.L627F Chr.17: 746164-746164 on Build GRCh38
1385279 c.1778A>T p.E593V Chr.17: 746265-746265 on Build GRCh38
6670426 c.1919T>C p.I640T Chr.17: 746124-746124 on Build GRCh38
1610644 c.1796C>G p.S599C Chr.17: 746247-746247 on Build GRCh38
3672538 c.1854C>A p.P618P Chr.17: 746189-746189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199702345 Chr.17: 746272-746272 on Build GRCh38
rs562143356 Chr.17: 746139-746139 on Build GRCh38
rs762712933 Chr.17: 746233-746233 on Build GRCh38
rs776242615 Chr.17: 746244-746244 on Build GRCh38
rs747763804 Chr.17: 746293-746293 on Build GRCh38
rs533913356 Chr.17: 746140-746140 on Build GRCh38
rs762736188 Chr.17: 746254-746254 on Build GRCh38
rs377606651 Chr.17: 746095-746095 on Build GRCh38
rs143008523 Chr.17: 746304-746304 on Build GRCh38
rs371459371 Chr.17: 746186-746186 on Build GRCh38
rs764724631 Chr.17: 746110-746110 on Build GRCh38
rs764982624 Chr.17: 746153-746153 on Build GRCh38
rs778323201 Chr.17: 746124-746124 on Build GRCh38
rs756630533 Chr.17: 746276-746276 on Build GRCh38
rs761785821 Chr.17: 746152-746152 on Build GRCh38
rs772928799 Chr.17: 746253-746253 on Build GRCh38
rs754463282 Chr.17: 746287-746287 on Build GRCh38
rs376416115 Chr.17: 746316-746316 on Build GRCh38
rs776310179 Chr.17: 746105-746107 on Build GRCh38
rs200492563 Chr.17: 746141-746141 on Build GRCh38
rs757544507 Chr.17: 746295-746295 on Build GRCh38
rs756721979 Chr.17: 746210-746210 on Build GRCh38
rs374700498 Chr.17: 746142-746142 on Build GRCh38
rs373700715 Chr.17: 746193-746193 on Build GRCh38
rs759844591 Chr.17: 746266-746266 on Build GRCh38
rs770262994 Chr.17: 746112-746112 on Build GRCh38
rs1009180395 Chr.17: 746289-746289 on Build GRCh38
rs746728929 Chr.17: 746236-746236 on Build GRCh38
rs750243511 Chr.17: 746155-746155 on Build GRCh38
rs748035562 Chr.17: 746245-746245 on Build GRCh38
rs372394762 Chr.17: 746299-746299 on Build GRCh38
rs779567946 Chr.17: 746222-746222 on Build GRCh38
rs758056773 Chr.17: 746213-746213 on Build GRCh38
rs200443736 Chr.17: 746161-746161 on Build GRCh38
rs370625526 Chr.17: 746096-746096 on Build GRCh38
rs764701907 Chr.17: 746281-746281 on Build GRCh38
rs1008625124 Chr.17: 746275-746275 on Build GRCh38
rs749939964 Chr.17: 746205-746205 on Build GRCh38
rs368565942 Chr.17: 746156-746156 on Build GRCh38
rs898791411 Chr.17: 746241-746241 on Build GRCh38
rs768025861 Chr.17: 746239-746239 on Build GRCh38
rs576980675 Chr.17: 746163-746163 on Build GRCh38
rs766254914 Chr.17: 746256-746256 on Build GRCh38
rs4968104 Chr.17: 746265-746265 on Build GRCh38
rs374674049 Chr.17: 746103-746103 on Build GRCh38
rs369912401 Chr.17: 746310-746310 on Build GRCh38
rs375214416 Chr.17: 746126-746126 on Build GRCh38
rs754999536 Chr.17: 746181-746181 on Build GRCh38
rs1046242682 Chr.17: 746191-746191 on Build GRCh38
rs769363855 Chr.17: 746315-746315 on Build GRCh38
rs763106997 Chr.17: 746160-746160 on Build GRCh38
rs916642992 Chr.17: 746173-746173 on Build GRCh38
rs910925 Chr.17: 746307-746307 on Build GRCh38
rs368058625 Chr.17: 746212-746212 on Build GRCh38

View Full Product Details