Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41091743-41091970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2A13
Entrez Gene ID
1553
Gene Name
cytochrome P450 family 2 subfamily A member 13
Gene Aliases
CPAD, CYP2A, CYPIIA13
UniGene
Hs.567252
Gene Chromosome Location
Chr.19: 41088451-41096195 on Build GRCh38
Transcripts
NM_000766.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1612291 c.724C>A p.Q242K Chr.19: 41091801-41091801 on Build GRCh38
4078410 c.741C>A p.F247L Chr.19: 41091818-41091818 on Build GRCh38
6150996 c.741C>T p.F247F Chr.19: 41091818-41091818 on Build GRCh38
5506533 c.805G>A p.D269N Chr.19: 41091882-41091882 on Build GRCh38
290363 c.810C>G p.S270S Chr.19: 41091887-41091887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142884995 Chr.19: 41091881-41091881 on Build GRCh38
rs143140637 Chr.19: 41091870-41091870 on Build GRCh38
rs1017810792 Chr.19: 41091767-41091767 on Build GRCh38
rs968454156 Chr.19: 41091815-41091817 on Build GRCh38
rs760027894 Chr.19: 41091882-41091882 on Build GRCh38
rs773368843 Chr.19: 41091796-41091796 on Build GRCh38
rs753094702 Chr.19: 41091886-41091886 on Build GRCh38
rs766691370 Chr.19: 41091806-41091806 on Build GRCh38
rs8192789 Chr.19: 41091846-41091846 on Build GRCh38
rs149101733 Chr.19: 41091858-41091858 on Build GRCh38
rs575322415 Chr.19: 41091913-41091913 on Build GRCh38
rs749571026 Chr.19: 41091933-41091933 on Build GRCh38
rs373570182 Chr.19: 41091936-41091936 on Build GRCh38
rs752102241 Chr.19: 41091810-41091810 on Build GRCh38
rs771009114 Chr.19: 41091828-41091832 on Build GRCh38
rs766050277 Chr.19: 41091821-41091821 on Build GRCh38
rs768630299 Chr.19: 41091847-41091847 on Build GRCh38
rs763570614 Chr.19: 41091819-41091819 on Build GRCh38
rs150934830 Chr.19: 41091912-41091912 on Build GRCh38
rs371543439 Chr.19: 41091925-41091925 on Build GRCh38
rs779970029 Chr.19: 41091843-41091843 on Build GRCh38
rs764806309 Chr.19: 41091898-41091898 on Build GRCh38
rs138836481 Chr.19: 41091910-41091910 on Build GRCh38
rs758107795 Chr.19: 41091903-41091903 on Build GRCh38
rs546024260 Chr.19: 41091949-41091949 on Build GRCh38
rs750146228 Chr.19: 41091900-41091900 on Build GRCh38
rs1031303266 Chr.19: 41091826-41091826 on Build GRCh38
rs773456747 Chr.19: 41091863-41091863 on Build GRCh38
rs763332469 Chr.19: 41091803-41091803 on Build GRCh38
rs529694842 Chr.19: 41091931-41091931 on Build GRCh38
rs780057919 Chr.19: 41091909-41091909 on Build GRCh38
rs777809591 Chr.19: 41091930-41091930 on Build GRCh38
rs781214870 Chr.19: 41091914-41091914 on Build GRCh38
rs753393303 Chr.19: 41091820-41091820 on Build GRCh38
rs766780855 Chr.19: 41091871-41091871 on Build GRCh38
rs758306132 Chr.19: 41091841-41091841 on Build GRCh38
rs765504747 Chr.19: 41091777-41091777 on Build GRCh38
rs778649369 Chr.19: 41091822-41091822 on Build GRCh38
rs145048388 Chr.19: 41091897-41091897 on Build GRCh38
rs745476785 Chr.19: 41091839-41091839 on Build GRCh38
rs768082507 Chr.19: 41091883-41091883 on Build GRCh38
rs373705937 Chr.19: 41091815-41091815 on Build GRCh38
rs138089886 Chr.19: 41091850-41091850 on Build GRCh38

View Full Product Details