Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 691386-691659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEAF1
Entrez Gene ID
10522
Gene Name
DEAF1, transcription factor
Gene Aliases
MRD24, NUDR, SPN, ZMYND5
UniGene
Hs.243994
Gene Chromosome Location
Chr.11: 644220-695754 on Build GRCh38
Transcripts
NM_001293634.1 NM_021008.3 XM_011519842.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6272789 c.290-9C>A p.? Chr.11: 691607-691607 on Build GRCh38
5080378 c.362C>T p.A121V Chr.11: 691526-691526 on Build GRCh38
4487847 c.323C>A p.A108D Chr.11: 691565-691565 on Build GRCh38
6531013 c.385C>G p.L129V Chr.11: 691503-691503 on Build GRCh38
6222692 c.309G>T p.V103V Chr.11: 691579-691579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542820751 Chr.11: 691413-691413 on Build GRCh38
rs368618561 Chr.11: 691469-691469 on Build GRCh38
rs757482202 Chr.11: 691508-691508 on Build GRCh38
rs747955550 Chr.11: 691492-691492 on Build GRCh38
rs767174593 Chr.11: 691457-691457 on Build GRCh38
rs762223267 Chr.11: 691540-691540 on Build GRCh38
rs11246268 Chr.11: 691636-691636 on Build GRCh38
rs771686625 Chr.11: 691550-691550 on Build GRCh38
rs1026794758 Chr.11: 691445-691445 on Build GRCh38
rs147580202 Chr.11: 691552-691552 on Build GRCh38
rs769877185 Chr.11: 691517-691517 on Build GRCh38
rs774729594 Chr.11: 691451-691451 on Build GRCh38
rs754133173 Chr.11: 691477-691477 on Build GRCh38
rs765917060 Chr.11: 691474-691474 on Build GRCh38
rs191496432 Chr.11: 691568-691568 on Build GRCh38
rs544636887 Chr.11: 691415-691415 on Build GRCh38
rs749825655 Chr.11: 691527-691527 on Build GRCh38
rs752503742 Chr.11: 691573-691573 on Build GRCh38
rs780147004 Chr.11: 691602-691602 on Build GRCh38
rs769208184 Chr.11: 691610-691610 on Build GRCh38
rs764474281 Chr.11: 691450-691450 on Build GRCh38
rs753836757 Chr.11: 691484-691484 on Build GRCh38
rs781475171 Chr.11: 691510-691510 on Build GRCh38
rs375400731 Chr.11: 691455-691455 on Build GRCh38
rs779468991 Chr.11: 691613-691613 on Build GRCh38
rs1055604057 Chr.11: 691407-691407 on Build GRCh38
rs769120267 Chr.11: 691528-691528 on Build GRCh38
rs755642833 Chr.11: 691470-691470 on Build GRCh38
rs755103869 Chr.11: 691600-691600 on Build GRCh38
rs952519056 Chr.11: 691448-691448 on Build GRCh38
rs371300797 Chr.11: 691531-691531 on Build GRCh38
rs543674601 Chr.11: 691439-691439 on Build GRCh38
rs563914647 Chr.11: 691443-691443 on Build GRCh38
rs558105606 Chr.11: 691427-691427 on Build GRCh38
rs750680707 Chr.11: 691593-691593 on Build GRCh38
rs529419459 Chr.11: 691444-691444 on Build GRCh38
rs971142820 Chr.11: 691468-691468 on Build GRCh38
rs765720888 Chr.11: 691436-691436 on Build GRCh38
rs1038817783 Chr.11: 691594-691594 on Build GRCh38
rs761411048 Chr.11: 691454-691454 on Build GRCh38
rs868811033 Chr.11: 691524-691524 on Build GRCh38
rs146959211 Chr.11: 691435-691435 on Build GRCh38
rs758219303 Chr.11: 691574-691574 on Build GRCh38
rs780059631 Chr.11: 691525-691525 on Build GRCh38
rs1006670567 Chr.11: 691449-691450 on Build GRCh38
rs746959227 Chr.11: 691606-691608 on Build GRCh38
rs753887079 Chr.11: 691408-691408 on Build GRCh38
rs756288901 Chr.11: 691599-691599 on Build GRCh38
rs866717763 Chr.11: 691425-691425 on Build GRCh38
rs746012451 Chr.11: 691514-691514 on Build GRCh38
rs924561482 Chr.11: 691475-691475 on Build GRCh38
rs867499392 Chr.11: 691546-691546 on Build GRCh38
rs748533024 Chr.11: 691614-691614 on Build GRCh38
rs773396409 Chr.11: 691632-691632 on Build GRCh38
rs778895447 Chr.11: 691489-691489 on Build GRCh38
rs753371450 Chr.11: 691563-691563 on Build GRCh38
rs772496038 Chr.11: 691615-691615 on Build GRCh38
rs932986185 Chr.11: 691612-691612 on Build GRCh38
rs754930479 Chr.11: 691488-691488 on Build GRCh38
rs749346352 Chr.11: 691607-691607 on Build GRCh38
rs374249965 Chr.11: 691585-691585 on Build GRCh38
rs143376874 Chr.11: 691572-691572 on Build GRCh38
rs777811134 Chr.11: 691587-691588 on Build GRCh38
rs772918456 Chr.11: 691551-691551 on Build GRCh38

View Full Product Details