Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88731240-88731513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13A
Entrez Gene ID
10144
Gene Name
family with sequence similarity 13 member A
Gene Aliases
ARHGAP48, FAM13A1
UniGene
Hs.714536
Gene Chromosome Location
Chr.4: 88725954-89111398 on Build GRCh38
Transcripts
NM_001015045.2 NM_001265578.1 NM_001265579.1 NM_001265580.1 NM_014883.3 XM_005262681.2 XM_005262682.2 XM_005262683.2 XM_005262684.3 XM_006714057.3 XM_011531516.1 XM_011531517.1 XM_011531518.1 XM_011531519.2 XM_017007624.1 XM_017007625.1 XM_017007626.1 XM_017007627.1 XM_017007628.1 XM_017007629.1 XM_017007630.1 XM_017007631.1 XM_017007632.1 XM_017007633.1 XM_017007634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594667 c.1919G>A p.R640Q Chr.4: 88731375-88731375 on Build GRCh38
5727255 c.2853C>T p.L951L Chr.4: 88731419-88731419 on Build GRCh38
1594668 c.1930C>T p.R644W Chr.4: 88731364-88731364 on Build GRCh38
6836375 c.2848G>C p.E950Q Chr.4: 88731424-88731424 on Build GRCh38
345663 c.2869G>T p.E957* Chr.4: 88731403-88731403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777693502 Chr.4: 88731313-88731313 on Build GRCh38
rs756540995 Chr.4: 88731488-88731488 on Build GRCh38
rs992353891 Chr.4: 88731336-88731336 on Build GRCh38
rs77546245 Chr.4: 88731453-88731453 on Build GRCh38
rs372262048 Chr.4: 88731277-88731277 on Build GRCh38
rs748202885 Chr.4: 88731283-88731283 on Build GRCh38
rs772942276 Chr.4: 88731381-88731385 on Build GRCh38
rs76146228 Chr.4: 88731292-88731292 on Build GRCh38
rs961632504 Chr.4: 88731375-88731375 on Build GRCh38
rs768399312 Chr.4: 88731383-88731383 on Build GRCh38
rs770054139 Chr.4: 88731290-88731291 on Build GRCh38
rs755191324 Chr.4: 88731276-88731276 on Build GRCh38
rs201929602 Chr.4: 88731387-88731387 on Build GRCh38
rs764434542 Chr.4: 88731454-88731454 on Build GRCh38
rs761059361 Chr.4: 88731298-88731298 on Build GRCh38
rs781232272 Chr.4: 88731468-88731468 on Build GRCh38
rs2276321 Chr.4: 88731287-88731287 on Build GRCh38
rs190742441 Chr.4: 88731293-88731293 on Build GRCh38
rs751126519 Chr.4: 88731295-88731295 on Build GRCh38
rs770976655 Chr.4: 88731330-88731330 on Build GRCh38
rs759460587 Chr.4: 88731291-88731291 on Build GRCh38
rs779041175 Chr.4: 88731303-88731303 on Build GRCh38
rs762421042 Chr.4: 88731405-88731405 on Build GRCh38
rs376511180 Chr.4: 88731439-88731440 on Build GRCh38
rs746942575 Chr.4: 88731288-88731288 on Build GRCh38
rs368810193 Chr.4: 88731397-88731397 on Build GRCh38
rs769740735 Chr.4: 88731364-88731364 on Build GRCh38
rs369933365 Chr.4: 88731363-88731363 on Build GRCh38
rs768086122 Chr.4: 88731419-88731419 on Build GRCh38
rs753156152 Chr.4: 88731449-88731449 on Build GRCh38
rs773733799 Chr.4: 88731394-88731394 on Build GRCh38
rs781290967 Chr.4: 88731332-88731332 on Build GRCh38
rs775155074 Chr.4: 88731366-88731366 on Build GRCh38
rs139055334 Chr.4: 88731269-88731269 on Build GRCh38
rs758455003 Chr.4: 88731307-88731307 on Build GRCh38
rs372551129 Chr.4: 88731416-88731416 on Build GRCh38
rs752896067 Chr.4: 88731305-88731305 on Build GRCh38
rs377481966 Chr.4: 88731440-88731440 on Build GRCh38
rs147595342 Chr.4: 88731430-88731431 on Build GRCh38
rs770786270 Chr.4: 88731432-88731435 on Build GRCh38
rs766942575 Chr.4: 88731299-88731299 on Build GRCh38
rs745819891 Chr.4: 88731471-88731471 on Build GRCh38
rs776562280 Chr.4: 88731290-88731290 on Build GRCh38
rs543048834 Chr.4: 88731289-88731289 on Build GRCh38
rs757416303 Chr.4: 88731458-88731458 on Build GRCh38
rs772050578 Chr.4: 88731284-88731284 on Build GRCh38
rs751043687 Chr.4: 88731376-88731376 on Build GRCh38
rs747180976 Chr.4: 88731326-88731326 on Build GRCh38
rs751533839 Chr.4: 88731456-88731456 on Build GRCh38
rs759856452 Chr.4: 88731433-88731433 on Build GRCh38
rs754190300 Chr.4: 88731301-88731301 on Build GRCh38

View Full Product Details