Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94571393-94571666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBP2
Entrez Gene ID
8789
Gene Name
fructose-bisphosphatase 2
Gene Aliases
-
UniGene
Hs.61255
Gene Chromosome Location
Chr.9: 94558714-94593832 on Build GRCh38
Transcripts
NM_003837.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6478437 c.511G>A p.A171T Chr.9: 94571518-94571518 on Build GRCh38
7216902 c.530C>G p.S177C Chr.9: 94571499-94571499 on Build GRCh38
3909121 c.543C>T p.G181G Chr.9: 94571486-94571486 on Build GRCh38
7296093 c.562G>A p.D188N Chr.9: 94571467-94571467 on Build GRCh38
5029820 c.544G>A p.V182M Chr.9: 94571485-94571485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762728707 Chr.9: 94571493-94571493 on Build GRCh38
rs747653168 Chr.9: 94571455-94571455 on Build GRCh38
rs150017904 Chr.9: 94571596-94571596 on Build GRCh38
rs759228397 Chr.9: 94571557-94571557 on Build GRCh38
rs755644556 Chr.9: 94571515-94571515 on Build GRCh38
rs747429104 Chr.9: 94571605-94571605 on Build GRCh38
rs748538007 Chr.9: 94571498-94571498 on Build GRCh38
rs770324642 Chr.9: 94571462-94571462 on Build GRCh38
rs201554574 Chr.9: 94571518-94571518 on Build GRCh38
rs752378188 Chr.9: 94571571-94571571 on Build GRCh38
rs747245526 Chr.9: 94571628-94571628 on Build GRCh38
rs142583559 Chr.9: 94571525-94571525 on Build GRCh38
rs569397172 Chr.9: 94571473-94571473 on Build GRCh38
rs552321762 Chr.9: 94571513-94571513 on Build GRCh38
rs751410821 Chr.9: 94571556-94571556 on Build GRCh38
rs200434975 Chr.9: 94571469-94571469 on Build GRCh38
rs372505356 Chr.9: 94571600-94571600 on Build GRCh38
rs780811651 Chr.9: 94571454-94571454 on Build GRCh38
rs894451633 Chr.9: 94571632-94571634 on Build GRCh38
rs1019374520 Chr.9: 94571538-94571538 on Build GRCh38
rs768983196 Chr.9: 94571642-94571642 on Build GRCh38
rs555016192 Chr.9: 94571570-94571570 on Build GRCh38
rs754526159 Chr.9: 94571453-94571453 on Build GRCh38
rs906645425 Chr.9: 94571638-94571638 on Build GRCh38
rs375435231 Chr.9: 94571423-94571423 on Build GRCh38
rs903100563 Chr.9: 94571594-94571594 on Build GRCh38
rs61753274 Chr.9: 94571542-94571542 on Build GRCh38
rs529636593 Chr.9: 94571463-94571463 on Build GRCh38
rs951116234 Chr.9: 94571520-94571520 on Build GRCh38
rs377577284 Chr.9: 94571617-94571617 on Build GRCh38
rs780495699 Chr.9: 94571627-94571627 on Build GRCh38
rs1003247586 Chr.9: 94571420-94571420 on Build GRCh38
rs751298618 Chr.9: 94571500-94571500 on Build GRCh38
rs369503726 Chr.9: 94571475-94571475 on Build GRCh38
rs41281156 Chr.9: 94571519-94571519 on Build GRCh38
rs749748809 Chr.9: 94571465-94571465 on Build GRCh38
rs199525966 Chr.9: 94571532-94571532 on Build GRCh38
rs779672395 Chr.9: 94571438-94571438 on Build GRCh38
rs755699444 Chr.9: 94571574-94571574 on Build GRCh38
rs766221270 Chr.9: 94571544-94571544 on Build GRCh38
rs536762747 Chr.9: 94571494-94571494 on Build GRCh38
rs754725767 Chr.9: 94571504-94571504 on Build GRCh38
rs887266082 Chr.9: 94571424-94571424 on Build GRCh38
rs73531436 Chr.9: 94571543-94571543 on Build GRCh38
rs775781859 Chr.9: 94571485-94571485 on Build GRCh38
rs554337531 Chr.9: 94571499-94571499 on Build GRCh38
rs920924311 Chr.9: 94571547-94571547 on Build GRCh38
rs779396492 Chr.9: 94571524-94571524 on Build GRCh38
rs111655429 Chr.9: 94571645-94571645 on Build GRCh38
rs200244771 Chr.9: 94571488-94571488 on Build GRCh38
rs139243565 Chr.9: 94571590-94571590 on Build GRCh38
rs144416702 Chr.9: 94571486-94571486 on Build GRCh38
rs746673929 Chr.9: 94571452-94571452 on Build GRCh38
rs145254842 Chr.9: 94571624-94571624 on Build GRCh38
rs201283011 Chr.9: 94571533-94571533 on Build GRCh38
rs868722501 Chr.9: 94571567-94571567 on Build GRCh38
rs772380672 Chr.9: 94571531-94571531 on Build GRCh38
rs777280479 Chr.9: 94571588-94571588 on Build GRCh38
rs372755885 Chr.9: 94571483-94571483 on Build GRCh38

View Full Product Details