Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133373167-133373398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECHS1
Entrez Gene ID
1892
Gene Name
enoyl-CoA hydratase, short chain 1
Gene Aliases
ECHS1D, SCEH
UniGene
Hs.76394
Gene Chromosome Location
Chr.10: 133362483-133373404 on Build GRCh38
Transcripts
NM_004092.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
427288 c.32T>C p.V11A Chr.10: 133373302-133373302 on Build GRCh38
5182127 c.53C>T p.P18L Chr.10: 133373281-133373281 on Build GRCh38
6862963 c.30C>T p.C10C Chr.10: 133373304-133373304 on Build GRCh38
6518463 c.3G>T p.M1I Chr.10: 133373331-133373331 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764488387 Chr.10: 133373291-133373291 on Build GRCh38
rs372408822 Chr.10: 133373326-133373326 on Build GRCh38
rs201315896 Chr.10: 133373221-133373221 on Build GRCh38
rs758406761 Chr.10: 133373373-133373373 on Build GRCh38
rs532849742 Chr.10: 133373308-133373308 on Build GRCh38
rs754378922 Chr.10: 133373341-133373341 on Build GRCh38
rs980366726 Chr.10: 133373334-133373334 on Build GRCh38
rs928449973 Chr.10: 133373335-133373335 on Build GRCh38
rs748120988 Chr.10: 133373299-133373299 on Build GRCh38
rs376728317 Chr.10: 133373213-133373213 on Build GRCh38
rs761612881 Chr.10: 133373370-133373370 on Build GRCh38
rs779521503 Chr.10: 133373212-133373212 on Build GRCh38
rs769232756 Chr.10: 133373199-133373199 on Build GRCh38
rs587776498 Chr.10: 133373329-133373329 on Build GRCh38
rs759853219 Chr.10: 133373267-133373267 on Build GRCh38
rs1044498257 Chr.10: 133373196-133373196 on Build GRCh38
rs1053158402 Chr.10: 133373351-133373351 on Build GRCh38
rs891791920 Chr.10: 133373237-133373237 on Build GRCh38
rs1051394800 Chr.10: 133373306-133373306 on Build GRCh38
rs370494262 Chr.10: 133373325-133373325 on Build GRCh38
rs752716314 Chr.10: 133373283-133373283 on Build GRCh38
rs761464256 Chr.10: 133373241-133373241 on Build GRCh38
rs774394880 Chr.10: 133373236-133373236 on Build GRCh38
rs747670108 Chr.10: 133373369-133373369 on Build GRCh38
rs768233927 Chr.10: 133373298-133373298 on Build GRCh38
rs765613112 Chr.10: 133373279-133373279 on Build GRCh38
rs947348158 Chr.10: 133373366-133373366 on Build GRCh38
rs776926045 Chr.10: 133373262-133373262 on Build GRCh38
rs764269107 Chr.10: 133373322-133373322 on Build GRCh38
rs1043679816 Chr.10: 133373254-133373254 on Build GRCh38
rs775833766 Chr.10: 133373311-133373311 on Build GRCh38
rs10466126 Chr.10: 133373302-133373302 on Build GRCh38
rs779720008 Chr.10: 133373296-133373296 on Build GRCh38
rs752132371 Chr.10: 133373292-133373292 on Build GRCh38
rs377287845 Chr.10: 133373269-133373269 on Build GRCh38
rs900684601 Chr.10: 133373255-133373255 on Build GRCh38
rs771935232 Chr.10: 133373300-133373300 on Build GRCh38
rs778498370 Chr.10: 133373355-133373355 on Build GRCh38
rs760615795 Chr.10: 133373305-133373305 on Build GRCh38
rs765896234 Chr.10: 133373331-133373331 on Build GRCh38
rs375706106 Chr.10: 133373328-133373328 on Build GRCh38
rs372816008 Chr.10: 133373315-133373315 on Build GRCh38
rs587776497 Chr.10: 133373332-133373332 on Build GRCh38
rs748723043 Chr.10: 133373297-133373297 on Build GRCh38
rs921686244 Chr.10: 133373289-133373289 on Build GRCh38
rs1030441878 Chr.10: 133373273-133373273 on Build GRCh38
rs772023125 Chr.10: 133373252-133373252 on Build GRCh38
rs942234917 Chr.10: 133373189-133373189 on Build GRCh38
rs540787638 Chr.10: 133373274-133373274 on Build GRCh38
rs763135144 Chr.10: 133373284-133373284 on Build GRCh38
rs938782213 Chr.10: 133373342-133373342 on Build GRCh38
rs750839674 Chr.10: 133373295-133373295 on Build GRCh38
rs781545463 Chr.10: 133373293-133373293 on Build GRCh38
rs375893006 Chr.10: 133373324-133373324 on Build GRCh38
rs1002931828 Chr.10: 133373361-133373361 on Build GRCh38
rs753557738 Chr.10: 133373333-133373333 on Build GRCh38
rs867367723 Chr.10: 133373347-133373347 on Build GRCh38

View Full Product Details