Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53050512-53050767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERVV-2
Entrez Gene ID
100271846
Gene Name
endogenous retrovirus group V member 2
Gene Aliases
ENVV2, HERV-V2
UniGene
Hs.420445
Gene Chromosome Location
Chr.19: 53044738-53051127 on Build GRCh38
Transcripts
NM_001191055.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7318329 c.1460C>T p.S487F Chr.19: 53050711-53050711 on Build GRCh38
4127848 c.1465A>G p.I489V Chr.19: 53050716-53050716 on Build GRCh38
3960414 c.1420G>T p.G474C Chr.19: 53050671-53050671 on Build GRCh38
6216724 c.1271A>G p.Y424C Chr.19: 53050522-53050522 on Build GRCh38
6201335 c.1283C>T p.T428M Chr.19: 53050534-53050534 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753373875 Chr.19: 53050616-53050616 on Build GRCh38
rs919874284 Chr.19: 53050601-53050601 on Build GRCh38
rs777337894 Chr.19: 53050615-53050615 on Build GRCh38
rs543168344 Chr.19: 53050714-53050714 on Build GRCh38
rs1022492186 Chr.19: 53050698-53050698 on Build GRCh38
rs900534755 Chr.19: 53050723-53050723 on Build GRCh38
rs191985458 Chr.19: 53050573-53050573 on Build GRCh38
rs73935209 Chr.19: 53050739-53050739 on Build GRCh38
rs768966221 Chr.19: 53050719-53050719 on Build GRCh38
rs1040363919 Chr.19: 53050690-53050690 on Build GRCh38
rs767342293 Chr.19: 53050583-53050583 on Build GRCh38
rs557259411 Chr.19: 53050572-53050572 on Build GRCh38
rs1020202805 Chr.19: 53050541-53050541 on Build GRCh38
rs759293997 Chr.19: 53050578-53050578 on Build GRCh38
rs894146788 Chr.19: 53050682-53050682 on Build GRCh38
rs1048408307 Chr.19: 53050623-53050623 on Build GRCh38
rs370475038 Chr.19: 53050596-53050596 on Build GRCh38
rs150188507 Chr.19: 53050677-53050677 on Build GRCh38
rs867175435 Chr.19: 53050715-53050715 on Build GRCh38
rs930804588 Chr.19: 53050610-53050610 on Build GRCh38
rs971759109 Chr.19: 53050545-53050545 on Build GRCh38
rs138743720 Chr.19: 53050702-53050702 on Build GRCh38
rs184054973 Chr.19: 53050599-53050599 on Build GRCh38
rs773416174 Chr.19: 53050558-53050558 on Build GRCh38
rs748998616 Chr.19: 53050574-53050574 on Build GRCh38
rs755802085 Chr.19: 53050602-53050602 on Build GRCh38
rs920730512 Chr.19: 53050608-53050608 on Build GRCh38
rs536198422 Chr.19: 53050575-53050575 on Build GRCh38
rs577778652 Chr.19: 53050725-53050725 on Build GRCh38
rs553135248 Chr.19: 53050706-53050706 on Build GRCh38
rs973834832 Chr.19: 53050604-53050604 on Build GRCh38
rs780587138 Chr.19: 53050704-53050704 on Build GRCh38
rs908118276 Chr.19: 53050627-53050627 on Build GRCh38
rs756703795 Chr.19: 53050666-53050666 on Build GRCh38

View Full Product Details