Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6622137-6622410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCHS1
Entrez Gene ID
8642
Gene Name
dachsous cadherin-related 1
Gene Aliases
CDH19, CDH25, CDHR6, FIB1, MVP2, PCDH16, VMLDS1
UniGene
Hs.199850
Gene Chromosome Location
Chr.11: 6621324-6655849 on Build GRCh38
Transcripts
NM_003737.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4656509 c.9410C>T p.A3137V Chr.11: 6622266-6622266 on Build GRCh38
6238989 c.9434G>C p.G3145A Chr.11: 6622242-6622242 on Build GRCh38
4965076 c.9393G>A p.G3131G Chr.11: 6622283-6622283 on Build GRCh38
290450 c.9435T>G p.G3145G Chr.11: 6622241-6622241 on Build GRCh38
6640726 c.9334C>G p.P3112A Chr.11: 6622342-6622342 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753513428 Chr.11: 6622216-6622216 on Build GRCh38
rs760382120 Chr.11: 6622258-6622258 on Build GRCh38
rs752885784 Chr.11: 6622370-6622370 on Build GRCh38
rs745419879 Chr.11: 6622241-6622241 on Build GRCh38
rs752799248 Chr.11: 6622219-6622219 on Build GRCh38
rs776120615 Chr.11: 6622335-6622335 on Build GRCh38
rs756412840 Chr.11: 6622283-6622283 on Build GRCh38
rs780212955 Chr.11: 6622273-6622273 on Build GRCh38
rs766624649 Chr.11: 6622254-6622254 on Build GRCh38
rs576965402 Chr.11: 6622343-6622343 on Build GRCh38
rs775100869 Chr.11: 6622255-6622255 on Build GRCh38
rs150591111 Chr.11: 6622342-6622343 on Build GRCh38
rs1052300354 Chr.11: 6622310-6622310 on Build GRCh38
rs370906695 Chr.11: 6622359-6622359 on Build GRCh38
rs769811934 Chr.11: 6622253-6622254 on Build GRCh38
rs781265531 Chr.11: 6622277-6622277 on Build GRCh38
rs749845517 Chr.11: 6622308-6622308 on Build GRCh38
rs546047071 Chr.11: 6622381-6622381 on Build GRCh38
rs899789828 Chr.11: 6622364-6622364 on Build GRCh38
rs537187492 Chr.11: 6622225-6622225 on Build GRCh38
rs750572199 Chr.11: 6622220-6622220 on Build GRCh38
rs776549537 Chr.11: 6622263-6622263 on Build GRCh38
rs767829471 Chr.11: 6622357-6622357 on Build GRCh38
rs758453347 Chr.11: 6622312-6622312 on Build GRCh38
rs187251510 Chr.11: 6622260-6622260 on Build GRCh38
rs145042783 Chr.11: 6622217-6622217 on Build GRCh38
rs755284630 Chr.11: 6622274-6622274 on Build GRCh38
rs753145923 Chr.11: 6622282-6622282 on Build GRCh38
rs770139460 Chr.11: 6622167-6622167 on Build GRCh38
rs763981960 Chr.11: 6622204-6622204 on Build GRCh38
rs751809147 Chr.11: 6622232-6622232 on Build GRCh38
rs775261458 Chr.11: 6622199-6622199 on Build GRCh38
rs770397594 Chr.11: 6622239-6622239 on Build GRCh38
rs768426007 Chr.11: 6622328-6622328 on Build GRCh38
rs768138201 Chr.11: 6622259-6622259 on Build GRCh38
rs929369248 Chr.11: 6622293-6622293 on Build GRCh38
rs919429958 Chr.11: 6622246-6622246 on Build GRCh38
rs1037910186 Chr.11: 6622236-6622236 on Build GRCh38
rs766449407 Chr.11: 6622271-6622271 on Build GRCh38
rs928281168 Chr.11: 6622186-6622186 on Build GRCh38
rs746490919 Chr.11: 6622327-6622327 on Build GRCh38
rs761513230 Chr.11: 6622265-6622265 on Build GRCh38
rs771730718 Chr.11: 6622242-6622242 on Build GRCh38
rs554635448 Chr.11: 6622176-6622176 on Build GRCh38
rs781629251 Chr.11: 6622238-6622238 on Build GRCh38
rs751726201 Chr.11: 6622351-6622351 on Build GRCh38
rs762834205 Chr.11: 6622267-6622267 on Build GRCh38
rs367962728 Chr.11: 6622298-6622298 on Build GRCh38
rs748691401 Chr.11: 6622164-6622164 on Build GRCh38
rs775835626 Chr.11: 6622163-6622163 on Build GRCh38
rs756320439 Chr.11: 6622371-6622371 on Build GRCh38
rs755085535 Chr.11: 6622235-6622235 on Build GRCh38
rs779726789 Chr.11: 6622316-6622316 on Build GRCh38
rs758447145 Chr.11: 6622221-6622221 on Build GRCh38
rs375851798 Chr.11: 6622356-6622356 on Build GRCh38
rs761475301 Chr.11: 6622342-6622342 on Build GRCh38
rs759112008 Chr.11: 6622187-6622187 on Build GRCh38
rs760356441 Chr.11: 6622203-6622203 on Build GRCh38
rs765265323 Chr.11: 6622266-6622266 on Build GRCh38
rs762887975 Chr.11: 6622347-6622347 on Build GRCh38
rs766288089 Chr.11: 6622348-6622348 on Build GRCh38
rs1011816592 Chr.11: 6622369-6622369 on Build GRCh38
rs754199674 Chr.11: 6622382-6622382 on Build GRCh38
rs767319271 Chr.11: 6622194-6622194 on Build GRCh38
rs371537644 Chr.11: 6622334-6622334 on Build GRCh38

View Full Product Details