Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 130865439-130865694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41L2
Entrez Gene ID
2037
Gene Name
erythrocyte membrane protein band 4.1 like 2
Gene Aliases
4.1-G, 4.1G
UniGene
Hs.486470
Gene Chromosome Location
Chr.6: 130829111-131063322 on Build GRCh38
Transcripts
NM_001135554.1 NM_001135555.3 NM_001199388.2 NM_001252660.1 NM_001431.3 XM_006715362.2 XM_011535524.1 XM_011535525.1 XM_011535527.1 XM_011535528.1 XM_011535529.1 XM_011535530.1 XM_011535534.1 XM_011535536.1 XM_017010349.1 XM_017010350.1 XM_017010351.1 XM_017010352.1 XM_017010353.1 XM_017010354.1 XM_017010355.1 XM_017010356.1 XM_017010357.1 XM_017010358.1 XM_017010359.1 XM_017010360.1 XM_017010361.1 XM_017010362.1 XM_017010363.1 XM_017010364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6993366 c.2739C>T p.G913G Chr.6: 130865626-130865626 on Build GRCh38
4681171 c.2807C>T p.T936M Chr.6: 130865558-130865558 on Build GRCh38
6221134 c.2743G>A p.A915T Chr.6: 130865622-130865622 on Build GRCh38
3366296 c.2749G>A p.G917S Chr.6: 130865616-130865616 on Build GRCh38
3172957 c.2813C>A p.T938N Chr.6: 130865552-130865552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61731764 Chr.6: 130865592-130865592 on Build GRCh38
rs942613502 Chr.6: 130865564-130865564 on Build GRCh38
rs778166095 Chr.6: 130865617-130865617 on Build GRCh38
rs759667681 Chr.6: 130865570-130865570 on Build GRCh38
rs778217407 Chr.6: 130865567-130865567 on Build GRCh38
rs779533012 Chr.6: 130865608-130865608 on Build GRCh38
rs144870570 Chr.6: 130865603-130865603 on Build GRCh38
rs765333180 Chr.6: 130865558-130865560 on Build GRCh38
rs977437253 Chr.6: 130865512-130865512 on Build GRCh38
rs764464330 Chr.6: 130865580-130865580 on Build GRCh38
rs775807331 Chr.6: 130865589-130865589 on Build GRCh38
rs757920221 Chr.6: 130865529-130865529 on Build GRCh38
rs202080247 Chr.6: 130865626-130865626 on Build GRCh38
rs61731766 Chr.6: 130865602-130865602 on Build GRCh38
rs779491573 Chr.6: 130865632-130865632 on Build GRCh38
rs752682279 Chr.6: 130865647-130865647 on Build GRCh38
rs1031648055 Chr.6: 130865540-130865540 on Build GRCh38
rs145851233 Chr.6: 130865568-130865568 on Build GRCh38
rs762740895 Chr.6: 130865651-130865652 on Build GRCh38
rs866176810 Chr.6: 130865515-130865515 on Build GRCh38
rs775824701 Chr.6: 130865651-130865651 on Build GRCh38
rs149169560 Chr.6: 130865582-130865582 on Build GRCh38
rs997198568 Chr.6: 130865468-130865468 on Build GRCh38
rs753806085 Chr.6: 130865598-130865598 on Build GRCh38
rs536763663 Chr.6: 130865665-130865665 on Build GRCh38
rs753857584 Chr.6: 130865495-130865495 on Build GRCh38
rs150978866 Chr.6: 130865593-130865593 on Build GRCh38
rs750036736 Chr.6: 130865659-130865659 on Build GRCh38
rs745522943 Chr.6: 130865527-130865527 on Build GRCh38
rs749789634 Chr.6: 130865625-130865625 on Build GRCh38
rs990214106 Chr.6: 130865587-130865587 on Build GRCh38
rs756444268 Chr.6: 130865609-130865609 on Build GRCh38
rs778336536 Chr.6: 130865510-130865510 on Build GRCh38
rs779768264 Chr.6: 130865537-130865537 on Build GRCh38
rs770310654 Chr.6: 130865479-130865479 on Build GRCh38
rs200774281 Chr.6: 130865497-130865497 on Build GRCh38
rs149001048 Chr.6: 130865578-130865578 on Build GRCh38
rs910852195 Chr.6: 130865557-130865557 on Build GRCh38
rs760962588 Chr.6: 130865559-130865559 on Build GRCh38
rs186411905 Chr.6: 130865569-130865569 on Build GRCh38
rs745890518 Chr.6: 130865642-130865642 on Build GRCh38
rs199859070 Chr.6: 130865558-130865558 on Build GRCh38
rs1029586511 Chr.6: 130865474-130865474 on Build GRCh38
rs772228963 Chr.6: 130865644-130865644 on Build GRCh38

View Full Product Details