Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115732816-115733065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASQ2
Entrez Gene ID
845
Gene Name
calsequestrin 2
Gene Aliases
PDIB2
UniGene
Hs.57975
Gene Chromosome Location
Chr.1: 115700003-115768805 on Build GRCh38
Transcripts
NM_001232.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
256538 c.585C>A p.F195L Chr.1: 115732922-115732922 on Build GRCh38
4760422 c.572C>T p.P191L Chr.1: 115732935-115732935 on Build GRCh38
4185995 c.541G>T p.A181S Chr.1: 115732966-115732966 on Build GRCh38
7308461 c.577delA p.I193fs*17 Chr.1: 115732930-115732930 on Build GRCh38
3975923 c.572C>A p.P191H Chr.1: 115732935-115732935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759490509 Chr.1: 115732858-115732858 on Build GRCh38
rs751862386 Chr.1: 115733020-115733020 on Build GRCh38
rs146664754 Chr.1: 115732940-115732940 on Build GRCh38
rs397516643 Chr.1: 115732927-115732929 on Build GRCh38
rs749906718 Chr.1: 115732918-115732918 on Build GRCh38
rs370268584 Chr.1: 115732989-115732989 on Build GRCh38
rs771091279 Chr.1: 115732951-115732951 on Build GRCh38
rs545407254 Chr.1: 115732980-115732980 on Build GRCh38
rs781262104 Chr.1: 115732960-115732960 on Build GRCh38
rs745879915 Chr.1: 115732966-115732966 on Build GRCh38
rs761583118 Chr.1: 115732988-115732988 on Build GRCh38
rs762922862 Chr.1: 115732974-115732974 on Build GRCh38
rs144118950 Chr.1: 115732879-115732879 on Build GRCh38
rs763932609 Chr.1: 115732889-115732889 on Build GRCh38
rs767163972 Chr.1: 115732913-115732913 on Build GRCh38
rs753949363 Chr.1: 115732966-115732966 on Build GRCh38
rs373683706 Chr.1: 115733019-115733019 on Build GRCh38
rs765025646 Chr.1: 115732866-115732866 on Build GRCh38
rs779225945 Chr.1: 115732928-115732928 on Build GRCh38
rs751215231 Chr.1: 115732892-115732892 on Build GRCh38
rs918086929 Chr.1: 115733012-115733012 on Build GRCh38
rs765899666 Chr.1: 115732999-115732999 on Build GRCh38
rs912740566 Chr.1: 115732964-115732964 on Build GRCh38
rs747274504 Chr.1: 115732944-115732944 on Build GRCh38
rs755863384 Chr.1: 115732959-115732965 on Build GRCh38
rs758909428 Chr.1: 115733014-115733014 on Build GRCh38
rs72554057 Chr.1: 115732967-115732967 on Build GRCh38
rs756935347 Chr.1: 115732902-115732902 on Build GRCh38
rs760173068 Chr.1: 115732994-115732994 on Build GRCh38
rs958175108 Chr.1: 115732865-115732865 on Build GRCh38
rs544256146 Chr.1: 115732880-115732880 on Build GRCh38
rs755558916 Chr.1: 115732926-115732926 on Build GRCh38
rs187366018 Chr.1: 115732993-115732993 on Build GRCh38
rs886039816 Chr.1: 115732968-115732968 on Build GRCh38
rs986840982 Chr.1: 115732953-115732953 on Build GRCh38
rs767418776 Chr.1: 115732871-115732872 on Build GRCh38
rs753244130 Chr.1: 115732929-115732929 on Build GRCh38
rs575282077 Chr.1: 115733031-115733031 on Build GRCh38
rs763955301 Chr.1: 115732961-115732961 on Build GRCh38
rs753194494 Chr.1: 115733013-115733013 on Build GRCh38
rs752653901 Chr.1: 115732892-115732894 on Build GRCh38
rs974066405 Chr.1: 115732919-115732919 on Build GRCh38
rs774246167 Chr.1: 115732986-115732986 on Build GRCh38
rs786205791 Chr.1: 115732900-115732900 on Build GRCh38
rs758748280 Chr.1: 115732931-115732931 on Build GRCh38
rs778031867 Chr.1: 115733017-115733017 on Build GRCh38

View Full Product Details