Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186156077-186156306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM149A
Entrez Gene ID
25854
Gene Name
family with sequence similarity 149 member A
Gene Aliases
MST119, MSTP119
UniGene
Hs.744961
Gene Chromosome Location
Chr.4: 186104798-186172663 on Build GRCh38
Transcripts
NM_001006655.2 NM_015398.2 XM_005262911.1 XM_005262912.1 XM_005262913.1 XM_011531834.1 XM_011531835.2 XM_011531836.1 XM_017007996.1 XM_017007997.1 XM_017007998.1 XM_017007999.1 XM_017008000.1 XM_017008001.1 XM_017008002.1 XM_017008003.1 XM_017008004.1 XM_017008005.1 XM_017008006.1 XM_017008007.1 XM_017008008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6099784 c.462G>T p.P154P Chr.4: 186156078-186156078 on Build GRCh38
6658621 c.483C>T p.A161A Chr.4: 186156099-186156099 on Build GRCh38
6451672 c.1336C>T p.R446C Chr.4: 186156079-186156079 on Build GRCh38
6099785 c.1335G>T p.P445P Chr.4: 186156078-186156078 on Build GRCh38
4950242 c.1446A>G p.T482T Chr.4: 186156189-186156189 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368121879 Chr.4: 186156191-186156191 on Build GRCh38
rs368819152 Chr.4: 186156201-186156201 on Build GRCh38
rs572376870 Chr.4: 186156242-186156242 on Build GRCh38
rs62350488 Chr.4: 186156272-186156272 on Build GRCh38
rs556222486 Chr.4: 186156113-186156113 on Build GRCh38
rs146614253 Chr.4: 186156099-186156099 on Build GRCh38
rs761877863 Chr.4: 186156136-186156136 on Build GRCh38
rs377314469 Chr.4: 186156220-186156220 on Build GRCh38
rs752308639 Chr.4: 186156206-186156206 on Build GRCh38
rs372208223 Chr.4: 186156114-186156114 on Build GRCh38
rs751842987 Chr.4: 186156172-186156172 on Build GRCh38
rs370895680 Chr.4: 186156226-186156226 on Build GRCh38
rs7670550 Chr.4: 186156239-186156239 on Build GRCh38
rs768119890 Chr.4: 186156100-186156100 on Build GRCh38
rs755439404 Chr.4: 186156177-186156177 on Build GRCh38
rs574207344 Chr.4: 186156193-186156193 on Build GRCh38
rs764269790 Chr.4: 186156117-186156117 on Build GRCh38
rs201494456 Chr.4: 186156124-186156124 on Build GRCh38
rs750887144 Chr.4: 186156154-186156154 on Build GRCh38
rs780974220 Chr.4: 186156198-186156198 on Build GRCh38
rs752462255 Chr.4: 186156200-186156200 on Build GRCh38
rs13114283 Chr.4: 186156283-186156283 on Build GRCh38
rs892777267 Chr.4: 186156253-186156253 on Build GRCh38
rs144604546 Chr.4: 186156102-186156102 on Build GRCh38
rs776769122 Chr.4: 186156240-186156240 on Build GRCh38
rs762037425 Chr.4: 186156241-186156241 on Build GRCh38
rs745650578 Chr.4: 186156235-186156235 on Build GRCh38
rs141088571 Chr.4: 186156137-186156137 on Build GRCh38
rs762755273 Chr.4: 186156219-186156219 on Build GRCh38
rs200916497 Chr.4: 186156184-186156184 on Build GRCh38
rs745898966 Chr.4: 186156279-186156283 on Build GRCh38
rs766730836 Chr.4: 186156167-186156167 on Build GRCh38
rs763394753 Chr.4: 186156156-186156156 on Build GRCh38
rs773881614 Chr.4: 186156233-186156233 on Build GRCh38

View Full Product Details