Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10123178-10123447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC7A
Entrez Gene ID
64581
Gene Name
C-type lectin domain family 7 member A
Gene Aliases
BGR, CANDF4, CD369, CLECSF12, DECTIN1, SCARE2
UniGene
Hs.143929
Gene Chromosome Location
Chr.12: 10116777-10130269 on Build GRCh38
Transcripts
NM_022570.4 NM_197947.2 NM_197948.2 NM_197949.2 NM_197950.2 NR_125336.1 XM_006719135.3 XM_017019822.1 XM_017019823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5937535 c.593C>T p.S198L Chr.12: 10123263-10123263 on Build GRCh38
3764367 c.420C>T p.P140P Chr.12: 10123298-10123298 on Build GRCh38
3870560 c.409C>T p.L137F Chr.12: 10123309-10123309 on Build GRCh38
5373978 c.388G>A p.D130N Chr.12: 10123330-10123330 on Build GRCh38
3455178 c.493-1G>A p.? Chr.12: 10123364-10123364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748416724 Chr.12: 10123395-10123402 on Build GRCh38
rs754607559 Chr.12: 10123284-10123284 on Build GRCh38
rs140318683 Chr.12: 10123309-10123309 on Build GRCh38
rs764328683 Chr.12: 10123345-10123345 on Build GRCh38
rs761405478 Chr.12: 10123335-10123336 on Build GRCh38
rs758175995 Chr.12: 10123299-10123299 on Build GRCh38
rs746882037 Chr.12: 10123391-10123391 on Build GRCh38
rs753357530 Chr.12: 10123283-10123283 on Build GRCh38
rs751632742 Chr.12: 10123395-10123400 on Build GRCh38
rs757043413 Chr.12: 10123395-10123396 on Build GRCh38
rs781087069 Chr.12: 10123395-10123398 on Build GRCh38
rs763262416 Chr.12: 10123335-10123335 on Build GRCh38
rs776839889 Chr.12: 10123313-10123313 on Build GRCh38
rs139108713 Chr.12: 10123402-10123403 on Build GRCh38
rs759776167 Chr.12: 10123210-10123210 on Build GRCh38
rs758389873 Chr.12: 10123394-10123395 on Build GRCh38
rs762167028 Chr.12: 10123363-10123363 on Build GRCh38
rs750161794 Chr.12: 10123393-10123396 on Build GRCh38
rs764643298 Chr.12: 10123234-10123234 on Build GRCh38
rs116323098 Chr.12: 10123208-10123208 on Build GRCh38
rs12313329 Chr.12: 10123388-10123388 on Build GRCh38
rs74060566 Chr.12: 10123402-10123402 on Build GRCh38
rs375085998 Chr.12: 10123256-10123256 on Build GRCh38
rs11053609 Chr.12: 10123383-10123383 on Build GRCh38
rs757257799 Chr.12: 10123392-10123392 on Build GRCh38
rs770109934 Chr.12: 10123331-10123331 on Build GRCh38
rs765614983 Chr.12: 10123214-10123214 on Build GRCh38
rs571106532 Chr.12: 10123404-10123404 on Build GRCh38
rs754852458 Chr.12: 10123396-10123397 on Build GRCh38
rs751966143 Chr.12: 10123236-10123236 on Build GRCh38
rs370481697 Chr.12: 10123405-10123406 on Build GRCh38
rs763268770 Chr.12: 10123229-10123229 on Build GRCh38
rs548792483 Chr.12: 10123302-10123302 on Build GRCh38
rs199729236 Chr.12: 10123411-10123411 on Build GRCh38
rs775992557 Chr.12: 10123227-10123227 on Build GRCh38
rs61531383 Chr.12: 10123407-10123408 on Build GRCh38
rs778599643 Chr.12: 10123294-10123294 on Build GRCh38
rs774785126 Chr.12: 10123353-10123353 on Build GRCh38
rs764846626 Chr.12: 10123382-10123382 on Build GRCh38
rs373306442 Chr.12: 10123379-10123379 on Build GRCh38
rs370583536 Chr.12: 10123321-10123321 on Build GRCh38
rs200173938 Chr.12: 10123292-10123292 on Build GRCh38
rs775627850 Chr.12: 10123334-10123334 on Build GRCh38
rs777363102 Chr.12: 10123389-10123389 on Build GRCh38
rs888069504 Chr.12: 10123398-10123398 on Build GRCh38
rs771406474 Chr.12: 10123406-10123407 on Build GRCh38
rs746011994 Chr.12: 10123326-10123326 on Build GRCh38
rs537788182 Chr.12: 10123376-10123376 on Build GRCh38
rs369160278 Chr.12: 10123373-10123373 on Build GRCh38
rs765806491 Chr.12: 10123260-10123260 on Build GRCh38
rs765021234 Chr.12: 10123376-10123377 on Build GRCh38
rs977293590 Chr.12: 10123217-10123217 on Build GRCh38
rs747943115 Chr.12: 10123295-10123295 on Build GRCh38
rs749653620 Chr.12: 10123404-10123405 on Build GRCh38
rs771007056 Chr.12: 10123310-10123310 on Build GRCh38
rs553551197 Chr.12: 10123414-10123414 on Build GRCh38
rs11053610 Chr.12: 10123408-10123408 on Build GRCh38
rs915585977 Chr.12: 10123380-10123380 on Build GRCh38

View Full Product Details