Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206648572-206648839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYRK3
Entrez Gene ID
8444
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 3
Gene Aliases
DYRK5, RED, REDK, hYAK3-2
UniGene
Hs.164267
Gene Chromosome Location
Chr.1: 206635536-206655158 on Build GRCh38
Transcripts
NM_001004023.1 NM_003582.2 XM_005273315.4 XM_011510061.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3482478 c.1327C>A p.P443T Chr.1: 206648630-206648630 on Build GRCh38
4468988 c.1514C>T p.S505F Chr.1: 206648772-206648772 on Build GRCh38
6650761 c.1395A>G p.T465T Chr.1: 206648653-206648653 on Build GRCh38
6650791 c.1457C>T p.A486V Chr.1: 206648655-206648655 on Build GRCh38
5643919 c.1418A>G p.Y473C Chr.1: 206648676-206648676 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201585345 Chr.1: 206648597-206648597 on Build GRCh38
rs267598333 Chr.1: 206648628-206648628 on Build GRCh38
rs376496214 Chr.1: 206648735-206648735 on Build GRCh38
rs781980943 Chr.1: 206648601-206648601 on Build GRCh38
rs199976942 Chr.1: 206648808-206648808 on Build GRCh38
rs202224978 Chr.1: 206648674-206648674 on Build GRCh38
rs979281038 Chr.1: 206648746-206648746 on Build GRCh38
rs202021957 Chr.1: 206648622-206648622 on Build GRCh38
rs781832844 Chr.1: 206648800-206648800 on Build GRCh38
rs201271373 Chr.1: 206648655-206648655 on Build GRCh38
rs782619318 Chr.1: 206648723-206648723 on Build GRCh38
rs782361196 Chr.1: 206648814-206648814 on Build GRCh38
rs782517154 Chr.1: 206648645-206648645 on Build GRCh38
rs201668436 Chr.1: 206648717-206648717 on Build GRCh38
rs782466407 Chr.1: 206648719-206648719 on Build GRCh38
rs961358453 Chr.1: 206648592-206648592 on Build GRCh38
rs200378422 Chr.1: 206648726-206648726 on Build GRCh38
rs145519561 Chr.1: 206648607-206648607 on Build GRCh38
rs782505105 Chr.1: 206648801-206648801 on Build GRCh38
rs199630138 Chr.1: 206648774-206648774 on Build GRCh38
rs199912384 Chr.1: 206648632-206648632 on Build GRCh38
rs200713840 Chr.1: 206648816-206648816 on Build GRCh38
rs201669944 Chr.1: 206648755-206648755 on Build GRCh38
rs782102407 Chr.1: 206648753-206648753 on Build GRCh38
rs782146383 Chr.1: 206648784-206648784 on Build GRCh38
rs782577641 Chr.1: 206648701-206648701 on Build GRCh38
rs200820369 Chr.1: 206648806-206648806 on Build GRCh38
rs782635938 Chr.1: 206648802-206648802 on Build GRCh38
rs201711218 Chr.1: 206648817-206648817 on Build GRCh38
rs371457547 Chr.1: 206648792-206648792 on Build GRCh38
rs200749690 Chr.1: 206648797-206648797 on Build GRCh38
rs782636886 Chr.1: 206648815-206648815 on Build GRCh38
rs201089459 Chr.1: 206648685-206648685 on Build GRCh38
rs782388724 Chr.1: 206648727-206648727 on Build GRCh38
rs782557396 Chr.1: 206648700-206648700 on Build GRCh38
rs782754005 Chr.1: 206648761-206648761 on Build GRCh38
rs782131237 Chr.1: 206648606-206648606 on Build GRCh38
rs954903871 Chr.1: 206648616-206648616 on Build GRCh38
rs201099154 Chr.1: 206648610-206648610 on Build GRCh38
rs199768627 Chr.1: 206648809-206648809 on Build GRCh38
rs782310002 Chr.1: 206648740-206648740 on Build GRCh38
rs189150355 Chr.1: 206648676-206648676 on Build GRCh38
rs781995280 Chr.1: 206648593-206648593 on Build GRCh38
rs371758530 Chr.1: 206648743-206648743 on Build GRCh38
rs781939615 Chr.1: 206648596-206648596 on Build GRCh38
rs200284299 Chr.1: 206648677-206648677 on Build GRCh38
rs782319337 Chr.1: 206648595-206648595 on Build GRCh38
rs782035317 Chr.1: 206648621-206648621 on Build GRCh38
rs782463760 Chr.1: 206648795-206648795 on Build GRCh38
rs782350176 Chr.1: 206648600-206648600 on Build GRCh38
rs781838488 Chr.1: 206648643-206648643 on Build GRCh38
rs200657106 Chr.1: 206648794-206648794 on Build GRCh38

View Full Product Details