Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95822392-95822665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP57
Entrez Gene ID
9702
Gene Name
centrosomal protein 57
Gene Aliases
MVA2, PIG8, TSP57
UniGene
Hs.101014
Gene Chromosome Location
Chr.11: 95790461-95832693 on Build GRCh38
Transcripts
NM_001243776.1 NM_001243777.1 NM_014679.4 XM_006718945.2 XM_006718946.2 XM_017018592.1 XM_017018593.1 XM_017018594.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7117506 c.806C>T p.A269V Chr.11: 95822524-95822524 on Build GRCh38
7117505 c.833C>T p.A278V Chr.11: 95822524-95822524 on Build GRCh38
933612 c.861T>C p.G287G Chr.11: 95822552-95822552 on Build GRCh38
6627090 c.850delT p.L284fs*9 Chr.11: 95822541-95822541 on Build GRCh38
933611 c.859G>A p.G287S Chr.11: 95822550-95822550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751982189 Chr.11: 95822575-95822576 on Build GRCh38
rs1029918025 Chr.11: 95822553-95822553 on Build GRCh38
rs766796929 Chr.11: 95822555-95822556 on Build GRCh38
rs759400733 Chr.11: 95822515-95822515 on Build GRCh38
rs936446188 Chr.11: 95822616-95822616 on Build GRCh38
rs749032542 Chr.11: 95822506-95822506 on Build GRCh38
rs747587524 Chr.11: 95822611-95822611 on Build GRCh38
rs774143047 Chr.11: 95822513-95822513 on Build GRCh38
rs758928678 Chr.11: 95822604-95822604 on Build GRCh38
rs978198476 Chr.11: 95822600-95822600 on Build GRCh38
rs767473854 Chr.11: 95822528-95822528 on Build GRCh38
rs909631282 Chr.11: 95822467-95822467 on Build GRCh38
rs750852994 Chr.11: 95822595-95822595 on Build GRCh38
rs377308306 Chr.11: 95822509-95822509 on Build GRCh38
rs1006047995 Chr.11: 95822442-95822442 on Build GRCh38
rs753994812 Chr.11: 95822587-95822587 on Build GRCh38
rs975538357 Chr.11: 95822484-95822484 on Build GRCh38
rs374754066 Chr.11: 95822502-95822502 on Build GRCh38
rs368923668 Chr.11: 95822483-95822483 on Build GRCh38
rs764825721 Chr.11: 95822619-95822619 on Build GRCh38
rs765633450 Chr.11: 95822594-95822594 on Build GRCh38
rs760640925 Chr.11: 95822561-95822561 on Build GRCh38
rs951046260 Chr.11: 95822436-95822436 on Build GRCh38
rs775590763 Chr.11: 95822537-95822537 on Build GRCh38
rs368401485 Chr.11: 95822625-95822625 on Build GRCh38
rs1016541823 Chr.11: 95822476-95822476 on Build GRCh38
rs757568493 Chr.11: 95822588-95822588 on Build GRCh38
rs201955313 Chr.11: 95822477-95822477 on Build GRCh38
rs372413990 Chr.11: 95822493-95822493 on Build GRCh38
rs780715748 Chr.11: 95822459-95822459 on Build GRCh38
rs186476356 Chr.11: 95822613-95822613 on Build GRCh38
rs550858215 Chr.11: 95822584-95822584 on Build GRCh38

View Full Product Details