Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123785514-123785787 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM91A1
Entrez Gene ID
157769
Gene Name
family with sequence similarity 91 member A1
Gene Aliases
-
UniGene
Hs.459174
Gene Chromosome Location
Chr.8: 123768439-123815452 on Build GRCh38
Transcripts
NM_001317917.1 NM_001317918.1 NM_144963.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
393049 c.936G>C p.K312N Chr.8: 123785715-123785715 on Build GRCh38
3644864 c.949G>A p.V317I Chr.8: 123785728-123785728 on Build GRCh38
3644863 c.880T>C p.F294L Chr.8: 123785659-123785659 on Build GRCh38
6112060 c.947C>T p.S316F Chr.8: 123785726-123785726 on Build GRCh38
4817993 c.850-1G>A p.? Chr.8: 123785628-123785628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370830528 Chr.8: 123785651-123785651 on Build GRCh38
rs1007118367 Chr.8: 123785700-123785700 on Build GRCh38
rs763148313 Chr.8: 123785613-123785613 on Build GRCh38
rs1036079180 Chr.8: 123785559-123785559 on Build GRCh38
rs775673542 Chr.8: 123785612-123785612 on Build GRCh38
rs901592514 Chr.8: 123785576-123785576 on Build GRCh38
rs757876509 Chr.8: 123785728-123785728 on Build GRCh38
rs764319555 Chr.8: 123785629-123785629 on Build GRCh38
rs754582941 Chr.8: 123785698-123785698 on Build GRCh38
rs114381396 Chr.8: 123785600-123785600 on Build GRCh38
rs764887456 Chr.8: 123785717-123785717 on Build GRCh38
rs1038616115 Chr.8: 123785662-123785662 on Build GRCh38
rs777219788 Chr.8: 123785736-123785736 on Build GRCh38
rs377644313 Chr.8: 123785635-123785635 on Build GRCh38
rs374948607 Chr.8: 123785738-123785738 on Build GRCh38
rs373378220 Chr.8: 123785727-123785727 on Build GRCh38
rs557275534 Chr.8: 123785739-123785739 on Build GRCh38
rs938217403 Chr.8: 123785694-123785694 on Build GRCh38
rs534296048 Chr.8: 123785554-123785554 on Build GRCh38
rs750754728 Chr.8: 123785693-123785693 on Build GRCh38
rs574127883 Chr.8: 123785634-123785634 on Build GRCh38
rs538594952 Chr.8: 123785689-123785689 on Build GRCh38
rs770759392 Chr.8: 123785597-123785597 on Build GRCh38

View Full Product Details