Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37007994-37008251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOSC8
Entrez Gene ID
11340
Gene Name
exosome component 8
Gene Aliases
CIP3, EAP2, OIP2, PCH1C, RRP43, Rrp43p, bA421P11.3, p9
UniGene
Hs.294041
Gene Chromosome Location
Chr.13: 37000373-37009614 on Build GRCh38
Transcripts
NM_181503.2 XM_006719763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7310649 c.581T>C p.V194A Chr.13: 37008150-37008150 on Build GRCh38
4599962 c.508A>G p.I170V Chr.13: 37008077-37008077 on Build GRCh38
1366711 c.578C>T p.P193L Chr.13: 37008147-37008147 on Build GRCh38
947165 c.549G>T p.K183N Chr.13: 37008118-37008118 on Build GRCh38
7189176 c.558T>C p.Y186Y Chr.13: 37008127-37008127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754724110 Chr.13: 37008162-37008162 on Build GRCh38
rs139322433 Chr.13: 37008174-37008174 on Build GRCh38
rs529051683 Chr.13: 37008199-37008199 on Build GRCh38
rs200992929 Chr.13: 37008138-37008138 on Build GRCh38
rs371593334 Chr.13: 37008143-37008143 on Build GRCh38
rs576415848 Chr.13: 37008052-37008052 on Build GRCh38
rs746063414 Chr.13: 37008071-37008071 on Build GRCh38
rs755856415 Chr.13: 37008040-37008040 on Build GRCh38
rs763330676 Chr.13: 37008077-37008077 on Build GRCh38
rs142300123 Chr.13: 37008038-37008038 on Build GRCh38
rs778769278 Chr.13: 37008046-37008046 on Build GRCh38
rs761368538 Chr.13: 37008027-37008027 on Build GRCh38
rs751122775 Chr.13: 37008123-37008123 on Build GRCh38
rs745774793 Chr.13: 37008179-37008186 on Build GRCh38
rs779713251 Chr.13: 37008041-37008041 on Build GRCh38
rs758385926 Chr.13: 37008187-37008187 on Build GRCh38
rs1014412265 Chr.13: 37008210-37008210 on Build GRCh38
rs367765614 Chr.13: 37008101-37008101 on Build GRCh38
rs924212382 Chr.13: 37008069-37008069 on Build GRCh38
rs775590997 Chr.13: 37008076-37008076 on Build GRCh38
rs770119494 Chr.13: 37008075-37008075 on Build GRCh38
rs945192906 Chr.13: 37008188-37008188 on Build GRCh38
rs752608874 Chr.13: 37008183-37008183 on Build GRCh38
rs757075598 Chr.13: 37008113-37008115 on Build GRCh38
rs770966591 Chr.13: 37008056-37008056 on Build GRCh38
rs746592008 Chr.13: 37008199-37008200 on Build GRCh38
rs778870949 Chr.13: 37008119-37008120 on Build GRCh38
rs769788723 Chr.13: 37008203-37008203 on Build GRCh38
rs774862843 Chr.13: 37008196-37008197 on Build GRCh38
rs753450309 Chr.13: 37008159-37008159 on Build GRCh38
rs753598965 Chr.13: 37008042-37008042 on Build GRCh38
rs150857216 Chr.13: 37008057-37008057 on Build GRCh38
rs978747582 Chr.13: 37008061-37008061 on Build GRCh38
rs765996165 Chr.13: 37008152-37008152 on Build GRCh38
rs745903345 Chr.13: 37008194-37008194 on Build GRCh38
rs750019667 Chr.13: 37008141-37008141 on Build GRCh38
rs748384256 Chr.13: 37008112-37008112 on Build GRCh38
rs769175023 Chr.13: 37008092-37008092 on Build GRCh38
rs762463961 Chr.13: 37008100-37008100 on Build GRCh38
rs781126029 Chr.13: 37008189-37008189 on Build GRCh38
rs781416113 Chr.13: 37008112-37008115 on Build GRCh38
rs774971786 Chr.13: 37008096-37008096 on Build GRCh38
rs751701456 Chr.13: 37008102-37008106 on Build GRCh38
rs755140645 Chr.13: 37008105-37008109 on Build GRCh38
rs754740846 Chr.13: 37008045-37008045 on Build GRCh38
rs759626150 Chr.13: 37008196-37008199 on Build GRCh38

View Full Product Details