Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172935081-172935316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC1
Entrez Gene ID
57222
Gene Name
endoplasmic reticulum-golgi intermediate compartment 1
Gene Aliases
ERGIC-32, ERGIC32, NET24
UniGene
Hs.509163
Gene Chromosome Location
Chr.5: 172834220-172952685 on Build GRCh38
Transcripts
NM_001031711.2 XM_006714892.1 XM_011534597.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6656080 c.715A>C p.T239P Chr.5: 172935260-172935260 on Build GRCh38
5780146 c.753A>G p.R251R Chr.5: 172935298-172935298 on Build GRCh38
1251480 c.703C>G p.L235V Chr.5: 172935248-172935248 on Build GRCh38
6656078 c.710C>T p.P237L Chr.5: 172935255-172935255 on Build GRCh38
5709647 c.682G>T p.A228S Chr.5: 172935227-172935227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761054660 Chr.5: 172935257-172935257 on Build GRCh38
rs763037748 Chr.5: 172935159-172935159 on Build GRCh38
rs768357064 Chr.5: 172935217-172935217 on Build GRCh38
rs547013747 Chr.5: 172935196-172935196 on Build GRCh38
rs771935390 Chr.5: 172935211-172935211 on Build GRCh38
rs754272525 Chr.5: 172935272-172935272 on Build GRCh38
rs749645712 Chr.5: 172935247-172935247 on Build GRCh38
rs755538561 Chr.5: 172935176-172935176 on Build GRCh38
rs369647590 Chr.5: 172935141-172935141 on Build GRCh38
rs267600553 Chr.5: 172935220-172935220 on Build GRCh38
rs774919836 Chr.5: 172935281-172935281 on Build GRCh38
rs745943189 Chr.5: 172935276-172935277 on Build GRCh38
rs200203918 Chr.5: 172935142-172935142 on Build GRCh38
rs1046167901 Chr.5: 172935140-172935140 on Build GRCh38
rs373899742 Chr.5: 172935215-172935215 on Build GRCh38
rs376307473 Chr.5: 172935240-172935240 on Build GRCh38
rs370260183 Chr.5: 172935244-172935244 on Build GRCh38
rs753290357 Chr.5: 172935248-172935248 on Build GRCh38
rs1027506523 Chr.5: 172935271-172935271 on Build GRCh38
rs757931584 Chr.5: 172935288-172935288 on Build GRCh38
rs769103026 Chr.5: 172935261-172935261 on Build GRCh38
rs566933192 Chr.5: 172935282-172935282 on Build GRCh38
rs376329857 Chr.5: 172935136-172935136 on Build GRCh38
rs767253308 Chr.5: 172935144-172935144 on Build GRCh38
rs369980778 Chr.5: 172935210-172935210 on Build GRCh38
rs967885321 Chr.5: 172935174-172935174 on Build GRCh38
rs779916330 Chr.5: 172935289-172935289 on Build GRCh38
rs141880390 Chr.5: 172935197-172935197 on Build GRCh38
rs200766378 Chr.5: 172935137-172935137 on Build GRCh38
rs201862360 Chr.5: 172935144-172935144 on Build GRCh38
rs113708386 Chr.5: 172935152-172935152 on Build GRCh38
rs944672324 Chr.5: 172935139-172935139 on Build GRCh38
rs775408246 Chr.5: 172935212-172935212 on Build GRCh38
rs945157864 Chr.5: 172935164-172935164 on Build GRCh38
rs983914293 Chr.5: 172935111-172935111 on Build GRCh38
rs773337014 Chr.5: 172935155-172935155 on Build GRCh38
rs778258760 Chr.5: 172935206-172935206 on Build GRCh38
rs77718424 Chr.5: 172935173-172935173 on Build GRCh38
rs774560942 Chr.5: 172935243-172935243 on Build GRCh38
rs148095919 Chr.5: 172935135-172935135 on Build GRCh38
rs201274324 Chr.5: 172935200-172935200 on Build GRCh38
rs200458222 Chr.5: 172935183-172935183 on Build GRCh38
rs756843543 Chr.5: 172935274-172935274 on Build GRCh38
rs775248401 Chr.5: 172935143-172935143 on Build GRCh38
rs763054803 Chr.5: 172935232-172935232 on Build GRCh38
rs746925479 Chr.5: 172935295-172935295 on Build GRCh38
rs199695105 Chr.5: 172935163-172935163 on Build GRCh38
rs1054655067 Chr.5: 172935153-172935153 on Build GRCh38
rs749866807 Chr.5: 172935283-172935283 on Build GRCh38
rs757370376 Chr.5: 172935143-172935144 on Build GRCh38
rs756553012 Chr.5: 172935194-172935194 on Build GRCh38
rs774935293 Chr.5: 172935115-172935115 on Build GRCh38
rs776536979 Chr.5: 172935230-172935230 on Build GRCh38

View Full Product Details