Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33621919-33622146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYZL1
Entrez Gene ID
9946
Gene Name
crystallin zeta like 1
Gene Aliases
4P11, QOH-1
UniGene
Hs.352671
Gene Chromosome Location
Chr.21: 33589341-33641854 on Build GRCh38
Transcripts
NM_145858.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5665160 c.187G>T p.G63W Chr.21: 33622026-33622026 on Build GRCh38
5607343 c.162G>A p.K54K Chr.21: 33622051-33622051 on Build GRCh38
419250 c.150G>A p.L50L Chr.21: 33622063-33622063 on Build GRCh38
3550421 c.181C>T p.P61S Chr.21: 33622032-33622032 on Build GRCh38
4910053 c.156A>C p.E52D Chr.21: 33622057-33622057 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767679950 Chr.21: 33622039-33622039 on Build GRCh38
rs1022422889 Chr.21: 33622072-33622072 on Build GRCh38
rs573109062 Chr.21: 33622110-33622110 on Build GRCh38
rs375789567 Chr.21: 33622075-33622075 on Build GRCh38
rs778844223 Chr.21: 33622101-33622101 on Build GRCh38
rs987927160 Chr.21: 33622053-33622053 on Build GRCh38
rs751989176 Chr.21: 33622024-33622024 on Build GRCh38
rs369350772 Chr.21: 33621992-33621992 on Build GRCh38
rs775046355 Chr.21: 33621991-33621991 on Build GRCh38
rs757318413 Chr.21: 33622087-33622087 on Build GRCh38
rs1031862936 Chr.21: 33622017-33622017 on Build GRCh38
rs763246778 Chr.21: 33622003-33622003 on Build GRCh38
rs557547779 Chr.21: 33622104-33622104 on Build GRCh38
rs747485370 Chr.21: 33621979-33621980 on Build GRCh38
rs1004462530 Chr.21: 33622079-33622079 on Build GRCh38
rs139699692 Chr.21: 33621997-33621997 on Build GRCh38
rs371780656 Chr.21: 33621989-33621989 on Build GRCh38
rs781503947 Chr.21: 33621962-33621965 on Build GRCh38
rs539492201 Chr.21: 33622085-33622085 on Build GRCh38
rs766717697 Chr.21: 33622009-33622009 on Build GRCh38
rs749050481 Chr.21: 33621958-33621958 on Build GRCh38
rs371802101 Chr.21: 33622029-33622029 on Build GRCh38
rs1039788469 Chr.21: 33621998-33621998 on Build GRCh38
rs756178055 Chr.21: 33622065-33622065 on Build GRCh38
rs779606802 Chr.21: 33622115-33622115 on Build GRCh38
rs752772127 Chr.21: 33622054-33622054 on Build GRCh38
rs778646128 Chr.21: 33621969-33621969 on Build GRCh38
rs745498862 Chr.21: 33621977-33621977 on Build GRCh38
rs773601013 Chr.21: 33622001-33622001 on Build GRCh38
rs770853755 Chr.21: 33621967-33621967 on Build GRCh38

View Full Product Details