Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 104349143-104349404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCSTAMP
Entrez Gene ID
81501
Gene Name
dendrocyte expressed seven transmembrane protein
Gene Aliases
FIND, TM7SF4, hDC-STAMP
UniGene
Hs.652230
Gene Chromosome Location
Chr.8: 104339087-104356874 on Build GRCh38
Transcripts
NM_001257317.1 NM_030788.3 XM_011517321.1 XM_011517324.1 XM_017013878.1 XM_017013879.1 XM_017013880.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4880191 c.639G>C p.K213N Chr.8: 104349191-104349191 on Build GRCh38
4149474 c.727G>A p.E243K Chr.8: 104349279-104349279 on Build GRCh38
4899845 c.807G>A p.L269L Chr.8: 104349359-104349359 on Build GRCh38
6318246 c.657G>T p.G219G Chr.8: 104349209-104349209 on Build GRCh38
1700446 c.814G>A p.E272K Chr.8: 104349366-104349366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144160852 Chr.8: 104349276-104349276 on Build GRCh38
rs748298141 Chr.8: 104349226-104349226 on Build GRCh38
rs767122336 Chr.8: 104349174-104349174 on Build GRCh38
rs141043655 Chr.8: 104349317-104349317 on Build GRCh38
rs566008351 Chr.8: 104349219-104349219 on Build GRCh38
rs905159606 Chr.8: 104349177-104349177 on Build GRCh38
rs766882223 Chr.8: 104349285-104349285 on Build GRCh38
rs749203316 Chr.8: 104349250-104349250 on Build GRCh38
rs148704252 Chr.8: 104349215-104349215 on Build GRCh38
rs757950369 Chr.8: 104349300-104349300 on Build GRCh38
rs777391980 Chr.8: 104349317-104349318 on Build GRCh38
rs545046616 Chr.8: 104349381-104349381 on Build GRCh38
rs759216839 Chr.8: 104349173-104349173 on Build GRCh38
rs760055630 Chr.8: 104349379-104349379 on Build GRCh38
rs200017505 Chr.8: 104349259-104349259 on Build GRCh38
rs750901832 Chr.8: 104349207-104349207 on Build GRCh38
rs866546975 Chr.8: 104349179-104349179 on Build GRCh38
rs775855383 Chr.8: 104349204-104349204 on Build GRCh38
rs146506048 Chr.8: 104349303-104349303 on Build GRCh38
rs755292234 Chr.8: 104349218-104349218 on Build GRCh38
rs766098559 Chr.8: 104349169-104349169 on Build GRCh38
rs752973542 Chr.8: 104349311-104349311 on Build GRCh38
rs763529427 Chr.8: 104349280-104349280 on Build GRCh38
rs200509068 Chr.8: 104349296-104349296 on Build GRCh38
rs763207658 Chr.8: 104349232-104349232 on Build GRCh38
rs770819792 Chr.8: 104349321-104349321 on Build GRCh38
rs752110585 Chr.8: 104349366-104349366 on Build GRCh38
rs540504854 Chr.8: 104349282-104349282 on Build GRCh38
rs775211373 Chr.8: 104349350-104349350 on Build GRCh38
rs745726484 Chr.8: 104349328-104349328 on Build GRCh38
rs759300750 Chr.8: 104349262-104349262 on Build GRCh38
rs761365381 Chr.8: 104349364-104349364 on Build GRCh38
rs749355177 Chr.8: 104349320-104349320 on Build GRCh38
rs755382087 Chr.8: 104349300-104349300 on Build GRCh38
rs756354649 Chr.8: 104349313-104349313 on Build GRCh38
rs537660977 Chr.8: 104349339-104349339 on Build GRCh38
rs139353637 Chr.8: 104349344-104349344 on Build GRCh38
rs776048966 Chr.8: 104349361-104349361 on Build GRCh38
rs774821780 Chr.8: 104349356-104349356 on Build GRCh38
rs766854020 Chr.8: 104349365-104349365 on Build GRCh38
rs79659609 Chr.8: 104349304-104349304 on Build GRCh38
rs765796550 Chr.8: 104349205-104349205 on Build GRCh38
rs144238227 Chr.8: 104349214-104349214 on Build GRCh38
rs922352502 Chr.8: 104349382-104349382 on Build GRCh38
rs149993718 Chr.8: 104349199-104349199 on Build GRCh38
rs758884670 Chr.8: 104349209-104349209 on Build GRCh38
rs767210591 Chr.8: 104349271-104349271 on Build GRCh38
rs775117480 Chr.8: 104349275-104349275 on Build GRCh38
rs190999185 Chr.8: 104349233-104349233 on Build GRCh38
rs770861535 Chr.8: 104349254-104349254 on Build GRCh38
rs143355193 Chr.8: 104349380-104349380 on Build GRCh38
rs760345015 Chr.8: 104349355-104349355 on Build GRCh38
rs763405051 Chr.8: 104349284-104349284 on Build GRCh38
rs752249205 Chr.8: 104349191-104349191 on Build GRCh38

View Full Product Details