Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 45455615-45455888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL18A1
Entrez Gene ID
80781
Gene Name
collagen type XVIII alpha 1 chain
Gene Aliases
KNO, KNO1, KS
UniGene
Hs.517356
Gene Chromosome Location
Chr.21: 45405137-45513720 on Build GRCh38
Transcripts
NM_030582.3 NM_130444.2 NM_130445.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3939528 c.292G>A p.E98K Chr.21: 45455822-45455822 on Build GRCh38
4582129 c.179C>T p.T60I Chr.21: 45455709-45455709 on Build GRCh38
3939529 c.292G>A p.E98K Chr.21: 45455822-45455822 on Build GRCh38
6632599 c.194delC p.R67fs*49 Chr.21: 45455724-45455724 on Build GRCh38
4004826 c.106G>A p.E36K Chr.21: 45455636-45455636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750868738 Chr.21: 45455867-45455867 on Build GRCh38
rs370000815 Chr.21: 45455854-45455854 on Build GRCh38
rs529405045 Chr.21: 45455844-45455844 on Build GRCh38
rs114139997 Chr.21: 45455861-45455861 on Build GRCh38
rs769637337 Chr.21: 45455654-45455654 on Build GRCh38
rs375958675 Chr.21: 45455769-45455770 on Build GRCh38
rs1031791883 Chr.21: 45455734-45455734 on Build GRCh38
rs867129226 Chr.21: 45455853-45455853 on Build GRCh38
rs760886724 Chr.21: 45455684-45455684 on Build GRCh38
rs775587295 Chr.21: 45455850-45455850 on Build GRCh38
rs200354859 Chr.21: 45455701-45455701 on Build GRCh38
rs61749002 Chr.21: 45455672-45455672 on Build GRCh38
rs746967987 Chr.21: 45455839-45455839 on Build GRCh38
rs772982058 Chr.21: 45455768-45455768 on Build GRCh38
rs752886739 Chr.21: 45455793-45455793 on Build GRCh38
rs764004091 Chr.21: 45455723-45455724 on Build GRCh38
rs948575157 Chr.21: 45455778-45455778 on Build GRCh38
rs199984023 Chr.21: 45455661-45455661 on Build GRCh38
rs773963114 Chr.21: 45455757-45455757 on Build GRCh38
rs754701304 Chr.21: 45455829-45455829 on Build GRCh38
rs764301199 Chr.21: 45455855-45455855 on Build GRCh38
rs201529109 Chr.21: 45455821-45455821 on Build GRCh38
rs764638296 Chr.21: 45455812-45455812 on Build GRCh38
rs748717777 Chr.21: 45455641-45455641 on Build GRCh38
rs373586271 Chr.21: 45455720-45455720 on Build GRCh38
rs781638267 Chr.21: 45455822-45455822 on Build GRCh38
rs970127668 Chr.21: 45455807-45455808 on Build GRCh38
rs749001958 Chr.21: 45455751-45455751 on Build GRCh38
rs377623821 Chr.21: 45455842-45455842 on Build GRCh38
rs748007378 Chr.21: 45455762-45455762 on Build GRCh38
rs770111210 Chr.21: 45455849-45455849 on Build GRCh38
rs776665393 Chr.21: 45455771-45455771 on Build GRCh38
rs745484237 Chr.21: 45455651-45455651 on Build GRCh38
rs199929202 Chr.21: 45455709-45455709 on Build GRCh38
rs928576283 Chr.21: 45455644-45455644 on Build GRCh38
rs767745134 Chr.21: 45455866-45455866 on Build GRCh38
rs375087150 Chr.21: 45455729-45455729 on Build GRCh38
rs762647237 Chr.21: 45455682-45455691 on Build GRCh38
rs779506536 Chr.21: 45455748-45455748 on Build GRCh38
rs563619668 Chr.21: 45455770-45455770 on Build GRCh38
rs747435605 Chr.21: 45455730-45455730 on Build GRCh38
rs370812546 Chr.21: 45455830-45455830 on Build GRCh38
rs979960718 Chr.21: 45455796-45455796 on Build GRCh38
rs753411491 Chr.21: 45455827-45455827 on Build GRCh38
rs61739237 Chr.21: 45455769-45455769 on Build GRCh38
rs61735029 Chr.21: 45455676-45455676 on Build GRCh38
rs375714903 Chr.21: 45455846-45455846 on Build GRCh38
rs368696106 Chr.21: 45455665-45455665 on Build GRCh38
rs759848880 Chr.21: 45455781-45455781 on Build GRCh38
rs773035029 Chr.21: 45455678-45455678 on Build GRCh38
rs765377882 Chr.21: 45455714-45455714 on Build GRCh38
rs149777818 Chr.21: 45455833-45455833 on Build GRCh38
rs914230 Chr.21: 45455650-45455650 on Build GRCh38
rs748689940 Chr.21: 45455662-45455662 on Build GRCh38
rs757709318 Chr.21: 45455731-45455731 on Build GRCh38
rs754968843 Chr.21: 45455710-45455710 on Build GRCh38
rs753119196 Chr.21: 45455719-45455719 on Build GRCh38
rs768003538 Chr.21: 45455752-45455752 on Build GRCh38
rs758784072 Chr.21: 45455643-45455643 on Build GRCh38
rs777643778 Chr.21: 45455836-45455836 on Build GRCh38
rs771588051 Chr.21: 45455834-45455834 on Build GRCh38
rs758958927 Chr.21: 45455795-45455795 on Build GRCh38
rs374634058 Chr.21: 45455860-45455860 on Build GRCh38
rs938697545 Chr.21: 45455652-45455652 on Build GRCh38
rs537048520 Chr.21: 45455764-45455764 on Build GRCh38
rs765197743 Chr.21: 45455788-45455788 on Build GRCh38
rs761835450 Chr.21: 45455674-45455674 on Build GRCh38

View Full Product Details