Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28933008-28933233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD19
Entrez Gene ID
930
Gene Name
CD19 molecule
Gene Aliases
B4, CVID3
UniGene
Hs.652262
Gene Chromosome Location
Chr.16: 28931735-28939347 on Build GRCh38
Transcripts
NM_001178098.1 NM_001770.5 XM_006721103.3 XM_011545981.1 XM_017023893.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1718359 c.514C>T p.P172S Chr.16: 28933069-28933069 on Build GRCh38
4550535 c.502G>A p.E168K Chr.16: 28933057-28933057 on Build GRCh38
5850652 c.518G>A p.C173Y Chr.16: 28933073-28933073 on Build GRCh38
7244180 c.520C>G p.L174V Chr.16: 28933075-28933075 on Build GRCh38
435059 c.487C>T p.R163C Chr.16: 28933042-28933042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751091345 Chr.16: 28933165-28933165 on Build GRCh38
rs761506815 Chr.16: 28933146-28933146 on Build GRCh38
rs1056128085 Chr.16: 28933055-28933055 on Build GRCh38
rs369028655 Chr.16: 28933142-28933142 on Build GRCh38
rs765136451 Chr.16: 28933147-28933147 on Build GRCh38
rs935022728 Chr.16: 28933034-28933034 on Build GRCh38
rs750438736 Chr.16: 28933069-28933069 on Build GRCh38
rs771361830 Chr.16: 28933041-28933041 on Build GRCh38
rs955325757 Chr.16: 28933187-28933187 on Build GRCh38
rs372281868 Chr.16: 28933124-28933124 on Build GRCh38
rs1029666682 Chr.16: 28933171-28933171 on Build GRCh38
rs768494152 Chr.16: 28933146-28933147 on Build GRCh38
rs767159408 Chr.16: 28933190-28933190 on Build GRCh38
rs746445038 Chr.16: 28933126-28933126 on Build GRCh38
rs915729237 Chr.16: 28933173-28933173 on Build GRCh38
rs968233112 Chr.16: 28933211-28933211 on Build GRCh38
rs769025685 Chr.16: 28933140-28933140 on Build GRCh38
rs568312450 Chr.16: 28933083-28933083 on Build GRCh38
rs372571439 Chr.16: 28933150-28933150 on Build GRCh38
rs749626059 Chr.16: 28933107-28933107 on Build GRCh38
rs747028041 Chr.16: 28933139-28933143 on Build GRCh38
rs1018349529 Chr.16: 28933202-28933202 on Build GRCh38
rs757945792 Chr.16: 28933124-28933125 on Build GRCh38
rs143769633 Chr.16: 28933029-28933029 on Build GRCh38
rs761027351 Chr.16: 28933047-28933047 on Build GRCh38
rs985557056 Chr.16: 28933197-28933197 on Build GRCh38
rs896098197 Chr.16: 28933070-28933070 on Build GRCh38
rs778061704 Chr.16: 28933097-28933097 on Build GRCh38
rs148200569 Chr.16: 28933082-28933082 on Build GRCh38
rs369297926 Chr.16: 28933158-28933158 on Build GRCh38
rs747198608 Chr.16: 28933139-28933139 on Build GRCh38
rs2904880 Chr.16: 28933075-28933075 on Build GRCh38
rs754564780 Chr.16: 28933188-28933188 on Build GRCh38
rs753721854 Chr.16: 28933050-28933050 on Build GRCh38
rs771403335 Chr.16: 28933108-28933108 on Build GRCh38
rs772257899 Chr.16: 28933131-28933131 on Build GRCh38
rs775757103 Chr.16: 28933134-28933134 on Build GRCh38
rs749762747 Chr.16: 28933032-28933032 on Build GRCh38
rs775841308 Chr.16: 28933046-28933046 on Build GRCh38
rs772346128 Chr.16: 28933043-28933043 on Build GRCh38
rs779854329 Chr.16: 28933129-28933137 on Build GRCh38
rs776423905 Chr.16: 28933211-28933212 on Build GRCh38
rs181541269 Chr.16: 28933151-28933151 on Build GRCh38
rs761763925 Chr.16: 28933051-28933051 on Build GRCh38
rs189652694 Chr.16: 28933064-28933064 on Build GRCh38
rs754656596 Chr.16: 28933086-28933086 on Build GRCh38
rs1043892288 Chr.16: 28933072-28933072 on Build GRCh38
rs561660848 Chr.16: 28933042-28933042 on Build GRCh38

View Full Product Details