Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 116357295-116357544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC172
Entrez Gene ID
374355
Gene Name
coiled-coil domain containing 172
Gene Aliases
C10orf96
UniGene
Hs.233407
Gene Chromosome Location
Chr.10: 116324097-116380029 on Build GRCh38
Transcripts
NM_198515.2 XM_006717820.3 XM_011539771.2 XM_017016206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7278987 c.491T>G p.L164R Chr.10: 116357422-116357422 on Build GRCh38
4011905 c.545T>C p.L182P Chr.10: 116357476-116357476 on Build GRCh38
4550919 c.513G>A p.L171L Chr.10: 116357444-116357444 on Build GRCh38
333503 c.506G>T p.S169I Chr.10: 116357437-116357437 on Build GRCh38
73883 c.464A>G p.E155G Chr.10: 116357395-116357395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925784672 Chr.10: 116357478-116357478 on Build GRCh38
rs890154839 Chr.10: 116357451-116357451 on Build GRCh38
rs11197637 Chr.10: 116357325-116357325 on Build GRCh38
rs769284084 Chr.10: 116357476-116357476 on Build GRCh38
rs765985314 Chr.10: 116357463-116357463 on Build GRCh38
rs916937786 Chr.10: 116357431-116357432 on Build GRCh38
rs1029954003 Chr.10: 116357417-116357417 on Build GRCh38
rs146274660 Chr.10: 116357465-116357465 on Build GRCh38
rs1018235751 Chr.10: 116357481-116357481 on Build GRCh38
rs768483146 Chr.10: 116357424-116357424 on Build GRCh38
rs368285625 Chr.10: 116357488-116357488 on Build GRCh38
rs201416704 Chr.10: 116357475-116357475 on Build GRCh38
rs977144953 Chr.10: 116357475-116357475 on Build GRCh38
rs777937742 Chr.10: 116357369-116357369 on Build GRCh38
rs771583951 Chr.10: 116357443-116357443 on Build GRCh38
rs758620555 Chr.10: 116357368-116357368 on Build GRCh38
rs779931764 Chr.10: 116357398-116357398 on Build GRCh38
rs757110543 Chr.10: 116357407-116357407 on Build GRCh38
rs774220410 Chr.10: 116357429-116357429 on Build GRCh38
rs772656092 Chr.10: 116357457-116357457 on Build GRCh38
rs758921480 Chr.10: 116357495-116357495 on Build GRCh38
rs747247815 Chr.10: 116357386-116357386 on Build GRCh38
rs201464805 Chr.10: 116357424-116357429 on Build GRCh38
rs749079391 Chr.10: 116357400-116357400 on Build GRCh38
rs897558732 Chr.10: 116357326-116357326 on Build GRCh38
rs754107376 Chr.10: 116357334-116357334 on Build GRCh38
rs774789466 Chr.10: 116357346-116357346 on Build GRCh38
rs748011212 Chr.10: 116357431-116357431 on Build GRCh38
rs765633744 Chr.10: 116357482-116357482 on Build GRCh38
rs779334471 Chr.10: 116357366-116357366 on Build GRCh38
rs751651171 Chr.10: 116357510-116357510 on Build GRCh38
rs530526423 Chr.10: 116357349-116357349 on Build GRCh38
rs551806683 Chr.10: 116357391-116357391 on Build GRCh38
rs764291193 Chr.10: 116357509-116357509 on Build GRCh38

View Full Product Details