Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 722777-723048 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC78
Entrez Gene ID
124093
Gene Name
coiled-coil domain containing 78
Gene Aliases
C16orf25, CNM4, JFP10, hsCCDC78
UniGene
Hs.381943
Gene Chromosome Location
Chr.16: 722578-726880 on Build GRCh38
Transcripts
NM_001031737.2 XM_006720838.1 XM_006720843.3 XM_011522356.1 XM_011522357.1 XM_011522358.2 XM_011522359.1 XM_011522360.1 XM_011522361.1 XM_011522362.1 XM_011522363.1 XM_011522364.1 XM_011522365.1 XM_011522366.1 XM_011522367.1 XM_011522368.1 XM_011522369.1 XM_011522370.1 XM_011522371.2 XM_017022929.1 XM_017022930.1 XM_017022931.1 XM_017022932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4403320 c.1235G>A p.G412E Chr.16: 722984-722984 on Build GRCh38
4845007 c.1248G>A p.L416L Chr.16: 722971-722971 on Build GRCh38
6622444 c.1277C>T p.T426M Chr.16: 722942-722942 on Build GRCh38
5851525 c.1240C>T p.R414* Chr.16: 722979-722979 on Build GRCh38
5484056 c.1233C>T p.S411S Chr.16: 722986-722986 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749463945 Chr.16: 722935-722935 on Build GRCh38
rs770751341 Chr.16: 722997-722997 on Build GRCh38
rs754424286 Chr.16: 722818-722818 on Build GRCh38
rs191749939 Chr.16: 722988-722988 on Build GRCh38
rs55652059 Chr.16: 722848-722848 on Build GRCh38
rs778293175 Chr.16: 722892-722892 on Build GRCh38
rs35715104 Chr.16: 722958-722959 on Build GRCh38
rs142180051 Chr.16: 722942-722942 on Build GRCh38
rs374237828 Chr.16: 722808-722808 on Build GRCh38
rs773837439 Chr.16: 722945-722945 on Build GRCh38
rs534538485 Chr.16: 722981-722981 on Build GRCh38
rs777647838 Chr.16: 722918-722918 on Build GRCh38
rs761180576 Chr.16: 722946-722946 on Build GRCh38
rs747634588 Chr.16: 722904-722904 on Build GRCh38
rs766184705 Chr.16: 722885-722885 on Build GRCh38
rs774987057 Chr.16: 722862-722862 on Build GRCh38
rs558615304 Chr.16: 722944-722944 on Build GRCh38
rs762243012 Chr.16: 722865-722865 on Build GRCh38
rs893572894 Chr.16: 722990-722990 on Build GRCh38
rs764064368 Chr.16: 722861-722861 on Build GRCh38
rs190128477 Chr.16: 722867-722867 on Build GRCh38
rs746845893 Chr.16: 722925-722925 on Build GRCh38
rs141141442 Chr.16: 723003-723003 on Build GRCh38
rs774139482 Chr.16: 723024-723028 on Build GRCh38
rs771507948 Chr.16: 723009-723009 on Build GRCh38
rs563037365 Chr.16: 723010-723010 on Build GRCh38
rs763586611 Chr.16: 723023-723023 on Build GRCh38
rs768794447 Chr.16: 722993-722993 on Build GRCh38
rs780967551 Chr.16: 722986-722986 on Build GRCh38
rs758288338 Chr.16: 722912-722912 on Build GRCh38
rs548607734 Chr.16: 722868-722868 on Build GRCh38
rs369081589 Chr.16: 722982-722982 on Build GRCh38
rs780307486 Chr.16: 722930-722930 on Build GRCh38
rs763065715 Chr.16: 722852-722852 on Build GRCh38
rs200333284 Chr.16: 722879-722879 on Build GRCh38
rs760280521 Chr.16: 722948-722948 on Build GRCh38
rs374749282 Chr.16: 722947-722947 on Build GRCh38
rs755855095 Chr.16: 722813-722813 on Build GRCh38
rs148276717 Chr.16: 722985-722985 on Build GRCh38
rs752371476 Chr.16: 723013-723013 on Build GRCh38
rs201006875 Chr.16: 722797-722797 on Build GRCh38
rs759125788 Chr.16: 722950-722950 on Build GRCh38
rs762424779 Chr.16: 723019-723019 on Build GRCh38
rs778968209 Chr.16: 722831-722831 on Build GRCh38
rs977097602 Chr.16: 722821-722821 on Build GRCh38
rs752178658 Chr.16: 722966-722966 on Build GRCh38
rs941301878 Chr.16: 723016-723016 on Build GRCh38
rs368002114 Chr.16: 722849-722849 on Build GRCh38
rs867886720 Chr.16: 722987-722987 on Build GRCh38
rs773923207 Chr.16: 723004-723004 on Build GRCh38
rs12599130 Chr.16: 722842-722842 on Build GRCh38
rs560848393 Chr.16: 722814-722814 on Build GRCh38
rs779816790 Chr.16: 722814-722814 on Build GRCh38
rs770686722 Chr.16: 722927-722927 on Build GRCh38
rs764893097 Chr.16: 722964-722964 on Build GRCh38
rs768853325 Chr.16: 722940-722940 on Build GRCh38
rs183115411 Chr.16: 722941-722941 on Build GRCh38
rs748850336 Chr.16: 722815-722815 on Build GRCh38
rs779169574 Chr.16: 723000-723000 on Build GRCh38
rs146067716 Chr.16: 722978-722978 on Build GRCh38
rs754584711 Chr.16: 722889-722889 on Build GRCh38
rs1045075829 Chr.16: 722975-722975 on Build GRCh38

View Full Product Details