Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 54907685-54907958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC68
Entrez Gene ID
80323
Gene Name
coiled-coil domain containing 68
Gene Aliases
SE57-1
UniGene
Hs.120790
Gene Chromosome Location
Chr.18: 54901509-54959539 on Build GRCh38
Transcripts
NM_001143829.1 NM_025214.2 XM_006722552.3 XM_011526198.2 XM_011526199.2 XM_011526200.2 XM_011526201.2 XM_011526202.1 XM_011526203.2 XM_017026011.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
473915 c.947C>T p.T316I Chr.18: 54907789-54907789 on Build GRCh38
1711385 c.919A>G p.R307G Chr.18: 54907817-54907817 on Build GRCh38
4072604 c.939T>C p.A313A Chr.18: 54907797-54907797 on Build GRCh38
231974 c.917C>T p.P306L Chr.18: 54907819-54907819 on Build GRCh38
1290837 c.889A>T p.T297S Chr.18: 54907847-54907847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748770244 Chr.18: 54907835-54907835 on Build GRCh38
rs1034889994 Chr.18: 54907889-54907889 on Build GRCh38
rs763041193 Chr.18: 54907789-54907789 on Build GRCh38
rs770090312 Chr.18: 54907932-54907932 on Build GRCh38
rs757559610 Chr.18: 54907807-54907807 on Build GRCh38
rs369270622 Chr.18: 54907751-54907751 on Build GRCh38
rs749204089 Chr.18: 54907748-54907748 on Build GRCh38
rs781661670 Chr.18: 54907819-54907819 on Build GRCh38
rs771760142 Chr.18: 54907869-54907869 on Build GRCh38
rs1944382 Chr.18: 54907771-54907771 on Build GRCh38
rs372144883 Chr.18: 54907737-54907737 on Build GRCh38
rs374855422 Chr.18: 54907744-54907744 on Build GRCh38
rs374785146 Chr.18: 54907777-54907777 on Build GRCh38
rs773100165 Chr.18: 54907886-54907886 on Build GRCh38
rs750035338 Chr.18: 54907825-54907825 on Build GRCh38
rs770011082 Chr.18: 54907896-54907896 on Build GRCh38
rs779406081 Chr.18: 54907834-54907834 on Build GRCh38
rs779678441 Chr.18: 54907746-54907746 on Build GRCh38
rs1013643618 Chr.18: 54907916-54907916 on Build GRCh38
rs781047467 Chr.18: 54907837-54907837 on Build GRCh38
rs754543160 Chr.18: 54907844-54907844 on Build GRCh38
rs1052741373 Chr.18: 54907881-54907881 on Build GRCh38
rs373869734 Chr.18: 54907887-54907887 on Build GRCh38
rs772010430 Chr.18: 54907763-54907763 on Build GRCh38
rs1023330775 Chr.18: 54907931-54907931 on Build GRCh38
rs745543570 Chr.18: 54907743-54907743 on Build GRCh38
rs369026613 Chr.18: 54907824-54907824 on Build GRCh38
rs761638313 Chr.18: 54907757-54907757 on Build GRCh38
rs150354176 Chr.18: 54907828-54907828 on Build GRCh38
rs764543714 Chr.18: 54907792-54907792 on Build GRCh38
rs757699312 Chr.18: 54907871-54907872 on Build GRCh38
rs748159066 Chr.18: 54907868-54907868 on Build GRCh38
rs774463178 Chr.18: 54907753-54907753 on Build GRCh38
rs752065242 Chr.18: 54907797-54907797 on Build GRCh38
rs529492910 Chr.18: 54907882-54907882 on Build GRCh38
rs370614984 Chr.18: 54907790-54907790 on Build GRCh38
rs188349449 Chr.18: 54907783-54907783 on Build GRCh38
rs780641712 Chr.18: 54907739-54907739 on Build GRCh38
rs748519399 Chr.18: 54907906-54907906 on Build GRCh38
rs778940317 Chr.18: 54907851-54907851 on Build GRCh38
rs759869367 Chr.18: 54907772-54907772 on Build GRCh38
rs762952972 Chr.18: 54907913-54907913 on Build GRCh38
rs554606364 Chr.18: 54907774-54907774 on Build GRCh38

View Full Product Details