Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157300776-157301007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP2
Entrez Gene ID
26999
Gene Name
cytoplasmic FMR1 interacting protein 2
Gene Aliases
PIR121
UniGene
Hs.519702
Gene Chromosome Location
Chr.5: 157266080-157395598 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1

SNP Detail

SNP ID Chromosome Location
rs761214987 Chr.5: 157300906-157300906 on Build GRCh38
rs574422657 Chr.5: 157300914-157300914 on Build GRCh38
rs754888547 Chr.5: 157300931-157300931 on Build GRCh38
rs777261535 Chr.5: 157300903-157300903 on Build GRCh38
rs764849957 Chr.5: 157300908-157300908 on Build GRCh38
rs369062847 Chr.5: 157300861-157300861 on Build GRCh38
rs752733312 Chr.5: 157300941-157300941 on Build GRCh38
rs373151265 Chr.5: 157300867-157300867 on Build GRCh38
rs528604984 Chr.5: 157300826-157300826 on Build GRCh38
rs376458256 Chr.5: 157300913-157300913 on Build GRCh38
rs750010808 Chr.5: 157300911-157300911 on Build GRCh38
rs766221459 Chr.5: 157300921-157300921 on Build GRCh38
rs565299805 Chr.5: 157300904-157300904 on Build GRCh38
rs374139112 Chr.5: 157300933-157300933 on Build GRCh38
rs749327820 Chr.5: 157300820-157300820 on Build GRCh38
rs769152996 Chr.5: 157300890-157300890 on Build GRCh38
rs746144507 Chr.5: 157300831-157300831 on Build GRCh38
rs982800387 Chr.5: 157300949-157300949 on Build GRCh38
rs369872275 Chr.5: 157300824-157300824 on Build GRCh38
rs761109295 Chr.5: 157300888-157300888 on Build GRCh38
rs751405478 Chr.5: 157300925-157300925 on Build GRCh38
rs370788970 Chr.5: 157300920-157300920 on Build GRCh38

View Full Product Details