Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49792791-49793062 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDHR4
Entrez Gene ID
389118
Gene Name
cadherin related family member 4
Gene Aliases
CDH29, PRO34300
UniGene
Hs.363312
Gene Chromosome Location
Chr.3: 49790732-49802741 on Build GRCh38
Transcripts
NM_001007540.3 XM_011533700.2 XM_011533701.2 XM_017006366.1 XM_017006367.1 XM_017006368.1 XM_017006369.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4149962 c.1906A>C p.T636P Chr.3: 49792943-49792943 on Build GRCh38
4616006 c.1912delC p.H638fs*12 Chr.3: 49792937-49792937 on Build GRCh38
6836037 c.1897G>T p.G633C Chr.3: 49792952-49792952 on Build GRCh38
5680769 c.1961C>A p.A654D Chr.3: 49792888-49792888 on Build GRCh38
5420736 c.1839G>T p.G613G Chr.3: 49793010-49793010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754847832 Chr.3: 49792966-49792966 on Build GRCh38
rs747315447 Chr.3: 49792870-49792870 on Build GRCh38
rs1031474908 Chr.3: 49793007-49793007 on Build GRCh38
rs868083022 Chr.3: 49792845-49792845 on Build GRCh38
rs34160951 Chr.3: 49792992-49792992 on Build GRCh38
rs748212081 Chr.3: 49792955-49792955 on Build GRCh38
rs966761357 Chr.3: 49792978-49792978 on Build GRCh38
rs773130212 Chr.3: 49793022-49793022 on Build GRCh38
rs530718899 Chr.3: 49792997-49792997 on Build GRCh38
rs932849375 Chr.3: 49792937-49792937 on Build GRCh38
rs185204365 Chr.3: 49792823-49792823 on Build GRCh38
rs775872473 Chr.3: 49792829-49792829 on Build GRCh38
rs758445851 Chr.3: 49792947-49792947 on Build GRCh38
rs759524530 Chr.3: 49793037-49793037 on Build GRCh38
rs368736976 Chr.3: 49792928-49792931 on Build GRCh38
rs779968203 Chr.3: 49792958-49792958 on Build GRCh38
rs746748949 Chr.3: 49792959-49792959 on Build GRCh38
rs764175337 Chr.3: 49792909-49792909 on Build GRCh38
rs768920368 Chr.3: 49792937-49792937 on Build GRCh38
rs763221367 Chr.3: 49793017-49793017 on Build GRCh38
rs548223769 Chr.3: 49792942-49792942 on Build GRCh38
rs370105554 Chr.3: 49792895-49792895 on Build GRCh38
rs757219790 Chr.3: 49792939-49792939 on Build GRCh38
rs561228168 Chr.3: 49792894-49792894 on Build GRCh38
rs906716592 Chr.3: 49792825-49792825 on Build GRCh38

View Full Product Details