Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 120158015-120158286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL14A1
Entrez Gene ID
7373
Gene Name
collagen type XIV alpha 1 chain
Gene Aliases
UND
UniGene
Hs.409662
Gene Chromosome Location
Chr.8: 120124468-120373575 on Build GRCh38
Transcripts
NM_021110.3 XM_005251059.3 XM_006716651.3 XM_017013809.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1313678 c.192G>A p.V64V Chr.8: 120158233-120158233 on Build GRCh38
6530517 c.110G>C p.R37T Chr.8: 120158151-120158151 on Build GRCh38
4514025 c.95C>T p.P32L Chr.8: 120158136-120158136 on Build GRCh38
3896451 c.109A>C p.R37R Chr.8: 120158150-120158150 on Build GRCh38
3644657 c.125C>T p.S42F Chr.8: 120158166-120158166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753829521 Chr.8: 120158061-120158061 on Build GRCh38
rs772865732 Chr.8: 120158118-120158118 on Build GRCh38
rs867973354 Chr.8: 120158187-120158187 on Build GRCh38
rs748067894 Chr.8: 120158053-120158053 on Build GRCh38
rs908564065 Chr.8: 120158165-120158165 on Build GRCh38
rs746558424 Chr.8: 120158238-120158238 on Build GRCh38
rs374404737 Chr.8: 120158098-120158098 on Build GRCh38
rs370278346 Chr.8: 120158133-120158133 on Build GRCh38
rs377749294 Chr.8: 120158131-120158131 on Build GRCh38
rs756367159 Chr.8: 120158195-120158195 on Build GRCh38
rs770454652 Chr.8: 120158246-120158246 on Build GRCh38
rs865802009 Chr.8: 120158127-120158127 on Build GRCh38
rs765045965 Chr.8: 120158137-120158137 on Build GRCh38
rs776188502 Chr.8: 120158258-120158258 on Build GRCh38
rs749845782 Chr.8: 120158092-120158092 on Build GRCh38
rs933955124 Chr.8: 120158094-120158094 on Build GRCh38
rs1038124013 Chr.8: 120158041-120158041 on Build GRCh38
rs868262301 Chr.8: 120158205-120158205 on Build GRCh38
rs780590042 Chr.8: 120158201-120158201 on Build GRCh38
rs780749441 Chr.8: 120158083-120158083 on Build GRCh38
rs746461865 Chr.8: 120158123-120158123 on Build GRCh38
rs778308536 Chr.8: 120158080-120158080 on Build GRCh38
rs146249434 Chr.8: 120158188-120158188 on Build GRCh38
rs745555649 Chr.8: 120158259-120158259 on Build GRCh38
rs551300076 Chr.8: 120158162-120158162 on Build GRCh38
rs201022344 Chr.8: 120158154-120158154 on Build GRCh38
rs769240742 Chr.8: 120158100-120158100 on Build GRCh38
rs762891720 Chr.8: 120158141-120158141 on Build GRCh38
rs767679736 Chr.8: 120158176-120158176 on Build GRCh38
rs562262486 Chr.8: 120158134-120158134 on Build GRCh38
rs757342098 Chr.8: 120158170-120158170 on Build GRCh38
rs755550648 Chr.8: 120158228-120158228 on Build GRCh38
rs749616534 Chr.8: 120158216-120158216 on Build GRCh38
rs867059241 Chr.8: 120158108-120158108 on Build GRCh38

View Full Product Details