Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49323962-49324199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf71
Entrez Gene ID
118461
Gene Name
chromosome 10 open reading frame 71
Gene Aliases
-
UniGene
Hs.585480
Gene Chromosome Location
Chr.10: 49297009-49330145 on Build GRCh38
Transcripts
NM_001135196.1 XM_005269476.4 XM_005269477.3 XM_005269478.4 XM_017015619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
918626 c.1519C>T p.R507W Chr.10: 49324064-49324064 on Build GRCh38
5605571 c.1603C>G p.Q535E Chr.10: 49324148-49324148 on Build GRCh38
1297290 c.1521G>A p.R507R Chr.10: 49324066-49324066 on Build GRCh38
3438354 c.1647G>A p.E549E Chr.10: 49324192-49324192 on Build GRCh38
684153 c.1490C>A p.P497H Chr.10: 49324035-49324035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114069700 Chr.10: 49324167-49324167 on Build GRCh38
rs376302106 Chr.10: 49324084-49324084 on Build GRCh38
rs749103332 Chr.10: 49324042-49324042 on Build GRCh38
rs765766954 Chr.10: 49324081-49324081 on Build GRCh38
rs143544986 Chr.10: 49324159-49324159 on Build GRCh38
rs775096407 Chr.10: 49324136-49324136 on Build GRCh38
rs1057168979 Chr.10: 49324051-49324051 on Build GRCh38
rs763325120 Chr.10: 49324175-49324175 on Build GRCh38
rs749458818 Chr.10: 49324165-49324165 on Build GRCh38
rs201064215 Chr.10: 49323992-49323992 on Build GRCh38
rs923314523 Chr.10: 49324147-49324147 on Build GRCh38
rs772077867 Chr.10: 49324134-49324134 on Build GRCh38
rs568594834 Chr.10: 49324093-49324093 on Build GRCh38
rs181600390 Chr.10: 49324061-49324061 on Build GRCh38
rs750453040 Chr.10: 49324004-49324004 on Build GRCh38
rs777081340 Chr.10: 49324079-49324079 on Build GRCh38
rs61453891 Chr.10: 49324110-49324110 on Build GRCh38
rs770842902 Chr.10: 49324060-49324060 on Build GRCh38
rs778888907 Chr.10: 49324118-49324118 on Build GRCh38
rs755143970 Chr.10: 49324100-49324100 on Build GRCh38
rs751670172 Chr.10: 49324099-49324099 on Build GRCh38
rs902909346 Chr.10: 49323989-49323989 on Build GRCh38
rs781404544 Chr.10: 49324105-49324105 on Build GRCh38
rs766482465 Chr.10: 49324098-49324098 on Build GRCh38
rs377285836 Chr.10: 49324040-49324040 on Build GRCh38
rs550545516 Chr.10: 49324065-49324065 on Build GRCh38
rs760558125 Chr.10: 49324133-49324144 on Build GRCh38
rs762091617 Chr.10: 49324080-49324080 on Build GRCh38
rs200257260 Chr.10: 49324106-49324106 on Build GRCh38
rs758472555 Chr.10: 49324011-49324011 on Build GRCh38
rs775875033 Chr.10: 49324071-49324071 on Build GRCh38
rs746534437 Chr.10: 49324137-49324137 on Build GRCh38
rs1005768469 Chr.10: 49324088-49324088 on Build GRCh38
rs569703214 Chr.10: 49324030-49324030 on Build GRCh38
rs201677101 Chr.10: 49324070-49324070 on Build GRCh38
rs554031001 Chr.10: 49324064-49324064 on Build GRCh38
rs750947211 Chr.10: 49324083-49324083 on Build GRCh38
rs747276832 Chr.10: 49324032-49324032 on Build GRCh38
rs757737048 Chr.10: 49323986-49323986 on Build GRCh38
rs368697466 Chr.10: 49324119-49324119 on Build GRCh38
rs972167545 Chr.10: 49324120-49324120 on Build GRCh38

View Full Product Details