Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43305953-43306208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIP3
Entrez Gene ID
401262
Gene Name
cysteine rich protein 3
Gene Aliases
TLP, TLP-A, bA480N24.2
UniGene
Hs.653165
Gene Chromosome Location
Chr.6: 43305473-43310705 on Build GRCh38
Transcripts
NM_206922.2 XM_005249103.3 XM_011514609.2 XM_017010856.1 XM_017010857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6547277 c.537C>A p.Y179* Chr.6: 43306083-43306083 on Build GRCh38
7335788 c.562_563CC>TT p.P188F Chr.6: 43306057-43306058 on Build GRCh38
3627614 c.616G>A p.G206S Chr.6: 43306004-43306004 on Build GRCh38
4802403 c.539T>C p.L180P Chr.6: 43306081-43306081 on Build GRCh38
1079543 c.507C>T p.V169V Chr.6: 43306113-43306113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767743121 Chr.6: 43306041-43306041 on Build GRCh38
rs754990737 Chr.6: 43306059-43306059 on Build GRCh38
rs562856023 Chr.6: 43306050-43306050 on Build GRCh38
rs745539913 Chr.6: 43306148-43306148 on Build GRCh38
rs1018148108 Chr.6: 43306067-43306067 on Build GRCh38
rs748401564 Chr.6: 43306142-43306143 on Build GRCh38
rs762413810 Chr.6: 43306037-43306037 on Build GRCh38
rs775105917 Chr.6: 43306157-43306157 on Build GRCh38
rs777408235 Chr.6: 43306100-43306100 on Build GRCh38
rs774491969 Chr.6: 43306134-43306137 on Build GRCh38
rs756963871 Chr.6: 43306113-43306113 on Build GRCh38
rs1004761361 Chr.6: 43306088-43306088 on Build GRCh38
rs371312241 Chr.6: 43306095-43306095 on Build GRCh38
rs370582150 Chr.6: 43306048-43306048 on Build GRCh38
rs563318442 Chr.6: 43306151-43306151 on Build GRCh38
rs895215195 Chr.6: 43306028-43306028 on Build GRCh38
rs745436154 Chr.6: 43305993-43305993 on Build GRCh38
rs142092139 Chr.6: 43306093-43306093 on Build GRCh38
rs139840752 Chr.6: 43306055-43306055 on Build GRCh38
rs772696657 Chr.6: 43306176-43306176 on Build GRCh38
rs752684319 Chr.6: 43306135-43306135 on Build GRCh38
rs746878328 Chr.6: 43306107-43306107 on Build GRCh38
rs756050567 Chr.6: 43306003-43306004 on Build GRCh38
rs953096780 Chr.6: 43306131-43306131 on Build GRCh38
rs867455206 Chr.6: 43305985-43305985 on Build GRCh38
rs41275952 Chr.6: 43306145-43306145 on Build GRCh38
rs780923110 Chr.6: 43306128-43306128 on Build GRCh38
rs200169587 Chr.6: 43306045-43306045 on Build GRCh38
rs748585750 Chr.6: 43306168-43306168 on Build GRCh38
rs903709473 Chr.6: 43306101-43306101 on Build GRCh38
rs368241763 Chr.6: 43306046-43306046 on Build GRCh38
rs372092478 Chr.6: 43306023-43306023 on Build GRCh38
rs774673052 Chr.6: 43306018-43306018 on Build GRCh38
rs551756582 Chr.6: 43306133-43306133 on Build GRCh38
rs769444881 Chr.6: 43306011-43306011 on Build GRCh38
rs150965756 Chr.6: 43306089-43306089 on Build GRCh38
rs929665317 Chr.6: 43305977-43305977 on Build GRCh38
rs543114859 Chr.6: 43306051-43306051 on Build GRCh38
rs773447648 Chr.6: 43306178-43306178 on Build GRCh38
rs374665833 Chr.6: 43306064-43306064 on Build GRCh38
rs1051159689 Chr.6: 43306005-43306005 on Build GRCh38
rs749095914 Chr.6: 43306023-43306024 on Build GRCh38
rs779898788 Chr.6: 43306004-43306025 on Build GRCh38

View Full Product Details