Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32040094-32040331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP21A2
Entrez Gene ID
1589
Gene Name
cytochrome P450 family 21 subfamily A member 2
Gene Aliases
CA21H, CAH1, CPS1, CYP21, CYP21B, P450c21B
UniGene
Hs.654479
Gene Chromosome Location
Chr.6: 32038316-32041670 on Build GRCh38
Transcripts
NM_000500.7 NM_001128590.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1443458 c.844G>T p.V282L Chr.6: 32040110-32040110 on Build GRCh38
6421987 c.863A>T p.D288V Chr.6: 32040129-32040129 on Build GRCh38
5040907 c.909G>C p.W303C Chr.6: 32040175-32040175 on Build GRCh38
7420141 c.916G>T p.V306F Chr.6: 32040182-32040182 on Build GRCh38
3347672 c.939+5G>A p.? Chr.6: 32040210-32040210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770199817 Chr.6: 32040116-32040116 on Build GRCh38
rs6442 Chr.6: 32040216-32040216 on Build GRCh38
rs72552753 Chr.6: 32040207-32040207 on Build GRCh38
rs267606756 Chr.6: 32040186-32040187 on Build GRCh38
rs151344501 Chr.6: 32040143-32040143 on Build GRCh38
rs374872656 Chr.6: 32040217-32040217 on Build GRCh38
rs750984938 Chr.6: 32040257-32040257 on Build GRCh38
rs746303150 Chr.6: 32040185-32040185 on Build GRCh38
rs775304709 Chr.6: 32040231-32040231 on Build GRCh38
rs6444 Chr.6: 32040296-32040296 on Build GRCh38
rs763782360 Chr.6: 32040239-32040239 on Build GRCh38
rs780412947 Chr.6: 32040209-32040209 on Build GRCh38
rs764955614 Chr.6: 32040224-32040224 on Build GRCh38
rs369087577 Chr.6: 32040211-32040211 on Build GRCh38
rs536088585 Chr.6: 32040200-32040200 on Build GRCh38
rs766779361 Chr.6: 32040237-32040237 on Build GRCh38
rs568758408 Chr.6: 32040183-32040183 on Build GRCh38
rs1027284604 Chr.6: 32040196-32040196 on Build GRCh38
rs961990168 Chr.6: 32040181-32040181 on Build GRCh38
rs6471 Chr.6: 32040110-32040110 on Build GRCh38
rs759308952 Chr.6: 32040152-32040152 on Build GRCh38
rs767069613 Chr.6: 32040256-32040256 on Build GRCh38
rs570785206 Chr.6: 32040122-32040122 on Build GRCh38
rs6443 Chr.6: 32040291-32040291 on Build GRCh38
rs994764994 Chr.6: 32040188-32040188 on Build GRCh38
rs765001985 Chr.6: 32040167-32040167 on Build GRCh38
rs606231200 Chr.6: 32040187-32040188 on Build GRCh38
rs759398534 Chr.6: 32040223-32040223 on Build GRCh38
rs774336886 Chr.6: 32040215-32040215 on Build GRCh38
rs770059546 Chr.6: 32040197-32040197 on Build GRCh38
rs537111179 Chr.6: 32040233-32040233 on Build GRCh38
rs557564791 Chr.6: 32040210-32040210 on Build GRCh38
rs775570889 Chr.6: 32040119-32040119 on Build GRCh38
rs397515532 Chr.6: 32040189-32040190 on Build GRCh38
rs201552310 Chr.6: 32040140-32040140 on Build GRCh38
rs752295510 Chr.6: 32040168-32040168 on Build GRCh38
rs758010702 Chr.6: 32040169-32040169 on Build GRCh38
rs751174502 Chr.6: 32040249-32040249 on Build GRCh38
rs776542176 Chr.6: 32040220-32040220 on Build GRCh38
rs777168794 Chr.6: 32040175-32040175 on Build GRCh38
rs151344505 Chr.6: 32040179-32040179 on Build GRCh38
rs768635138 Chr.6: 32040131-32040131 on Build GRCh38
rs991620424 Chr.6: 32040205-32040205 on Build GRCh38
rs143877763 Chr.6: 32040139-32040139 on Build GRCh38
rs753678267 Chr.6: 32040151-32040151 on Build GRCh38
rs148622274 Chr.6: 32040149-32040149 on Build GRCh38
rs756824528 Chr.6: 32040255-32040255 on Build GRCh38
rs751072381 Chr.6: 32040178-32040178 on Build GRCh38

View Full Product Details