Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31492284-31492517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf58
Entrez Gene ID
64755
Gene Name
chromosome 16 open reading frame 58
Gene Aliases
RUS
UniGene
Hs.499102
Gene Chromosome Location
Chr.16: 31489464-31508419 on Build GRCh38
Transcripts
NM_022744.3 XM_017023570.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6203666 c.1088-8C>G p.? Chr.16: 31492348-31492348 on Build GRCh38
1609248 c.1102_1110delGTTCTGAAC p.V368_N370delVLN Chr.16: 31492318-31492326 on Build GRCh38
6614505 c.1141G>A p.A381T Chr.16: 31492287-31492287 on Build GRCh38
3509287 c.1094T>C p.V365A Chr.16: 31492334-31492334 on Build GRCh38
970310 c.1144G>A p.A382T Chr.16: 31492284-31492284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200612071 Chr.16: 31492373-31492373 on Build GRCh38
rs751398606 Chr.16: 31492377-31492377 on Build GRCh38
rs1001041350 Chr.16: 31492317-31492317 on Build GRCh38
rs775899419 Chr.16: 31492313-31492313 on Build GRCh38
rs760909745 Chr.16: 31492318-31492318 on Build GRCh38
rs199591213 Chr.16: 31492310-31492310 on Build GRCh38
rs368694327 Chr.16: 31492312-31492312 on Build GRCh38
rs781000937 Chr.16: 31492382-31492382 on Build GRCh38
rs937604206 Chr.16: 31492467-31492467 on Build GRCh38
rs765301117 Chr.16: 31492367-31492367 on Build GRCh38
rs769367407 Chr.16: 31492303-31492303 on Build GRCh38
rs779566850 Chr.16: 31492304-31492304 on Build GRCh38
rs750388463 Chr.16: 31492368-31492368 on Build GRCh38
rs947463625 Chr.16: 31492408-31492408 on Build GRCh38
rs577507252 Chr.16: 31492341-31492341 on Build GRCh38
rs752327291 Chr.16: 31492383-31492383 on Build GRCh38
rs892116245 Chr.16: 31492412-31492412 on Build GRCh38
rs575313022 Chr.16: 31492407-31492407 on Build GRCh38
rs927318027 Chr.16: 31492466-31492466 on Build GRCh38
rs980598654 Chr.16: 31492479-31492479 on Build GRCh38
rs373658198 Chr.16: 31492380-31492380 on Build GRCh38
rs755752123 Chr.16: 31492385-31492385 on Build GRCh38
rs201491629 Chr.16: 31492369-31492369 on Build GRCh38
rs1009210290 Chr.16: 31492425-31492425 on Build GRCh38
rs970655051 Chr.16: 31492447-31492447 on Build GRCh38
rs768889568 Chr.16: 31492323-31492323 on Build GRCh38
rs369485430 Chr.16: 31492365-31492365 on Build GRCh38
rs4536493 Chr.16: 31492430-31492430 on Build GRCh38
rs541989256 Chr.16: 31492463-31492463 on Build GRCh38

View Full Product Details