Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37566528-37566775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42EP1
Entrez Gene ID
11135
Gene Name
CDC42 effector protein 1
Gene Aliases
BORG5, CEP1, MSE55
UniGene
Hs.225356
Gene Chromosome Location
Chr.22: 37559778-37569405 on Build GRCh38
Transcripts
NM_152243.2 XM_006724107.1 XM_006724108.1 XM_006724109.1 XM_017028541.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5981847 c.423C>A p.D141E Chr.22: 37566772-37566772 on Build GRCh38
3554186 c.243C>T p.F81F Chr.22: 37566592-37566592 on Build GRCh38
1416147 c.282_283insC p.R97fs*78 Chr.22: 37566631-37566632 on Build GRCh38
1416146 c.227G>A p.R76H Chr.22: 37566576-37566576 on Build GRCh38
5974375 c.213C>T p.S71S Chr.22: 37566562-37566562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748248737 Chr.22: 37566551-37566551 on Build GRCh38
rs140945384 Chr.22: 37566628-37566628 on Build GRCh38
rs768436129 Chr.22: 37566686-37566686 on Build GRCh38
rs1011747026 Chr.22: 37566630-37566630 on Build GRCh38
rs995090347 Chr.22: 37566604-37566604 on Build GRCh38
rs370072833 Chr.22: 37566623-37566623 on Build GRCh38
rs111307192 Chr.22: 37566733-37566733 on Build GRCh38
rs899369484 Chr.22: 37566689-37566689 on Build GRCh38
rs774081807 Chr.22: 37566748-37566748 on Build GRCh38
rs376901875 Chr.22: 37566734-37566734 on Build GRCh38
rs138108680 Chr.22: 37566749-37566749 on Build GRCh38
rs747116423 Chr.22: 37566618-37566618 on Build GRCh38
rs757940883 Chr.22: 37566582-37566582 on Build GRCh38
rs969629486 Chr.22: 37566682-37566682 on Build GRCh38
rs376654024 Chr.22: 37566593-37566593 on Build GRCh38
rs773216227 Chr.22: 37566555-37566555 on Build GRCh38
rs201283947 Chr.22: 37566635-37566635 on Build GRCh38
rs956831361 Chr.22: 37566625-37566625 on Build GRCh38
rs763778405 Chr.22: 37566584-37566584 on Build GRCh38
rs748423187 Chr.22: 37566605-37566605 on Build GRCh38
rs201030338 Chr.22: 37566709-37566709 on Build GRCh38
rs751855498 Chr.22: 37566728-37566728 on Build GRCh38
rs61739954 Chr.22: 37566576-37566576 on Build GRCh38
rs200018136 Chr.22: 37566636-37566636 on Build GRCh38
rs758694944 Chr.22: 37566718-37566718 on Build GRCh38
rs773879023 Chr.22: 37566688-37566688 on Build GRCh38
rs73418177 Chr.22: 37566550-37566550 on Build GRCh38
rs199905468 Chr.22: 37566662-37566662 on Build GRCh38
rs369211517 Chr.22: 37566585-37566585 on Build GRCh38
rs991988332 Chr.22: 37566560-37566560 on Build GRCh38
rs762710520 Chr.22: 37566683-37566683 on Build GRCh38
rs571270968 Chr.22: 37566669-37566669 on Build GRCh38
rs769657329 Chr.22: 37566677-37566677 on Build GRCh38
rs202138466 Chr.22: 37566703-37566703 on Build GRCh38
rs759701710 Chr.22: 37566740-37566740 on Build GRCh38
rs953392403 Chr.22: 37566610-37566610 on Build GRCh38
rs765059393 Chr.22: 37566567-37566567 on Build GRCh38
rs139353065 Chr.22: 37566639-37566639 on Build GRCh38
rs765724113 Chr.22: 37566705-37566705 on Build GRCh38
rs771780348 Chr.22: 37566753-37566753 on Build GRCh38
rs757605019 Chr.22: 37566730-37566730 on Build GRCh38
rs774440829 Chr.22: 37566631-37566632 on Build GRCh38
rs772224448 Chr.22: 37566607-37566607 on Build GRCh38
rs751206521 Chr.22: 37566632-37566632 on Build GRCh38
rs371962479 Chr.22: 37566638-37566638 on Build GRCh38
rs771080796 Chr.22: 37566621-37566621 on Build GRCh38
rs767836939 Chr.22: 37566680-37566703 on Build GRCh38
rs769736700 Chr.22: 37566626-37566626 on Build GRCh38
rs779105093 Chr.22: 37566597-37566597 on Build GRCh38
rs148893156 Chr.22: 37566692-37566692 on Build GRCh38
rs781374875 Chr.22: 37566671-37566671 on Build GRCh38
rs373084609 Chr.22: 37566670-37566670 on Build GRCh38
rs758778225 Chr.22: 37566655-37566655 on Build GRCh38
rs775490487 Chr.22: 37566678-37566678 on Build GRCh38
rs778052874 Chr.22: 37566617-37566617 on Build GRCh38
rs149573903 Chr.22: 37566559-37566559 on Build GRCh38
rs762035931 Chr.22: 37566632-37566632 on Build GRCh38
rs1019471800 Chr.22: 37566623-37566624 on Build GRCh38
rs764483507 Chr.22: 37566719-37566719 on Build GRCh38
rs150186371 Chr.22: 37566631-37566631 on Build GRCh38
rs916954381 Chr.22: 37566658-37566658 on Build GRCh38
rs745922297 Chr.22: 37566676-37566676 on Build GRCh38
rs759509615 Chr.22: 37566624-37566624 on Build GRCh38
rs528047239 Chr.22: 37566704-37566704 on Build GRCh38
rs775196114 Chr.22: 37566627-37566627 on Build GRCh38
rs373648442 Chr.22: 37566586-37566586 on Build GRCh38
rs145680167 Chr.22: 37566563-37566563 on Build GRCh38
rs1036124428 Chr.22: 37566654-37566654 on Build GRCh38
rs754285888 Chr.22: 37566587-37566587 on Build GRCh38
rs760780041 Chr.22: 37566556-37566556 on Build GRCh38
rs947968701 Chr.22: 37566575-37566575 on Build GRCh38
rs776700659 Chr.22: 37566562-37566562 on Build GRCh38

View Full Product Details