Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36412957-36413226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAF1B
Entrez Gene ID
8208
Gene Name
chromatin assembly factor 1 subunit B
Gene Aliases
CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7
UniGene
Hs.75238
Gene Chromosome Location
Chr.21: 36380145-36416827 on Build GRCh38
Transcripts
NM_005441.2 XM_011529755.2 XM_017028477.1 XM_017028478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1713971 c.1256C>T p.T419I Chr.21: 36413078-36413078 on Build GRCh38
3550544 c.1152G>A p.L384L Chr.21: 36412974-36412974 on Build GRCh38
1200934 c.1295C>T p.T432M Chr.21: 36413117-36413117 on Build GRCh38
1723512 c.1376C>T p.P459L Chr.21: 36413198-36413198 on Build GRCh38
7159068 c.1301C>G p.P434R Chr.21: 36413123-36413123 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989630220 Chr.21: 36413101-36413101 on Build GRCh38
rs968847405 Chr.21: 36413129-36413129 on Build GRCh38
rs763354487 Chr.21: 36413146-36413146 on Build GRCh38
rs759822351 Chr.21: 36413172-36413172 on Build GRCh38
rs756979003 Chr.21: 36413197-36413197 on Build GRCh38
rs566995930 Chr.21: 36413125-36413125 on Build GRCh38
rs752859731 Chr.21: 36413187-36413187 on Build GRCh38
rs201581262 Chr.21: 36413080-36413080 on Build GRCh38
rs748125049 Chr.21: 36413018-36413018 on Build GRCh38
rs958641879 Chr.21: 36412996-36412996 on Build GRCh38
rs756889046 Chr.21: 36413113-36413113 on Build GRCh38
rs745523995 Chr.21: 36413117-36413117 on Build GRCh38
rs754955180 Chr.21: 36412998-36412998 on Build GRCh38
rs776964270 Chr.21: 36413057-36413057 on Build GRCh38
rs760502373 Chr.21: 36413190-36413190 on Build GRCh38
rs771435098 Chr.21: 36413118-36413118 on Build GRCh38
rs771625691 Chr.21: 36413053-36413053 on Build GRCh38
rs139594054 Chr.21: 36412995-36412995 on Build GRCh38
rs774679537 Chr.21: 36413164-36413164 on Build GRCh38
rs751847865 Chr.21: 36413077-36413077 on Build GRCh38
rs767663024 Chr.21: 36413097-36413097 on Build GRCh38
rs955157014 Chr.21: 36413183-36413183 on Build GRCh38
rs768948362 Chr.21: 36413084-36413101 on Build GRCh38
rs759297500 Chr.21: 36413100-36413100 on Build GRCh38
rs777396815 Chr.21: 36413104-36413104 on Build GRCh38
rs767469369 Chr.21: 36413174-36413174 on Build GRCh38
rs1010287406 Chr.21: 36413121-36413121 on Build GRCh38
rs1012811346 Chr.21: 36413005-36413005 on Build GRCh38
rs777642815 Chr.21: 36413033-36413033 on Build GRCh38
rs1036027346 Chr.21: 36413153-36413153 on Build GRCh38
rs760522489 Chr.21: 36413188-36413188 on Build GRCh38
rs764075222 Chr.21: 36413189-36413189 on Build GRCh38
rs147369174 Chr.21: 36412986-36412986 on Build GRCh38
rs780937851 Chr.21: 36413014-36413014 on Build GRCh38
rs975332177 Chr.21: 36413163-36413163 on Build GRCh38
rs376013076 Chr.21: 36413067-36413067 on Build GRCh38
rs748914337 Chr.21: 36413038-36413038 on Build GRCh38
rs115050809 Chr.21: 36413066-36413066 on Build GRCh38
rs866013286 Chr.21: 36413135-36413135 on Build GRCh38
rs766768506 Chr.21: 36413075-36413075 on Build GRCh38
rs773845071 Chr.21: 36413051-36413051 on Build GRCh38
rs376201329 Chr.21: 36413093-36413093 on Build GRCh38
rs368621019 Chr.21: 36413204-36413204 on Build GRCh38
rs142952612 Chr.21: 36413124-36413124 on Build GRCh38
rs770607149 Chr.21: 36413042-36413042 on Build GRCh38
rs771430207 Chr.21: 36413150-36413150 on Build GRCh38
rs755821149 Chr.21: 36413022-36413022 on Build GRCh38
rs762405550 Chr.21: 36413065-36413065 on Build GRCh38
rs749146474 Chr.21: 36413112-36413112 on Build GRCh38
rs148582638 Chr.21: 36413102-36413102 on Build GRCh38
rs369251042 Chr.21: 36413088-36413088 on Build GRCh38
rs745438796 Chr.21: 36413052-36413052 on Build GRCh38
rs201486922 Chr.21: 36413205-36413205 on Build GRCh38
rs535924263 Chr.21: 36413134-36413134 on Build GRCh38
rs375927375 Chr.21: 36413081-36413081 on Build GRCh38

View Full Product Details