Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 130184141-130184366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHSY3
Entrez Gene ID
337876
Gene Name
chondroitin sulfate synthase 3
Gene Aliases
CHSY2, CSS3
UniGene
Hs.213137
Gene Chromosome Location
Chr.5: 129903415-130186637 on Build GRCh38
Transcripts
NM_175856.4 XM_011543363.2 XM_011543364.2 XM_011543365.2 XM_017009434.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
362365 c.1216C>A p.H406N Chr.5: 130184358-130184358 on Build GRCh38
6900650 c.1177C>T p.L393F Chr.5: 130184319-130184319 on Build GRCh38
1432929 c.1130G>T p.G377V Chr.5: 130184272-130184272 on Build GRCh38
448534 c.1192A>T p.R398W Chr.5: 130184334-130184334 on Build GRCh38
5867227 c.1141G>A p.D381N Chr.5: 130184283-130184283 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746317547 Chr.5: 130184179-130184179 on Build GRCh38
rs78391857 Chr.5: 130184297-130184297 on Build GRCh38
rs10066410 Chr.5: 130184181-130184181 on Build GRCh38
rs963488186 Chr.5: 130184313-130184313 on Build GRCh38
rs778495962 Chr.5: 130184293-130184293 on Build GRCh38
rs147857014 Chr.5: 130184305-130184305 on Build GRCh38
rs959475642 Chr.5: 130184266-130184266 on Build GRCh38
rs1033562678 Chr.5: 130184265-130184265 on Build GRCh38
rs967993676 Chr.5: 130184240-130184240 on Build GRCh38
rs765897274 Chr.5: 130184263-130184263 on Build GRCh38
rs775669247 Chr.5: 130184202-130184202 on Build GRCh38
rs144535982 Chr.5: 130184280-130184280 on Build GRCh38
rs770373198 Chr.5: 130184321-130184321 on Build GRCh38
rs764267674 Chr.5: 130184213-130184213 on Build GRCh38
rs549621285 Chr.5: 130184214-130184214 on Build GRCh38
rs141425789 Chr.5: 130184308-130184308 on Build GRCh38
rs750368142 Chr.5: 130184249-130184249 on Build GRCh38
rs567825362 Chr.5: 130184261-130184261 on Build GRCh38
rs908546652 Chr.5: 130184330-130184330 on Build GRCh38
rs148654806 Chr.5: 130184235-130184235 on Build GRCh38
rs753337951 Chr.5: 130184270-130184270 on Build GRCh38
rs751880121 Chr.5: 130184206-130184207 on Build GRCh38
rs747822104 Chr.5: 130184294-130184294 on Build GRCh38
rs999081268 Chr.5: 130184246-130184246 on Build GRCh38
rs768771576 Chr.5: 130184339-130184339 on Build GRCh38
rs756006904 Chr.5: 130184251-130184251 on Build GRCh38
rs758559327 Chr.5: 130184204-130184204 on Build GRCh38
rs757707487 Chr.5: 130184302-130184302 on Build GRCh38
rs776038578 Chr.5: 130184325-130184325 on Build GRCh38
rs749363556 Chr.5: 130184327-130184327 on Build GRCh38
rs902190145 Chr.5: 130184243-130184243 on Build GRCh38

View Full Product Details