Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40983248-40983495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTPS1
Entrez Gene ID
1503
Gene Name
CTP synthase 1
Gene Aliases
CTPS, IMD24
UniGene
Hs.473087
Gene Chromosome Location
Chr.1: 40979299-41012565 on Build GRCh38
Transcripts
NM_001905.3 NR_125440.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419221 c.44G>A p.G15E Chr.1: 40983334-40983334 on Build GRCh38
4426419 c.50G>T p.G17V Chr.1: 40983340-40983340 on Build GRCh38
7121208 c.152C>A p.S51Y Chr.1: 40983442-40983442 on Build GRCh38
4426418 c.50G>T p.G17V Chr.1: 40983340-40983340 on Build GRCh38
3419222 c.44G>A p.G15E Chr.1: 40983334-40983334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757447163 Chr.1: 40983427-40983427 on Build GRCh38
rs1020690759 Chr.1: 40983355-40983355 on Build GRCh38
rs767820852 Chr.1: 40983308-40983308 on Build GRCh38
rs779248936 Chr.1: 40983446-40983446 on Build GRCh38
rs759914190 Chr.1: 40983303-40983303 on Build GRCh38
rs868098496 Chr.1: 40983328-40983328 on Build GRCh38
rs769386291 Chr.1: 40983296-40983296 on Build GRCh38
rs757354207 Chr.1: 40983281-40983282 on Build GRCh38
rs545128484 Chr.1: 40983348-40983348 on Build GRCh38
rs761020732 Chr.1: 40983380-40983380 on Build GRCh38
rs772880925 Chr.1: 40983297-40983297 on Build GRCh38
rs756725880 Chr.1: 40983364-40983364 on Build GRCh38
rs764713994 Chr.1: 40983383-40983383 on Build GRCh38
rs367595948 Chr.1: 40983280-40983280 on Build GRCh38
rs753922614 Chr.1: 40983422-40983422 on Build GRCh38

View Full Product Details