Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 4428805-4429064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHAF1A
Entrez Gene ID
10036
Gene Name
chromatin assembly factor 1 subunit A
Gene Aliases
CAF-1, CAF1, CAF1B, CAF1P150, P150
UniGene
Hs.79018
Gene Chromosome Location
Chr.19: 4402596-4448322 on Build GRCh38
Transcripts
NM_005483.2 XM_011527605.2 XM_011527606.2 XM_011527607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5623342 c.1583G>A p.R528Q Chr.19: 4428869-4428869 on Build GRCh38
1711628 c.1550C>T p.P517L Chr.19: 4428836-4428836 on Build GRCh38
70166 c.1544G>A p.R515Q Chr.19: 4428830-4428830 on Build GRCh38
4079003 c.1576T>C p.S526P Chr.19: 4428862-4428862 on Build GRCh38
3797195 c.1536C>G p.L512L Chr.19: 4428822-4428822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758799950 Chr.19: 4428855-4428855 on Build GRCh38
rs922268599 Chr.19: 4429044-4429044 on Build GRCh38
rs1025597434 Chr.19: 4429005-4429005 on Build GRCh38
rs200533816 Chr.19: 4428859-4428859 on Build GRCh38
rs1017355485 Chr.19: 4428948-4428948 on Build GRCh38
rs755489585 Chr.19: 4428864-4428864 on Build GRCh38
rs62130978 Chr.19: 4429010-4429010 on Build GRCh38
rs1042797056 Chr.19: 4429000-4429000 on Build GRCh38
rs190635280 Chr.19: 4429033-4429033 on Build GRCh38
rs781268125 Chr.19: 4428865-4428865 on Build GRCh38
rs549885781 Chr.19: 4428963-4428963 on Build GRCh38
rs753288525 Chr.19: 4428830-4428830 on Build GRCh38
rs548926211 Chr.19: 4428987-4428987 on Build GRCh38
rs777908696 Chr.19: 4428901-4428901 on Build GRCh38
rs772063695 Chr.19: 4428910-4428910 on Build GRCh38
rs374387229 Chr.19: 4429026-4429026 on Build GRCh38
rs185776398 Chr.19: 4429011-4429011 on Build GRCh38
rs762781914 Chr.19: 4428901-4428903 on Build GRCh38
rs756891683 Chr.19: 4428846-4428846 on Build GRCh38
rs144320472 Chr.19: 4428863-4428863 on Build GRCh38
rs768505753 Chr.19: 4428934-4428934 on Build GRCh38
rs148041261 Chr.19: 4429024-4429024 on Build GRCh38
rs373495353 Chr.19: 4428840-4428840 on Build GRCh38
rs755864927 Chr.19: 4428951-4428951 on Build GRCh38
rs910999517 Chr.19: 4428877-4428877 on Build GRCh38
rs1040700537 Chr.19: 4428923-4428923 on Build GRCh38
rs374801348 Chr.19: 4428909-4428909 on Build GRCh38
rs376393055 Chr.19: 4428929-4428929 on Build GRCh38
rs534602987 Chr.19: 4428992-4428992 on Build GRCh38
rs528900843 Chr.19: 4428986-4428986 on Build GRCh38
rs890376410 Chr.19: 4429025-4429025 on Build GRCh38
rs34219547 Chr.19: 4428852-4428852 on Build GRCh38
rs1035698602 Chr.19: 4428990-4428990 on Build GRCh38
rs770012981 Chr.19: 4428869-4428869 on Build GRCh38
rs557573700 Chr.19: 4428997-4428997 on Build GRCh38
rs910934930 Chr.19: 4429012-4429012 on Build GRCh38
rs750776567 Chr.19: 4428854-4428854 on Build GRCh38
rs760619745 Chr.19: 4428930-4428930 on Build GRCh38
rs983479688 Chr.19: 4428984-4428984 on Build GRCh38
rs774005633 Chr.19: 4429042-4429058 on Build GRCh38
rs964116656 Chr.19: 4428961-4428961 on Build GRCh38
rs763636448 Chr.19: 4428828-4428828 on Build GRCh38
rs565829724 Chr.19: 4428964-4428964 on Build GRCh38
rs769099831 Chr.19: 4429008-4429008 on Build GRCh38
rs758859910 Chr.19: 4428970-4428970 on Build GRCh38
rs752206987 Chr.19: 4428868-4428868 on Build GRCh38
rs942337796 Chr.19: 4428906-4428906 on Build GRCh38
rs181359600 Chr.19: 4428988-4428988 on Build GRCh38
rs963837448 Chr.19: 4429019-4429019 on Build GRCh38

View Full Product Details