Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78280084-78280309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA4
Entrez Gene ID
55466
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
Gene Aliases
MST104, MSTP104, PRO1472
UniGene
Hs.513053
Gene Chromosome Location
Chr.15: 78264145-78282196 on Build GRCh38
Transcripts
NM_001130182.1 NM_001130183.1 NM_018602.3 XM_005254535.3 XM_006720601.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6234916 c.938delT p.P314fs*13 Chr.15: 78280285-78280285 on Build GRCh38
6885812 c.988C>T p.P330S Chr.15: 78280254-78280254 on Build GRCh38
6021371 c.1018C>T p.L340F Chr.15: 78280284-78280284 on Build GRCh38
5761391 c.954A>G p.K318K Chr.15: 78280121-78280121 on Build GRCh38
6885813 c.907C>T p.P303S Chr.15: 78280254-78280254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367669738 Chr.15: 78280155-78280155 on Build GRCh38
rs765335788 Chr.15: 78280274-78280274 on Build GRCh38
rs888729730 Chr.15: 78280242-78280242 on Build GRCh38
rs763055743 Chr.15: 78280287-78280287 on Build GRCh38
rs752673309 Chr.15: 78280277-78280277 on Build GRCh38
rs760454951 Chr.15: 78280233-78280233 on Build GRCh38
rs537704664 Chr.15: 78280165-78280165 on Build GRCh38
rs745622164 Chr.15: 78280164-78280165 on Build GRCh38
rs780201667 Chr.15: 78280182-78280182 on Build GRCh38
rs367900015 Chr.15: 78280184-78280184 on Build GRCh38
rs377049810 Chr.15: 78280214-78280214 on Build GRCh38
rs559035230 Chr.15: 78280203-78280203 on Build GRCh38
rs776855714 Chr.15: 78280253-78280253 on Build GRCh38
rs376841411 Chr.15: 78280170-78280170 on Build GRCh38
rs759691137 Chr.15: 78280261-78280261 on Build GRCh38
rs568522577 Chr.15: 78280138-78280138 on Build GRCh38
rs1036083655 Chr.15: 78280181-78280181 on Build GRCh38
rs150822161 Chr.15: 78280135-78280135 on Build GRCh38
rs759674662 Chr.15: 78280157-78280157 on Build GRCh38
rs766631828 Chr.15: 78280152-78280152 on Build GRCh38
rs778523363 Chr.15: 78280201-78280201 on Build GRCh38
rs371265771 Chr.15: 78280158-78280158 on Build GRCh38
rs772050961 Chr.15: 78280241-78280241 on Build GRCh38
rs910362803 Chr.15: 78280284-78280284 on Build GRCh38
rs369988433 Chr.15: 78280215-78280215 on Build GRCh38
rs202244455 Chr.15: 78280245-78280245 on Build GRCh38
rs757636547 Chr.15: 78280179-78280179 on Build GRCh38
rs751907163 Chr.15: 78280176-78280176 on Build GRCh38
rs768282373 Chr.15: 78280186-78280186 on Build GRCh38
rs944480343 Chr.15: 78280167-78280167 on Build GRCh38
rs772832121 Chr.15: 78280109-78280109 on Build GRCh38

View Full Product Details