Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73200979-73201252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
UniGene
Hs.103555
Gene Chromosome Location
Chr.17: 73193034-73208507 on Build GRCh38
Transcripts
NM_018714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7257086 c.1335G>C p.L445F Chr.17: 73201162-73201162 on Build GRCh38
6991564 c.1317T>C p.S439S Chr.17: 73201144-73201144 on Build GRCh38
5387191 c.1282-1G>A p.? Chr.17: 73201108-73201108 on Build GRCh38
3755789 c.1357G>C p.E453Q Chr.17: 73201184-73201184 on Build GRCh38
1179553 c.1348C>T p.Q450* Chr.17: 73201175-73201175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775282875 Chr.17: 73201111-73201111 on Build GRCh38
rs778207262 Chr.17: 73201075-73201075 on Build GRCh38
rs140102593 Chr.17: 73201142-73201142 on Build GRCh38
rs779485368 Chr.17: 73201187-73201187 on Build GRCh38
rs767686661 Chr.17: 73201143-73201143 on Build GRCh38
rs752942423 Chr.17: 73201146-73201146 on Build GRCh38
rs756380726 Chr.17: 73201172-73201172 on Build GRCh38
rs745305711 Chr.17: 73201192-73201192 on Build GRCh38
rs374059941 Chr.17: 73201006-73201006 on Build GRCh38
rs951443945 Chr.17: 73201189-73201189 on Build GRCh38
rs193921130 Chr.17: 73201175-73201175 on Build GRCh38
rs749722293 Chr.17: 73201182-73201182 on Build GRCh38
rs185537858 Chr.17: 73201038-73201038 on Build GRCh38
rs62621249 Chr.17: 73201184-73201184 on Build GRCh38
rs935124894 Chr.17: 73201047-73201047 on Build GRCh38
rs749775763 Chr.17: 73201087-73201087 on Build GRCh38
rs771511506 Chr.17: 73201101-73201101 on Build GRCh38
rs75558335 Chr.17: 73201045-73201045 on Build GRCh38
rs751375454 Chr.17: 73201097-73201097 on Build GRCh38
rs192967636 Chr.17: 73201023-73201023 on Build GRCh38
rs1052606294 Chr.17: 73201051-73201051 on Build GRCh38
rs764002722 Chr.17: 73201124-73201124 on Build GRCh38
rs188853520 Chr.17: 73201106-73201106 on Build GRCh38
rs1008905656 Chr.17: 73201094-73201094 on Build GRCh38
rs761623437 Chr.17: 73201136-73201136 on Build GRCh38
rs143699348 Chr.17: 73201177-73201177 on Build GRCh38
rs775233683 Chr.17: 73201205-73201205 on Build GRCh38
rs140675600 Chr.17: 73201194-73201194 on Build GRCh38
rs763976755 Chr.17: 73201110-73201111 on Build GRCh38
rs760505703 Chr.17: 73201118-73201118 on Build GRCh38
rs768462230 Chr.17: 73201222-73201222 on Build GRCh38
rs753767488 Chr.17: 73201151-73201186 on Build GRCh38
rs567335371 Chr.17: 73201137-73201137 on Build GRCh38
rs1026130 Chr.17: 73201061-73201061 on Build GRCh38
rs899721252 Chr.17: 73201125-73201125 on Build GRCh38
rs1039650034 Chr.17: 73201112-73201112 on Build GRCh38
rs770179791 Chr.17: 73201062-73201062 on Build GRCh38
rs746636606 Chr.17: 73201216-73201216 on Build GRCh38
rs35401348 Chr.17: 73201221-73201222 on Build GRCh38
rs776615420 Chr.17: 73201130-73201130 on Build GRCh38
rs886053369 Chr.17: 73201145-73201145 on Build GRCh38
rs773727583 Chr.17: 73201102-73201102 on Build GRCh38

View Full Product Details