Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133328776-133329037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALY
Entrez Gene ID
50632
Gene Name
calcyon neuron specific vesicular protein
Gene Aliases
DRD1IP, NSG3
UniGene
Hs.148680
Gene Chromosome Location
Chr.10: 133325423-133336983 on Build GRCh38
Transcripts
NM_001321617.1 NM_015722.3 XM_005252685.4 XM_011539836.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359332 c.20G>A p.S7N Chr.10: 133328970-133328970 on Build GRCh38
6518458 c.73G>T p.D25Y Chr.10: 133328917-133328917 on Build GRCh38
4593471 c.113T>C p.L38P Chr.10: 133328877-133328877 on Build GRCh38
6395132 c.34C>T p.P12S Chr.10: 133328956-133328956 on Build GRCh38
4481332 c.24C>T p.F8F Chr.10: 133328966-133328966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866303467 Chr.10: 133328970-133328970 on Build GRCh38
rs146132141 Chr.10: 133328957-133328957 on Build GRCh38
rs917041117 Chr.10: 133328807-133328807 on Build GRCh38
rs771848704 Chr.10: 133328956-133328956 on Build GRCh38
rs187662105 Chr.10: 133328871-133328871 on Build GRCh38
rs139888672 Chr.10: 133328923-133328923 on Build GRCh38
rs139948953 Chr.10: 133328961-133328961 on Build GRCh38
rs764580709 Chr.10: 133328836-133328836 on Build GRCh38
rs767252678 Chr.10: 133328813-133328813 on Build GRCh38
rs963953728 Chr.10: 133328990-133328990 on Build GRCh38
rs748798784 Chr.10: 133328929-133328929 on Build GRCh38
rs182655149 Chr.10: 133328815-133328815 on Build GRCh38
rs200316803 Chr.10: 133328863-133328863 on Build GRCh38
rs775747454 Chr.10: 133328989-133328989 on Build GRCh38
rs375019333 Chr.10: 133328837-133328837 on Build GRCh38
rs757620477 Chr.10: 133328864-133328864 on Build GRCh38
rs780762993 Chr.10: 133328870-133328870 on Build GRCh38
rs748142791 Chr.10: 133328953-133328953 on Build GRCh38
rs765391462 Chr.10: 133328964-133328964 on Build GRCh38
rs1010540981 Chr.10: 133328966-133328966 on Build GRCh38
rs765318658 Chr.10: 133328828-133328828 on Build GRCh38
rs922688664 Chr.10: 133328803-133328803 on Build GRCh38
rs372078376 Chr.10: 133328920-133328920 on Build GRCh38
rs145969782 Chr.10: 133328908-133328908 on Build GRCh38
rs201476239 Chr.10: 133328832-133328832 on Build GRCh38
rs1018613874 Chr.10: 133328977-133328977 on Build GRCh38
rs542453532 Chr.10: 133328939-133328939 on Build GRCh38
rs142942870 Chr.10: 133328933-133328933 on Build GRCh38
rs376926790 Chr.10: 133328995-133328995 on Build GRCh38
rs949826999 Chr.10: 133328808-133328808 on Build GRCh38
rs1014839980 Chr.10: 133328845-133328845 on Build GRCh38
rs747094733 Chr.10: 133328976-133328976 on Build GRCh38
rs1056464610 Chr.10: 133328833-133328833 on Build GRCh38
rs41283333 Chr.10: 133328816-133328816 on Build GRCh38
rs752542242 Chr.10: 133328827-133328839 on Build GRCh38
rs759696777 Chr.10: 133328817-133328817 on Build GRCh38
rs758844712 Chr.10: 133328834-133328834 on Build GRCh38
rs930100181 Chr.10: 133328810-133328810 on Build GRCh38

View Full Product Details