Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 168237225-168237484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX60
Entrez Gene ID
55601
Gene Name
DEXD/H-box helicase 60
Gene Aliases
-
UniGene
Hs.591710
Gene Chromosome Location
Chr.4: 168216291-168318807 on Build GRCh38
Transcripts
NM_017631.5 XM_011532103.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4839961 c.4336G>T p.E1446* Chr.4: 168237361-168237361 on Build GRCh38
3601833 c.4301G>A p.G1434E Chr.4: 168237396-168237396 on Build GRCh38
3601831 c.4343C>T p.S1448F Chr.4: 168237354-168237354 on Build GRCh38
4420249 c.4294C>T p.P1432S Chr.4: 168237403-168237403 on Build GRCh38
5682531 c.4301G>C p.G1434A Chr.4: 168237396-168237396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3805396 Chr.4: 168237380-168237380 on Build GRCh38
rs750482016 Chr.4: 168237290-168237290 on Build GRCh38
rs764802195 Chr.4: 168237259-168237260 on Build GRCh38
rs781493292 Chr.4: 168237419-168237419 on Build GRCh38
rs1055023729 Chr.4: 168237299-168237299 on Build GRCh38
rs867583844 Chr.4: 168237391-168237391 on Build GRCh38
rs6552559 Chr.4: 168237320-168237320 on Build GRCh38
rs3828560 Chr.4: 168237338-168237338 on Build GRCh38
rs901667272 Chr.4: 168237255-168237255 on Build GRCh38
rs538006739 Chr.4: 168237336-168237336 on Build GRCh38
rs371285949 Chr.4: 168237263-168237263 on Build GRCh38
rs755906629 Chr.4: 168237314-168237314 on Build GRCh38
rs747118964 Chr.4: 168237330-168237330 on Build GRCh38
rs774843415 Chr.4: 168237446-168237446 on Build GRCh38
rs755116660 Chr.4: 168237418-168237418 on Build GRCh38
rs773671007 Chr.4: 168237358-168237358 on Build GRCh38
rs375550836 Chr.4: 168237288-168237288 on Build GRCh38
rs749683161 Chr.4: 168237439-168237439 on Build GRCh38
rs763335214 Chr.4: 168237364-168237364 on Build GRCh38
rs765781389 Chr.4: 168237267-168237267 on Build GRCh38
rs753708323 Chr.4: 168237403-168237403 on Build GRCh38
rs369165821 Chr.4: 168237261-168237261 on Build GRCh38
rs996036276 Chr.4: 168237259-168237259 on Build GRCh38
rs760378001 Chr.4: 168237390-168237390 on Build GRCh38
rs200182828 Chr.4: 168237287-168237287 on Build GRCh38
rs761382805 Chr.4: 168237260-168237261 on Build GRCh38
rs771497232 Chr.4: 168237366-168237366 on Build GRCh38
rs770036866 Chr.4: 168237357-168237357 on Build GRCh38
rs756395961 Chr.4: 168237427-168237427 on Build GRCh38
rs765245036 Chr.4: 168237407-168237407 on Build GRCh38
rs781581408 Chr.4: 168237337-168237337 on Build GRCh38
rs75558995 Chr.4: 168237436-168237436 on Build GRCh38
rs751118344 Chr.4: 168237281-168237281 on Build GRCh38
rs777691714 Chr.4: 168237316-168237316 on Build GRCh38
rs771283821 Chr.4: 168237445-168237445 on Build GRCh38
rs145749472 Chr.4: 168237324-168237324 on Build GRCh38
rs370496559 Chr.4: 168237415-168237415 on Build GRCh38
rs763775908 Chr.4: 168237396-168237396 on Build GRCh38
rs139699246 Chr.4: 168237319-168237319 on Build GRCh38
rs374886685 Chr.4: 168237369-168237369 on Build GRCh38
rs998333419 Chr.4: 168237345-168237345 on Build GRCh38
rs752430979 Chr.4: 168237310-168237310 on Build GRCh38
rs190525669 Chr.4: 168237423-168237423 on Build GRCh38
rs367825763 Chr.4: 168237382-168237382 on Build GRCh38

View Full Product Details